Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169518262-169518634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
UniGene
Hs.30054
Gene Chromosome Location
Chr.1: 169511954-169586632 on Build GRCh38
Transcripts
NM_000130.4 XM_017000660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899254 c.6264G>T p.K2088N Chr.1: 169518493-169518493 on Build GRCh38
6657763 c.6294C>T p.P2098P Chr.1: 169518463-169518463 on Build GRCh38
899253 c.6344A>G p.K2115R Chr.1: 169518413-169518413 on Build GRCh38
3418298 c.6264G>A p.K2088K Chr.1: 169518493-169518493 on Build GRCh38
6059170 c.6280G>C p.D2094H Chr.1: 169518477-169518477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751215852 Chr.1: 169518542-169518542 on Build GRCh38
rs776601500 Chr.1: 169518355-169518355 on Build GRCh38
rs951637458 Chr.1: 169518302-169518302 on Build GRCh38
rs767355058 Chr.1: 169518393-169518393 on Build GRCh38
rs759579691 Chr.1: 169518349-169518349 on Build GRCh38
rs760084344 Chr.1: 169518453-169518453 on Build GRCh38
rs764974779 Chr.1: 169518397-169518397 on Build GRCh38
rs145706683 Chr.1: 169518602-169518602 on Build GRCh38
rs964572005 Chr.1: 169518343-169518343 on Build GRCh38
rs772248818 Chr.1: 169518458-169518458 on Build GRCh38
rs377357737 Chr.1: 169518399-169518399 on Build GRCh38
rs373540139 Chr.1: 169518364-169518364 on Build GRCh38
rs759698433 Chr.1: 169518362-169518362 on Build GRCh38
rs958968876 Chr.1: 169518337-169518337 on Build GRCh38
rs757163642 Chr.1: 169518610-169518610 on Build GRCh38
rs762791810 Chr.1: 169518520-169518520 on Build GRCh38
rs764184897 Chr.1: 169518389-169518389 on Build GRCh38
rs879214285 Chr.1: 169518416-169518416 on Build GRCh38
rs886141992 Chr.1: 169518596-169518596 on Build GRCh38
rs746211961 Chr.1: 169518454-169518454 on Build GRCh38
rs201709883 Chr.1: 169518498-169518498 on Build GRCh38
rs987302774 Chr.1: 169518438-169518438 on Build GRCh38
rs991184224 Chr.1: 169518359-169518359 on Build GRCh38
rs370754868 Chr.1: 169518407-169518407 on Build GRCh38
rs779469373 Chr.1: 169518582-169518582 on Build GRCh38
rs775024856 Chr.1: 169518387-169518404 on Build GRCh38
rs749823356 Chr.1: 169518443-169518443 on Build GRCh38
rs35369423 Chr.1: 169518448-169518448 on Build GRCh38
rs112171464 Chr.1: 169518463-169518463 on Build GRCh38
rs779729300 Chr.1: 169518404-169518404 on Build GRCh38
rs560451001 Chr.1: 169518348-169518348 on Build GRCh38
rs267598156 Chr.1: 169518430-169518430 on Build GRCh38
rs6013 Chr.1: 169518583-169518583 on Build GRCh38
rs551194028 Chr.1: 169518598-169518598 on Build GRCh38
rs780611660 Chr.1: 169518437-169518437 on Build GRCh38
rs201142225 Chr.1: 169518516-169518516 on Build GRCh38
rs527837316 Chr.1: 169518504-169518504 on Build GRCh38
rs775843029 Chr.1: 169518459-169518459 on Build GRCh38
rs369317367 Chr.1: 169518560-169518560 on Build GRCh38
rs200646894 Chr.1: 169518435-169518435 on Build GRCh38
rs758039101 Chr.1: 169518403-169518403 on Build GRCh38
rs775340722 Chr.1: 169518412-169518412 on Build GRCh38
rs118203910 Chr.1: 169518453-169518453 on Build GRCh38
rs564167068 Chr.1: 169518552-169518552 on Build GRCh38
rs375632547 Chr.1: 169518383-169518383 on Build GRCh38
rs747170145 Chr.1: 169518472-169518472 on Build GRCh38
rs887173832 Chr.1: 169518557-169518557 on Build GRCh38
rs148045855 Chr.1: 169518541-169518541 on Build GRCh38
rs376834131 Chr.1: 169518561-169518561 on Build GRCh38
rs200435082 Chr.1: 169518499-169518499 on Build GRCh38
rs772316521 Chr.1: 169518367-169518367 on Build GRCh38
rs202227122 Chr.1: 169518449-169518449 on Build GRCh38
rs761745537 Chr.1: 169518396-169518396 on Build GRCh38
rs751124066 Chr.1: 169518434-169518434 on Build GRCh38
rs768862607 Chr.1: 169518477-169518477 on Build GRCh38
rs1036373668 Chr.1: 169518322-169518322 on Build GRCh38
rs752228138 Chr.1: 169518573-169518573 on Build GRCh38
rs1003012822 Chr.1: 169518318-169518318 on Build GRCh38
rs183596349 Chr.1: 169518381-169518381 on Build GRCh38
rs765202347 Chr.1: 169518503-169518503 on Build GRCh38

View Full Product Details