Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17734681-17735260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BID
Entrez Gene ID
637
Gene Name
BH3 interacting domain death agonist
Gene Aliases
FP497
UniGene
Hs.517145
Gene Chromosome Location
Chr.22: 17734140-17774665 on Build GRCh38
Transcripts
NM_001196.3 NM_001244567.1 NM_001244569.1 NM_001244570.1 NM_001244572.1 NM_197966.2 NM_197967.2

SNP Detail

SNP ID Chromosome Location
rs566559199 Chr.22: 17735088-17735088 on Build GRCh38
rs149980557 Chr.22: 17734867-17734867 on Build GRCh38
rs140522332 Chr.22: 17734736-17734736 on Build GRCh38
rs544801192 Chr.22: 17735163-17735163 on Build GRCh38
rs775548793 Chr.22: 17735036-17735037 on Build GRCh38
rs878903696 Chr.22: 17734901-17734901 on Build GRCh38
rs974594453 Chr.22: 17734975-17734975 on Build GRCh38
rs776421618 Chr.22: 17735201-17735202 on Build GRCh38
rs567224393 Chr.22: 17734939-17734939 on Build GRCh38
rs550824939 Chr.22: 17734983-17734984 on Build GRCh38
rs868546130 Chr.22: 17735132-17735132 on Build GRCh38
rs767247848 Chr.22: 17734986-17734986 on Build GRCh38
rs924617168 Chr.22: 17734748-17734748 on Build GRCh38
rs934663245 Chr.22: 17735077-17735077 on Build GRCh38
rs189629146 Chr.22: 17734847-17734847 on Build GRCh38
rs559033635 Chr.22: 17735143-17735143 on Build GRCh38
rs762735083 Chr.22: 17734790-17734790 on Build GRCh38
rs955417854 Chr.22: 17734990-17734990 on Build GRCh38
rs972884127 Chr.22: 17735120-17735120 on Build GRCh38
rs982241307 Chr.22: 17734761-17734761 on Build GRCh38
rs765460407 Chr.22: 17735131-17735131 on Build GRCh38
rs934882557 Chr.22: 17734749-17734749 on Build GRCh38
rs1044551485 Chr.22: 17735219-17735219 on Build GRCh38
rs934111724 Chr.22: 17734723-17734723 on Build GRCh38
rs1032258315 Chr.22: 17734732-17734732 on Build GRCh38
rs754320600 Chr.22: 17735090-17735090 on Build GRCh38
rs201491655 Chr.22: 17735202-17735203 on Build GRCh38
rs931532156 Chr.22: 17734831-17734832 on Build GRCh38
rs144073937 Chr.22: 17735077-17735078 on Build GRCh38
rs773681981 Chr.22: 17734917-17734917 on Build GRCh38
rs1015088366 Chr.22: 17735073-17735073 on Build GRCh38
rs184390244 Chr.22: 17734743-17734743 on Build GRCh38
rs146895750 Chr.22: 17735202-17735202 on Build GRCh38
rs772178520 Chr.22: 17734970-17734970 on Build GRCh38
rs931584832 Chr.22: 17735159-17735159 on Build GRCh38
rs928849703 Chr.22: 17734775-17734775 on Build GRCh38
rs990577606 Chr.22: 17735097-17735097 on Build GRCh38
rs553335102 Chr.22: 17734803-17734803 on Build GRCh38
rs8190357 Chr.22: 17735031-17735032 on Build GRCh38
rs368957466 Chr.22: 17735176-17735176 on Build GRCh38
rs867495953 Chr.22: 17734801-17734801 on Build GRCh38
rs536191527 Chr.22: 17734947-17734947 on Build GRCh38
rs981909996 Chr.22: 17735050-17735050 on Build GRCh38
rs1054725467 Chr.22: 17734756-17734756 on Build GRCh38
rs986863151 Chr.22: 17735004-17735004 on Build GRCh38
rs907374422 Chr.22: 17735226-17735226 on Build GRCh38
rs1012201043 Chr.22: 17735219-17735220 on Build GRCh38
rs8190356 Chr.22: 17735067-17735068 on Build GRCh38
rs957723018 Chr.22: 17734733-17734733 on Build GRCh38
rs1053636037 Chr.22: 17735203-17735203 on Build GRCh38
rs575617011 Chr.22: 17735158-17735158 on Build GRCh38
rs893405205 Chr.22: 17734757-17734757 on Build GRCh38
rs555207372 Chr.22: 17735005-17735005 on Build GRCh38
rs1008339916 Chr.22: 17734894-17734894 on Build GRCh38
rs948895442 Chr.22: 17734759-17734759 on Build GRCh38
rs575412458 Chr.22: 17735211-17735212 on Build GRCh38
rs573203205 Chr.22: 17734851-17734852 on Build GRCh38
rs1023318068 Chr.22: 17735008-17735008 on Build GRCh38
rs147656401 Chr.22: 17734924-17734924 on Build GRCh38
rs886125448 Chr.22: 17734885-17734885 on Build GRCh38
rs920055440 Chr.22: 17735126-17735126 on Build GRCh38
rs771123079 Chr.22: 17734726-17734727 on Build GRCh38
rs8190355 Chr.22: 17735118-17735118 on Build GRCh38
rs893728437 Chr.22: 17734994-17734994 on Build GRCh38
rs536211373 Chr.22: 17734776-17734776 on Build GRCh38
rs569215031 Chr.22: 17735031-17735031 on Build GRCh38
rs552836548 Chr.22: 17735074-17735074 on Build GRCh38
rs1016169829 Chr.22: 17734938-17734938 on Build GRCh38
rs759313178 Chr.22: 17734970-17734970 on Build GRCh38

View Full Product Details