Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58937406-58937962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
-
UniGene
Hs.368434
Gene Chromosome Location
Chr.3: 58716415-59050078 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533461.1 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Gene Symbol
C3orf67-AS1
Entrez Gene ID
101929238
Gene Name
C3orf67 antisense RNA 1
Gene Aliases
-
UniGene
Hs.683953
Gene Chromosome Location
Chr.3: 58824471-59019093 on Build GRCh38
Transcripts
NM_198463.2 XM_005264929.2 XM_006713013.3 XM_011533451.2 XM_011533453.2 XM_011533455.2 XM_011533456.1 XM_011533457.1 XM_011533458.2 XM_011533459.2 XM_011533461.1 XM_017005848.1 XM_017005849.1 XM_017005850.1 XM_017005851.1 NR_110820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
480515 c.17T>G p.I6S Chr.3: 58937649-58937649 on Build GRCh38
6616053 c.10C>T p.R4C Chr.3: 58937656-58937656 on Build GRCh38
4968708 c.16A>C p.I6L Chr.3: 58937650-58937650 on Build GRCh38
6300137 c.12T>C p.R4R Chr.3: 58937654-58937654 on Build GRCh38
6300136 c.12T>C p.R4R Chr.3: 58937654-58937654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs60853394 Chr.3: 58937489-58937489 on Build GRCh38
rs757899307 Chr.3: 58937764-58937764 on Build GRCh38
rs747932906 Chr.3: 58937795-58937795 on Build GRCh38
rs765063239 Chr.3: 58937665-58937665 on Build GRCh38
rs771766526 Chr.3: 58937797-58937797 on Build GRCh38
rs930407707 Chr.3: 58937473-58937473 on Build GRCh38
rs770922449 Chr.3: 58937716-58937716 on Build GRCh38
rs569684184 Chr.3: 58937472-58937472 on Build GRCh38
rs770207975 Chr.3: 58937721-58937721 on Build GRCh38
rs867198925 Chr.3: 58937462-58937462 on Build GRCh38
rs776174601 Chr.3: 58937732-58937732 on Build GRCh38
rs907615511 Chr.3: 58937748-58937748 on Build GRCh38
rs773140937 Chr.3: 58937799-58937799 on Build GRCh38
rs563555694 Chr.3: 58937731-58937731 on Build GRCh38
rs768588266 Chr.3: 58937627-58937627 on Build GRCh38
rs781507784 Chr.3: 58937717-58937717 on Build GRCh38
rs760132330 Chr.3: 58937442-58937442 on Build GRCh38
rs771996035 Chr.3: 58937726-58937726 on Build GRCh38
rs941694474 Chr.3: 58937565-58937565 on Build GRCh38
rs1042468487 Chr.3: 58937707-58937707 on Build GRCh38
rs990926576 Chr.3: 58937523-58937523 on Build GRCh38
rs540865500 Chr.3: 58937433-58937433 on Build GRCh38
rs753490536 Chr.3: 58937664-58937664 on Build GRCh38
rs369007424 Chr.3: 58937589-58937589 on Build GRCh38
rs542740822 Chr.3: 58937772-58937772 on Build GRCh38
rs749927587 Chr.3: 58937654-58937654 on Build GRCh38
rs761806884 Chr.3: 58937647-58937647 on Build GRCh38
rs550930994 Chr.3: 58937869-58937869 on Build GRCh38
rs773360252 Chr.3: 58937638-58937638 on Build GRCh38
rs1008724624 Chr.3: 58937524-58937524 on Build GRCh38
rs546831138 Chr.3: 58937807-58937807 on Build GRCh38
rs753124047 Chr.3: 58937684-58937684 on Build GRCh38
rs532334424 Chr.3: 58937776-58937776 on Build GRCh38
rs781417589 Chr.3: 58937765-58937765 on Build GRCh38
rs970916376 Chr.3: 58937785-58937785 on Build GRCh38
rs907570632 Chr.3: 58937562-58937562 on Build GRCh38
rs760123795 Chr.3: 58937655-58937655 on Build GRCh38
rs756381323 Chr.3: 58937774-58937774 on Build GRCh38
rs944416442 Chr.3: 58937753-58937753 on Build GRCh38
rs762098933 Chr.3: 58937459-58937459 on Build GRCh38
rs141165549 Chr.3: 58937666-58937666 on Build GRCh38
rs757371530 Chr.3: 58937874-58937874 on Build GRCh38
rs555503215 Chr.3: 58937525-58937525 on Build GRCh38
rs776084777 Chr.3: 58937812-58937812 on Build GRCh38
rs1032522256 Chr.3: 58937494-58937494 on Build GRCh38
rs368902952 Chr.3: 58937437-58937437 on Build GRCh38
rs532667701 Chr.3: 58937859-58937859 on Build GRCh38
rs752021547 Chr.3: 58937761-58937761 on Build GRCh38
rs145528529 Chr.3: 58937800-58937800 on Build GRCh38
rs753060406 Chr.3: 58937741-58937741 on Build GRCh38
rs900380201 Chr.3: 58937583-58937583 on Build GRCh38
rs758215193 Chr.3: 58937729-58937729 on Build GRCh38
rs553296082 Chr.3: 58937587-58937587 on Build GRCh38
rs764905876 Chr.3: 58937739-58937739 on Build GRCh38
rs375106645 Chr.3: 58937613-58937613 on Build GRCh38
rs776521908 Chr.3: 58937522-58937522 on Build GRCh38
rs1000043831 Chr.3: 58937558-58937558 on Build GRCh38
rs746077209 Chr.3: 58937718-58937718 on Build GRCh38
rs779231705 Chr.3: 58937679-58937679 on Build GRCh38
rs756364851 Chr.3: 58937613-58937614 on Build GRCh38
rs916705673 Chr.3: 58937605-58937605 on Build GRCh38
rs879193675 Chr.3: 58937497-58937497 on Build GRCh38
rs760163592 Chr.3: 58937730-58937730 on Build GRCh38
rs375862184 Chr.3: 58937569-58937569 on Build GRCh38
rs190915720 Chr.3: 58937501-58937501 on Build GRCh38
rs765907486 Chr.3: 58937484-58937484 on Build GRCh38
rs781462446 Chr.3: 58937902-58937902 on Build GRCh38
rs761560449 Chr.3: 58937723-58937723 on Build GRCh38
rs369534957 Chr.3: 58937652-58937652 on Build GRCh38
rs996060692 Chr.3: 58937541-58937541 on Build GRCh38
rs545378264 Chr.3: 58937656-58937656 on Build GRCh38
rs1040240120 Chr.3: 58937755-58937755 on Build GRCh38
rs751851846 Chr.3: 58937851-58937851 on Build GRCh38
rs572814311 Chr.3: 58937634-58937634 on Build GRCh38
rs890281294 Chr.3: 58937671-58937671 on Build GRCh38
rs531000950 Chr.3: 58937756-58937756 on Build GRCh38
rs900424025 Chr.3: 58937536-58937536 on Build GRCh38
rs1022320475 Chr.3: 58937552-58937552 on Build GRCh38
rs1031541548 Chr.3: 58937858-58937858 on Build GRCh38
rs865808935 Chr.3: 58937712-58937712 on Build GRCh38
rs573736631 Chr.3: 58937561-58937561 on Build GRCh38
rs758919936 Chr.3: 58937685-58937685 on Build GRCh38
rs565197134 Chr.3: 58937837-58937837 on Build GRCh38
rs752497716 Chr.3: 58937695-58937695 on Build GRCh38
rs968019864 Chr.3: 58937554-58937554 on Build GRCh38
rs202214979 Chr.3: 58937693-58937693 on Build GRCh38

View Full Product Details