Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 9119161-9119650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf72
Entrez Gene ID
29035
Gene Name
chromosome 16 open reading frame 72
Gene Aliases
PRO0149
UniGene
Hs.221497
Gene Chromosome Location
Chr.16: 9091329-9119698 on Build GRCh38
Transcripts
NM_014117.2

SNP Detail

SNP ID Chromosome Location
rs1016868051 Chr.16: 9119533-9119533 on Build GRCh38
rs559365255 Chr.16: 9119285-9119285 on Build GRCh38
rs762148189 Chr.16: 9119261-9119261 on Build GRCh38
rs940421343 Chr.16: 9119203-9119203 on Build GRCh38
rs973065276 Chr.16: 9119455-9119455 on Build GRCh38
rs992721721 Chr.16: 9119431-9119431 on Build GRCh38
rs115683539 Chr.16: 9119340-9119340 on Build GRCh38
rs1003153891 Chr.16: 9119210-9119210 on Build GRCh38
rs376662525 Chr.16: 9119613-9119613 on Build GRCh38
rs778854307 Chr.16: 9119581-9119581 on Build GRCh38
rs1043097632 Chr.16: 9119529-9119529 on Build GRCh38
rs1041046108 Chr.16: 9119269-9119269 on Build GRCh38
rs6777 Chr.16: 9119562-9119562 on Build GRCh38
rs1001767363 Chr.16: 9119396-9119396 on Build GRCh38
rs1050315602 Chr.16: 9119486-9119486 on Build GRCh38
rs79581216 Chr.16: 9119445-9119445 on Build GRCh38
rs766430703 Chr.16: 9119454-9119454 on Build GRCh38
rs907518675 Chr.16: 9119446-9119446 on Build GRCh38
rs763768693 Chr.16: 9119241-9119241 on Build GRCh38
rs954669859 Chr.16: 9119573-9119573 on Build GRCh38
rs958836021 Chr.16: 9119249-9119249 on Build GRCh38
rs946029537 Chr.16: 9119523-9119523 on Build GRCh38
rs545306663 Chr.16: 9119185-9119185 on Build GRCh38
rs942505282 Chr.16: 9119438-9119438 on Build GRCh38
rs750938757 Chr.16: 9119324-9119324 on Build GRCh38
rs947146922 Chr.16: 9119449-9119449 on Build GRCh38
rs890025167 Chr.16: 9119251-9119251 on Build GRCh38
rs370030244 Chr.16: 9119182-9119182 on Build GRCh38
rs551653442 Chr.16: 9119423-9119423 on Build GRCh38
rs919281036 Chr.16: 9119388-9119388 on Build GRCh38
rs920386081 Chr.16: 9119247-9119247 on Build GRCh38
rs563616182 Chr.16: 9119186-9119186 on Build GRCh38
rs775026118 Chr.16: 9119618-9119618 on Build GRCh38
rs1013078351 Chr.16: 9119514-9119514 on Build GRCh38
rs760327299 Chr.16: 9119200-9119200 on Build GRCh38
rs187201907 Chr.16: 9119365-9119365 on Build GRCh38
rs776306519 Chr.16: 9119250-9119250 on Build GRCh38
rs771927674 Chr.16: 9119616-9119616 on Build GRCh38
rs931781860 Chr.16: 9119482-9119482 on Build GRCh38
rs530729674 Chr.16: 9119196-9119196 on Build GRCh38
rs753460071 Chr.16: 9119349-9119349 on Build GRCh38
rs993959400 Chr.16: 9119552-9119552 on Build GRCh38
rs911414620 Chr.16: 9119488-9119488 on Build GRCh38
rs1057125697 Chr.16: 9119319-9119319 on Build GRCh38
rs912484220 Chr.16: 9119425-9119425 on Build GRCh38
rs781538345 Chr.16: 9119530-9119530 on Build GRCh38
rs542411259 Chr.16: 9119248-9119248 on Build GRCh38
rs189111738 Chr.16: 9119184-9119184 on Build GRCh38
rs561148113 Chr.16: 9119280-9119280 on Build GRCh38
rs532018680 Chr.16: 9119422-9119422 on Build GRCh38
rs904573396 Chr.16: 9119536-9119536 on Build GRCh38
rs902823836 Chr.16: 9119473-9119473 on Build GRCh38
rs942557944 Chr.16: 9119183-9119183 on Build GRCh38
rs373227496 Chr.16: 9119300-9119300 on Build GRCh38
rs7188669 Chr.16: 9119584-9119584 on Build GRCh38
rs377083587 Chr.16: 9119475-9119475 on Build GRCh38
rs748437472 Chr.16: 9119539-9119539 on Build GRCh38
rs1041307384 Chr.16: 9119499-9119499 on Build GRCh38
rs755145180 Chr.16: 9119513-9119513 on Build GRCh38
rs987439253 Chr.16: 9119576-9119576 on Build GRCh38
rs961972795 Chr.16: 9119598-9119598 on Build GRCh38
rs181605820 Chr.16: 9119284-9119284 on Build GRCh38
rs944245502 Chr.16: 9119495-9119495 on Build GRCh38
rs537755967 Chr.16: 9119457-9119457 on Build GRCh38
rs751619984 Chr.16: 9119474-9119474 on Build GRCh38
rs1037424925 Chr.16: 9119240-9119240 on Build GRCh38
rs567870959 Chr.16: 9119567-9119567 on Build GRCh38

View Full Product Details