Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 116092368-116092967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD10
Entrez Gene ID
51397
Gene Name
COMM domain containing 10
Gene Aliases
PTD002
UniGene
Hs.483136
Gene Chromosome Location
Chr.5: 116084592-116446462 on Build GRCh38
Transcripts
NM_001308080.1 NM_016144.3 XM_011543458.2 XM_011543460.2 XM_017009553.1 XM_017009555.1 XM_017009556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
260255 c.322G>T p.A108S Chr.5: 116092623-116092623 on Build GRCh38
4449824 c.344_347delCTAT p.S115fs*19 Chr.5: 116092645-116092648 on Build GRCh38
149901 c.336G>T p.T112T Chr.5: 116092637-116092637 on Build GRCh38
4141384 c.387G>T p.L129L Chr.5: 116092688-116092688 on Build GRCh38
4428581 c.361G>A p.V121I Chr.5: 116092662-116092662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760914074 Chr.5: 116092660-116092660 on Build GRCh38
rs192970063 Chr.5: 116092428-116092428 on Build GRCh38
rs1018936426 Chr.5: 116092876-116092876 on Build GRCh38
rs891296572 Chr.5: 116092860-116092860 on Build GRCh38
rs930018457 Chr.5: 116092882-116092882 on Build GRCh38
rs745355043 Chr.5: 116092704-116092704 on Build GRCh38
rs952583778 Chr.5: 116092484-116092484 on Build GRCh38
rs151325955 Chr.5: 116092606-116092606 on Build GRCh38
rs768504981 Chr.5: 116092722-116092722 on Build GRCh38
rs775191934 Chr.5: 116092647-116092647 on Build GRCh38
rs771732405 Chr.5: 116092501-116092501 on Build GRCh38
rs769701802 Chr.5: 116092641-116092641 on Build GRCh38
rs753094653 Chr.5: 116092675-116092675 on Build GRCh38
rs780732647 Chr.5: 116092701-116092701 on Build GRCh38
rs930075535 Chr.5: 116092832-116092832 on Build GRCh38
rs956637705 Chr.5: 116092776-116092776 on Build GRCh38
rs746739074 Chr.5: 116092691-116092691 on Build GRCh38
rs957078185 Chr.5: 116092938-116092938 on Build GRCh38
rs1025621738 Chr.5: 116092689-116092689 on Build GRCh38
rs763549331 Chr.5: 116092709-116092710 on Build GRCh38
rs201107215 Chr.5: 116092632-116092632 on Build GRCh38
rs777217887 Chr.5: 116092655-116092655 on Build GRCh38
rs981357728 Chr.5: 116092739-116092740 on Build GRCh38
rs111935686 Chr.5: 116092773-116092773 on Build GRCh38
rs776950650 Chr.5: 116092734-116092734 on Build GRCh38
rs550973441 Chr.5: 116092570-116092570 on Build GRCh38
rs200313896 Chr.5: 116092712-116092712 on Build GRCh38
rs10069656 Chr.5: 116092747-116092747 on Build GRCh38
rs978974126 Chr.5: 116092854-116092854 on Build GRCh38
rs10478276 Chr.5: 116092480-116092480 on Build GRCh38
rs1032709204 Chr.5: 116092925-116092925 on Build GRCh38
rs756886920 Chr.5: 116092695-116092695 on Build GRCh38
rs774013583 Chr.5: 116092724-116092724 on Build GRCh38
rs982384823 Chr.5: 116092759-116092759 on Build GRCh38
rs867116086 Chr.5: 116092855-116092855 on Build GRCh38
rs747812694 Chr.5: 116092499-116092499 on Build GRCh38
rs570575160 Chr.5: 116092844-116092844 on Build GRCh38
rs746888932 Chr.5: 116092565-116092565 on Build GRCh38
rs753944491 Chr.5: 116092670-116092670 on Build GRCh38
rs574085904 Chr.5: 116092673-116092673 on Build GRCh38
rs766510815 Chr.5: 116092583-116092583 on Build GRCh38
rs987914270 Chr.5: 116092784-116092784 on Build GRCh38
rs772694743 Chr.5: 116092721-116092732 on Build GRCh38
rs766633557 Chr.5: 116092665-116092665 on Build GRCh38
rs868050477 Chr.5: 116092525-116092525 on Build GRCh38
rs551280738 Chr.5: 116092881-116092881 on Build GRCh38
rs370201279 Chr.5: 116092792-116092792 on Build GRCh38
rs769156596 Chr.5: 116092495-116092495 on Build GRCh38
rs148059527 Chr.5: 116092756-116092756 on Build GRCh38
rs537786572 Chr.5: 116092556-116092556 on Build GRCh38
rs766948867 Chr.5: 116092711-116092712 on Build GRCh38
rs905875238 Chr.5: 116092425-116092425 on Build GRCh38
rs1041585805 Chr.5: 116092880-116092880 on Build GRCh38
rs756285400 Chr.5: 116092573-116092573 on Build GRCh38
rs779355071 Chr.5: 116092498-116092498 on Build GRCh38
rs1047195725 Chr.5: 116092839-116092839 on Build GRCh38
rs745867369 Chr.5: 116092640-116092640 on Build GRCh38
rs1129495 Chr.5: 116092684-116092684 on Build GRCh38
rs775812234 Chr.5: 116092706-116092707 on Build GRCh38
rs969213089 Chr.5: 116092518-116092518 on Build GRCh38
rs141808376 Chr.5: 116092932-116092932 on Build GRCh38
rs996227642 Chr.5: 116092897-116092897 on Build GRCh38
rs759398485 Chr.5: 116092552-116092552 on Build GRCh38
rs914929373 Chr.5: 116092636-116092636 on Build GRCh38
rs770375919 Chr.5: 116092546-116092546 on Build GRCh38
rs957169526 Chr.5: 116092686-116092686 on Build GRCh38
rs760261793 Chr.5: 116092516-116092516 on Build GRCh38
rs760243106 Chr.5: 116092726-116092727 on Build GRCh38
rs772636021 Chr.5: 116092515-116092515 on Build GRCh38
rs746262442 Chr.5: 116092799-116092799 on Build GRCh38
rs758800834 Chr.5: 116092681-116092681 on Build GRCh38
rs77798383 Chr.5: 116092716-116092716 on Build GRCh38
rs772515888 Chr.5: 116092803-116092803 on Build GRCh38
rs755528213 Chr.5: 116092605-116092605 on Build GRCh38
rs1129494 Chr.5: 116092637-116092637 on Build GRCh38
rs746582688 Chr.5: 116092618-116092618 on Build GRCh38
rs530792697 Chr.5: 116092849-116092849 on Build GRCh38
rs375816472 Chr.5: 116092615-116092615 on Build GRCh38
rs200380253 Chr.5: 116092663-116092663 on Build GRCh38
rs766676570 Chr.5: 116092730-116092730 on Build GRCh38
rs571057040 Chr.5: 116092596-116092596 on Build GRCh38
rs568460246 Chr.5: 116092483-116092483 on Build GRCh38
rs984051981 Chr.5: 116092493-116092493 on Build GRCh38
rs769257187 Chr.5: 116092708-116092708 on Build GRCh38
rs558248470 Chr.5: 116092608-116092608 on Build GRCh38
rs942863943 Chr.5: 116092867-116092867 on Build GRCh38
rs548689433 Chr.5: 116092768-116092768 on Build GRCh38
rs765803405 Chr.5: 116092674-116092674 on Build GRCh38
rs1008401275 Chr.5: 116092862-116092862 on Build GRCh38
rs965399054 Chr.5: 116092479-116092479 on Build GRCh38
rs140746680 Chr.5: 116092613-116092613 on Build GRCh38
rs10478275 Chr.5: 116092399-116092399 on Build GRCh38
rs6894046 Chr.5: 116092742-116092742 on Build GRCh38
rs898555827 Chr.5: 116092823-116092823 on Build GRCh38

View Full Product Details