Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140847966-140848560 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Gene Symbol
PCDHA5
Entrez Gene ID
56143
Gene Name
protocadherin alpha 5
Gene Aliases
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140821776-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Gene Symbol
PCDHA6
Entrez Gene ID
56142
Gene Name
protocadherin alpha 6
Gene Aliases
CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140827922-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Gene Symbol
PCDHA7
Entrez Gene ID
56141
Gene Name
protocadherin alpha 7
Gene Aliases
CNR4, CNRN4, CNRS4, CRNR4, PCDH-ALPHA7
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140834384-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Gene Symbol
PCDHA8
Entrez Gene ID
56140
Gene Name
protocadherin alpha 8
Gene Aliases
PCDH-ALPHA8
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140841322-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Gene Symbol
PCDHA9
Entrez Gene ID
9752
Gene Name
protocadherin alpha 9
Gene Aliases
PCDH-ALPHA9
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140847772-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_014005.4 NM_031857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6011971 c.2T>A p.M1K Chr.5: 140848497-140848497 on Build GRCh38
3851061 c.17G>A p.R6Q Chr.5: 140848512-140848512 on Build GRCh38
1310650 c.28G>C p.E10Q Chr.5: 140848523-140848523 on Build GRCh38
6169227 c.63C>T p.L21L Chr.5: 140848558-140848558 on Build GRCh38
3851060 c.17G>A p.R6Q Chr.5: 140848512-140848512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973292437 Chr.5: 140848537-140848537 on Build GRCh38
rs782638492 Chr.5: 140848488-140848488 on Build GRCh38
rs373689042 Chr.5: 140848501-140848501 on Build GRCh38
rs183772404 Chr.5: 140848127-140848127 on Build GRCh38
rs782242681 Chr.5: 140848517-140848517 on Build GRCh38
rs954845616 Chr.5: 140848293-140848293 on Build GRCh38
rs534218988 Chr.5: 140848444-140848444 on Build GRCh38
rs370634862 Chr.5: 140848495-140848495 on Build GRCh38
rs781824936 Chr.5: 140848473-140848473 on Build GRCh38
rs116084528 Chr.5: 140848069-140848069 on Build GRCh38
rs964795646 Chr.5: 140848275-140848275 on Build GRCh38
rs916399183 Chr.5: 140848364-140848364 on Build GRCh38
rs749997840 Chr.5: 140848103-140848103 on Build GRCh38
rs114595741 Chr.5: 140848080-140848080 on Build GRCh38
rs566166825 Chr.5: 140848195-140848195 on Build GRCh38
rs558543350 Chr.5: 140848404-140848404 on Build GRCh38
rs782601054 Chr.5: 140848499-140848499 on Build GRCh38
rs1011742191 Chr.5: 140848200-140848200 on Build GRCh38
rs755444526 Chr.5: 140848316-140848316 on Build GRCh38
rs375489955 Chr.5: 140848285-140848285 on Build GRCh38
rs251352 Chr.5: 140848291-140848291 on Build GRCh38
rs782793064 Chr.5: 140848413-140848413 on Build GRCh38
rs188149099 Chr.5: 140848252-140848252 on Build GRCh38
rs782776989 Chr.5: 140848320-140848320 on Build GRCh38
rs547902835 Chr.5: 140848164-140848164 on Build GRCh38
rs1021743216 Chr.5: 140848253-140848253 on Build GRCh38
rs1003138626 Chr.5: 140848175-140848175 on Build GRCh38
rs782646562 Chr.5: 140848503-140848503 on Build GRCh38
rs4151686 Chr.5: 140848312-140848312 on Build GRCh38
rs941809355 Chr.5: 140848042-140848042 on Build GRCh38
rs1030799361 Chr.5: 140848457-140848457 on Build GRCh38
rs574135691 Chr.5: 140848024-140848024 on Build GRCh38
rs553949488 Chr.5: 140848521-140848521 on Build GRCh38
rs1052101954 Chr.5: 140848403-140848403 on Build GRCh38
rs781909881 Chr.5: 140848493-140848493 on Build GRCh38
rs781798725 Chr.5: 140848428-140848428 on Build GRCh38
rs983897735 Chr.5: 140848468-140848468 on Build GRCh38
rs906204704 Chr.5: 140848390-140848390 on Build GRCh38
rs963313279 Chr.5: 140848520-140848520 on Build GRCh38
rs536867910 Chr.5: 140848483-140848483 on Build GRCh38
rs782158413 Chr.5: 140848448-140848448 on Build GRCh38
rs782334908 Chr.5: 140848502-140848502 on Build GRCh38
rs782613962 Chr.5: 140848366-140848366 on Build GRCh38
rs181132008 Chr.5: 140848111-140848111 on Build GRCh38
rs781855358 Chr.5: 140848357-140848357 on Build GRCh38
rs115415885 Chr.5: 140848043-140848043 on Build GRCh38
rs569179390 Chr.5: 140848270-140848270 on Build GRCh38
rs4151688 Chr.5: 140848406-140848406 on Build GRCh38
rs891964710 Chr.5: 140848198-140848198 on Build GRCh38
rs4151689 Chr.5: 140848414-140848414 on Build GRCh38
rs181978789 Chr.5: 140848296-140848296 on Build GRCh38
rs764710818 Chr.5: 140848079-140848079 on Build GRCh38
rs934950988 Chr.5: 140848399-140848399 on Build GRCh38
rs782415361 Chr.5: 140848529-140848529 on Build GRCh38
rs4151687 Chr.5: 140848387-140848387 on Build GRCh38
rs782065425 Chr.5: 140848535-140848535 on Build GRCh38
rs782806998 Chr.5: 140848449-140848449 on Build GRCh38
rs973261439 Chr.5: 140848334-140848334 on Build GRCh38
rs782524885 Chr.5: 140848487-140848487 on Build GRCh38
rs571649192 Chr.5: 140848385-140848385 on Build GRCh38
rs782051282 Chr.5: 140848458-140848458 on Build GRCh38
rs782015840 Chr.5: 140848533-140848533 on Build GRCh38
rs116232683 Chr.5: 140848031-140848031 on Build GRCh38
rs189573491 Chr.5: 140847992-140847992 on Build GRCh38
rs551344579 Chr.5: 140848335-140848335 on Build GRCh38
rs143939535 Chr.5: 140848422-140848422 on Build GRCh38
rs782748098 Chr.5: 140848466-140848466 on Build GRCh38
rs941918814 Chr.5: 140848329-140848329 on Build GRCh38
rs781806569 Chr.5: 140848456-140848456 on Build GRCh38

View Full Product Details