Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113002742-113003341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OAS2
Entrez Gene ID
4939
Gene Name
2'-5'-oligoadenylate synthetase 2
Gene Aliases
-
UniGene
Hs.414332
Gene Chromosome Location
Chr.12: 112978395-113011723 on Build GRCh38
Transcripts
NM_002535.2 NM_016817.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4400941 c.1138A>C p.N380H Chr.12: 113003061-113003061 on Build GRCh38
7144921 c.1148G>T p.R383L Chr.12: 113003071-113003071 on Build GRCh38
1298939 c.1053C>A p.F351L Chr.12: 113002976-113002976 on Build GRCh38
396888 c.1020C>G p.L340L Chr.12: 113002943-113002943 on Build GRCh38
6265601 c.1026G>T p.T342T Chr.12: 113002949-113002949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551044967 Chr.12: 113003270-113003270 on Build GRCh38
rs757252857 Chr.12: 113003122-113003122 on Build GRCh38
rs758263196 Chr.12: 113002966-113002967 on Build GRCh38
rs745515783 Chr.12: 113003044-113003044 on Build GRCh38
rs771647147 Chr.12: 113003051-113003051 on Build GRCh38
rs574968398 Chr.12: 113002948-113002948 on Build GRCh38
rs560824430 Chr.12: 113003317-113003317 on Build GRCh38
rs562079346 Chr.12: 113003141-113003141 on Build GRCh38
rs985160141 Chr.12: 113002861-113002861 on Build GRCh38
rs769473601 Chr.12: 113002773-113002773 on Build GRCh38
rs746687058 Chr.12: 113003135-113003135 on Build GRCh38
rs370758817 Chr.12: 113003120-113003120 on Build GRCh38
rs200404382 Chr.12: 113003070-113003070 on Build GRCh38
rs763845869 Chr.12: 113003112-113003112 on Build GRCh38
rs987643503 Chr.12: 113003239-113003239 on Build GRCh38
rs756741874 Chr.12: 113003029-113003029 on Build GRCh38
rs533594916 Chr.12: 113003318-113003318 on Build GRCh38
rs778830527 Chr.12: 113003126-113003126 on Build GRCh38
rs375557231 Chr.12: 113002864-113002864 on Build GRCh38
rs529185320 Chr.12: 113002781-113002781 on Build GRCh38
rs370660600 Chr.12: 113002966-113002966 on Build GRCh38
rs1021578635 Chr.12: 113002798-113002798 on Build GRCh38
rs759283739 Chr.12: 113003136-113003136 on Build GRCh38
rs1014950681 Chr.12: 113003177-113003177 on Build GRCh38
rs755318769 Chr.12: 113002981-113002981 on Build GRCh38
rs755908961 Chr.12: 113003111-113003111 on Build GRCh38
rs142235676 Chr.12: 113003274-113003274 on Build GRCh38
rs777724485 Chr.12: 113003015-113003015 on Build GRCh38
rs748780958 Chr.12: 113002887-113002887 on Build GRCh38
rs142826885 Chr.12: 113003043-113003043 on Build GRCh38
rs189886076 Chr.12: 113002799-113002799 on Build GRCh38
rs752475666 Chr.12: 113002999-113002999 on Build GRCh38
rs767853072 Chr.12: 113002983-113002983 on Build GRCh38
rs922154783 Chr.12: 113002845-113002845 on Build GRCh38
rs774505880 Chr.12: 113003098-113003098 on Build GRCh38
rs763055902 Chr.12: 113002968-113002968 on Build GRCh38
rs773227838 Chr.12: 113002862-113002862 on Build GRCh38
rs765719615 Chr.12: 113002962-113002962 on Build GRCh38
rs147623979 Chr.12: 113003144-113003144 on Build GRCh38
rs184906370 Chr.12: 113002765-113002765 on Build GRCh38
rs762111508 Chr.12: 113003313-113003313 on Build GRCh38
rs1056936200 Chr.12: 113003113-113003113 on Build GRCh38
rs983617843 Chr.12: 113003247-113003247 on Build GRCh38
rs909434474 Chr.12: 113003248-113003248 on Build GRCh38
rs533081787 Chr.12: 113003241-113003241 on Build GRCh38
rs963599395 Chr.12: 113003252-113003252 on Build GRCh38
rs766129779 Chr.12: 113003026-113003026 on Build GRCh38
rs202215696 Chr.12: 113002953-113002953 on Build GRCh38
rs748498814 Chr.12: 113003071-113003071 on Build GRCh38
rs770094178 Chr.12: 113003075-113003075 on Build GRCh38
rs771505985 Chr.12: 113003152-113003152 on Build GRCh38
rs927309427 Chr.12: 113003261-113003261 on Build GRCh38
rs749881286 Chr.12: 113003128-113003128 on Build GRCh38
rs770178595 Chr.12: 113003138-113003138 on Build GRCh38
rs965129241 Chr.12: 113003201-113003201 on Build GRCh38
rs1025416298 Chr.12: 113003222-113003222 on Build GRCh38
rs950947042 Chr.12: 113003240-113003240 on Build GRCh38
rs974199071 Chr.12: 113002847-113002847 on Build GRCh38
rs1017972979 Chr.12: 113003148-113003148 on Build GRCh38
rs374072119 Chr.12: 113002776-113002776 on Build GRCh38
rs947034222 Chr.12: 113003290-113003290 on Build GRCh38
rs751817154 Chr.12: 113002979-113002979 on Build GRCh38
rs922010362 Chr.12: 113003207-113003207 on Build GRCh38
rs1053096513 Chr.12: 113003262-113003262 on Build GRCh38
rs150642424 Chr.12: 113003101-113003101 on Build GRCh38
rs749225940 Chr.12: 113003020-113003020 on Build GRCh38
rs772814355 Chr.12: 113003026-113003026 on Build GRCh38
rs891430056 Chr.12: 113003004-113003004 on Build GRCh38
rs2072137 Chr.12: 113003116-113003116 on Build GRCh38
rs374251982 Chr.12: 113002895-113002895 on Build GRCh38
rs373335209 Chr.12: 113003210-113003210 on Build GRCh38
rs546184269 Chr.12: 113003054-113003054 on Build GRCh38
rs984090888 Chr.12: 113003007-113003007 on Build GRCh38
rs980047435 Chr.12: 113003260-113003260 on Build GRCh38
rs973879418 Chr.12: 113002811-113002811 on Build GRCh38
rs763425188 Chr.12: 113003091-113003091 on Build GRCh38
rs909507974 Chr.12: 113003017-113003017 on Build GRCh38
rs762846971 Chr.12: 113002952-113002952 on Build GRCh38
rs550386906 Chr.12: 113003193-113003193 on Build GRCh38
rs751303051 Chr.12: 113003038-113003038 on Build GRCh38
rs777581337 Chr.12: 113002885-113002885 on Build GRCh38
rs752892060 Chr.12: 113003108-113003108 on Build GRCh38
rs963696894 Chr.12: 113003183-113003183 on Build GRCh38
rs932131515 Chr.12: 113002857-113002857 on Build GRCh38
rs768999781 Chr.12: 113002945-113002945 on Build GRCh38
rs1010581201 Chr.12: 113002792-113002792 on Build GRCh38
rs755280606 Chr.12: 113003090-113003090 on Build GRCh38
rs759701630 Chr.12: 113003100-113003100 on Build GRCh38
rs766558000 Chr.12: 113003095-113003095 on Build GRCh38
rs766613230 Chr.12: 113002977-113002977 on Build GRCh38
rs745573049 Chr.12: 113002922-113002922 on Build GRCh38
rs374760083 Chr.12: 113003129-113003129 on Build GRCh38
rs773965576 Chr.12: 113002916-113002916 on Build GRCh38
rs181599146 Chr.12: 113003009-113003009 on Build GRCh38

View Full Product Details