Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 7787739-7788269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PER3
Entrez Gene ID
8863
Gene Name
period circadian clock 3
Gene Aliases
FASPS3, GIG13
UniGene
Hs.162200
Gene Chromosome Location
Chr.1: 7784285-7845181 on Build GRCh38
Transcripts
NM_001289861.1 NM_001289862.1 NM_001289863.1 NM_001289864.1 NM_016831.2 XM_005263524.3 XM_011542385.2 XM_011542390.2 XM_017002723.1 XM_017002724.1 XM_017002725.1 XM_017002726.1 XM_017002727.1 XM_017002728.1 XM_017002729.1 XM_017002730.1 XM_017002731.1 XM_017002732.1 XM_017002733.1 XM_017002734.1 XM_017002735.1 XM_017002736.1 XM_017002737.1 XM_017002738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5616759 c.543G>A p.A181A Chr.1: 7788197-7788197 on Build GRCh38
6905367 c.443C>G p.S148C Chr.1: 7788097-7788097 on Build GRCh38
6191703 c.474G>C p.K158N Chr.1: 7788128-7788128 on Build GRCh38
4010012 c.478G>C p.D160H Chr.1: 7788132-7788132 on Build GRCh38
5025175 c.536T>C p.F179S Chr.1: 7788190-7788190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772027021 Chr.1: 7788171-7788171 on Build GRCh38
rs750025676 Chr.1: 7788197-7788197 on Build GRCh38
rs549704202 Chr.1: 7788124-7788124 on Build GRCh38
rs758743497 Chr.1: 7788057-7788057 on Build GRCh38
rs765816057 Chr.1: 7788050-7788050 on Build GRCh38
rs1014230694 Chr.1: 7788242-7788242 on Build GRCh38
rs754936631 Chr.1: 7788216-7788216 on Build GRCh38
rs1004510176 Chr.1: 7787925-7787925 on Build GRCh38
rs1006572035 Chr.1: 7787917-7787917 on Build GRCh38
rs371175980 Chr.1: 7787973-7787973 on Build GRCh38
rs113068661 Chr.1: 7788082-7788082 on Build GRCh38
rs1039554603 Chr.1: 7788084-7788084 on Build GRCh38
rs375445968 Chr.1: 7788123-7788123 on Build GRCh38
rs764458292 Chr.1: 7788061-7788061 on Build GRCh38
rs760310021 Chr.1: 7788200-7788200 on Build GRCh38
rs780654670 Chr.1: 7788073-7788073 on Build GRCh38
rs746102872 Chr.1: 7788238-7788238 on Build GRCh38
rs1025716093 Chr.1: 7788227-7788227 on Build GRCh38
rs150445246 Chr.1: 7788196-7788196 on Build GRCh38
rs759624365 Chr.1: 7788114-7788114 on Build GRCh38
rs754947224 Chr.1: 7788165-7788165 on Build GRCh38
rs926792047 Chr.1: 7788005-7788005 on Build GRCh38
rs755897834 Chr.1: 7788063-7788063 on Build GRCh38
rs769189782 Chr.1: 7788193-7788193 on Build GRCh38
rs776823126 Chr.1: 7788099-7788099 on Build GRCh38
rs774356456 Chr.1: 7788088-7788088 on Build GRCh38
rs995571610 Chr.1: 7787978-7787978 on Build GRCh38
rs1027954614 Chr.1: 7787900-7787900 on Build GRCh38
rs1010179828 Chr.1: 7787856-7787856 on Build GRCh38
rs765899798 Chr.1: 7788205-7788205 on Build GRCh38
rs1045184575 Chr.1: 7787781-7787781 on Build GRCh38
rs1042338429 Chr.1: 7788053-7788053 on Build GRCh38
rs777438003 Chr.1: 7788234-7788234 on Build GRCh38
rs535750061 Chr.1: 7787773-7787773 on Build GRCh38
rs773508531 Chr.1: 7788007-7788007 on Build GRCh38
rs755591980 Chr.1: 7788137-7788137 on Build GRCh38
rs779990925 Chr.1: 7788081-7788081 on Build GRCh38
rs368619694 Chr.1: 7788026-7788026 on Build GRCh38
rs925248960 Chr.1: 7787785-7787785 on Build GRCh38
rs747221099 Chr.1: 7788094-7788094 on Build GRCh38
rs994748578 Chr.1: 7787892-7787892 on Build GRCh38
rs764365010 Chr.1: 7787809-7787809 on Build GRCh38
rs765256883 Chr.1: 7788116-7788116 on Build GRCh38
rs756958718 Chr.1: 7788068-7788068 on Build GRCh38
rs761421650 Chr.1: 7788133-7788133 on Build GRCh38
rs979887576 Chr.1: 7787983-7787983 on Build GRCh38
rs143936373 Chr.1: 7788195-7788195 on Build GRCh38
rs750063430 Chr.1: 7788136-7788136 on Build GRCh38
rs535936436 Chr.1: 7788194-7788194 on Build GRCh38
rs779843477 Chr.1: 7788098-7788099 on Build GRCh38
rs373305861 Chr.1: 7788016-7788016 on Build GRCh38
rs754126880 Chr.1: 7787827-7787827 on Build GRCh38
rs116020316 Chr.1: 7787903-7787903 on Build GRCh38
rs763378200 Chr.1: 7788121-7788121 on Build GRCh38
rs559800857 Chr.1: 7787967-7787967 on Build GRCh38
rs768430140 Chr.1: 7788083-7788083 on Build GRCh38
rs1017983072 Chr.1: 7787943-7787943 on Build GRCh38
rs228727 Chr.1: 7787776-7787776 on Build GRCh38
rs150199185 Chr.1: 7788142-7788142 on Build GRCh38
rs766564077 Chr.1: 7788013-7788013 on Build GRCh38
rs779433185 Chr.1: 7788140-7788140 on Build GRCh38
rs113509050 Chr.1: 7788160-7788160 on Build GRCh38
rs191955069 Chr.1: 7788143-7788143 on Build GRCh38
rs948103382 Chr.1: 7787777-7787777 on Build GRCh38
rs202065261 Chr.1: 7788010-7788010 on Build GRCh38
rs776011723 Chr.1: 7788119-7788119 on Build GRCh38
rs781517960 Chr.1: 7788179-7788179 on Build GRCh38
rs371669928 Chr.1: 7788225-7788225 on Build GRCh38
rs898245898 Chr.1: 7788215-7788215 on Build GRCh38
rs886790940 Chr.1: 7788232-7788232 on Build GRCh38
rs890501867 Chr.1: 7787759-7787759 on Build GRCh38
rs753180203 Chr.1: 7788055-7788055 on Build GRCh38
rs748860326 Chr.1: 7788180-7788182 on Build GRCh38
rs758212151 Chr.1: 7788230-7788230 on Build GRCh38
rs763490378 Chr.1: 7788186-7788186 on Build GRCh38
rs747997827 Chr.1: 7788152-7788152 on Build GRCh38
rs547035195 Chr.1: 7788180-7788180 on Build GRCh38
rs1051358141 Chr.1: 7787872-7787872 on Build GRCh38
rs531385815 Chr.1: 7788077-7788077 on Build GRCh38
rs891706025 Chr.1: 7787839-7787839 on Build GRCh38
rs7556518 Chr.1: 7787786-7787786 on Build GRCh38
rs200046179 Chr.1: 7788218-7788218 on Build GRCh38
rs755609184 Chr.1: 7788079-7788079 on Build GRCh38
rs900374576 Chr.1: 7787849-7787849 on Build GRCh38
rs753337798 Chr.1: 7788211-7788211 on Build GRCh38
rs767114073 Chr.1: 7788134-7788134 on Build GRCh38
rs778831783 Chr.1: 7788151-7788151 on Build GRCh38
rs925352263 Chr.1: 7788109-7788109 on Build GRCh38
rs771329983 Chr.1: 7788098-7788098 on Build GRCh38
rs141195095 Chr.1: 7788184-7788184 on Build GRCh38
rs200948632 Chr.1: 7788064-7788064 on Build GRCh38
rs1023983693 Chr.1: 7787950-7787950 on Build GRCh38

View Full Product Details