Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31105506-31106103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SELM
Entrez Gene ID
140606
Gene Name
selenoprotein M
Gene Aliases
SEPM
UniGene
Hs.55940
Gene Chromosome Location
Chr.22: 31104777-31107565 on Build GRCh38
Transcripts
NM_080430.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6768972 c.154C>T p.R52C Chr.22: 31105941-31105941 on Build GRCh38
6378410 c.143G>T p.*48L Chr.22: 31105952-31105952 on Build GRCh38
4793154 c.173C>A p.A58D Chr.22: 31105685-31105685 on Build GRCh38
4724656 c.160A>G p.K54E Chr.22: 31105935-31105935 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs561670110 Chr.22: 31106023-31106023 on Build GRCh38
rs544103699 Chr.22: 31105914-31105917 on Build GRCh38
rs373767032 Chr.22: 31105674-31105674 on Build GRCh38
rs777873669 Chr.22: 31105960-31105960 on Build GRCh38
rs112745491 Chr.22: 31105657-31105657 on Build GRCh38
rs776436408 Chr.22: 31105669-31105669 on Build GRCh38
rs560373620 Chr.22: 31105964-31105964 on Build GRCh38
rs35563586 Chr.22: 31105527-31105528 on Build GRCh38
rs372862700 Chr.22: 31105929-31105929 on Build GRCh38
rs772173012 Chr.22: 31105640-31105640 on Build GRCh38
rs762345221 Chr.22: 31105895-31105895 on Build GRCh38
rs775535772 Chr.22: 31105894-31105894 on Build GRCh38
rs575684940 Chr.22: 31105792-31105792 on Build GRCh38
rs769712132 Chr.22: 31105892-31105892 on Build GRCh38
rs113640912 Chr.22: 31105968-31105968 on Build GRCh38
rs1029163470 Chr.22: 31105622-31105622 on Build GRCh38
rs756460604 Chr.22: 31106049-31106049 on Build GRCh38
rs2283875 Chr.22: 31105847-31105847 on Build GRCh38
rs755484623 Chr.22: 31105721-31105721 on Build GRCh38
rs901346657 Chr.22: 31106046-31106046 on Build GRCh38
rs11705035 Chr.22: 31106076-31106076 on Build GRCh38
rs776348151 Chr.22: 31105886-31105886 on Build GRCh38
rs553271664 Chr.22: 31105749-31105749 on Build GRCh38
rs991375283 Chr.22: 31105770-31105770 on Build GRCh38
rs754260849 Chr.22: 31105720-31105720 on Build GRCh38
rs370035098 Chr.22: 31105748-31105748 on Build GRCh38
rs375827780 Chr.22: 31105774-31105774 on Build GRCh38
rs748274421 Chr.22: 31105632-31105632 on Build GRCh38
rs779069281 Chr.22: 31105618-31105618 on Build GRCh38
rs771142645 Chr.22: 31105883-31105883 on Build GRCh38
rs903431712 Chr.22: 31105569-31105569 on Build GRCh38
rs11705054 Chr.22: 31105995-31105995 on Build GRCh38
rs999085785 Chr.22: 31105580-31105580 on Build GRCh38
rs762587461 Chr.22: 31105680-31105680 on Build GRCh38
rs758854708 Chr.22: 31105950-31105950 on Build GRCh38
rs535568284 Chr.22: 31105544-31105544 on Build GRCh38
rs1033156255 Chr.22: 31105581-31105581 on Build GRCh38
rs200661049 Chr.22: 31105943-31105943 on Build GRCh38
rs776373563 Chr.22: 31105716-31105716 on Build GRCh38
rs915768462 Chr.22: 31106015-31106015 on Build GRCh38
rs903209161 Chr.22: 31105840-31105840 on Build GRCh38
rs572753057 Chr.22: 31105982-31105982 on Build GRCh38
rs751600040 Chr.22: 31105961-31105961 on Build GRCh38
rs375741208 Chr.22: 31105725-31105725 on Build GRCh38
rs758548837 Chr.22: 31105737-31105737 on Build GRCh38
rs375349301 Chr.22: 31105867-31105867 on Build GRCh38
rs149176411 Chr.22: 31105841-31105841 on Build GRCh38
rs760045678 Chr.22: 31105941-31105941 on Build GRCh38
rs905200853 Chr.22: 31105686-31105686 on Build GRCh38
rs1052252537 Chr.22: 31105655-31105655 on Build GRCh38
rs763620807 Chr.22: 31105685-31105685 on Build GRCh38
rs773854891 Chr.22: 31106006-31106006 on Build GRCh38
rs912730491 Chr.22: 31105552-31105552 on Build GRCh38
rs370198471 Chr.22: 31105676-31105676 on Build GRCh38
rs58114992 Chr.22: 31105778-31105778 on Build GRCh38
rs550783006 Chr.22: 31105658-31105658 on Build GRCh38
rs896309660 Chr.22: 31105663-31105663 on Build GRCh38
rs547988236 Chr.22: 31105561-31105561 on Build GRCh38
rs768481721 Chr.22: 31106001-31106001 on Build GRCh38
rs758032723 Chr.22: 31105819-31105819 on Build GRCh38
rs368307324 Chr.22: 31105903-31105903 on Build GRCh38
rs540540257 Chr.22: 31105940-31105940 on Build GRCh38
rs751271893 Chr.22: 31105691-31105691 on Build GRCh38
rs182689622 Chr.22: 31105644-31105644 on Build GRCh38
rs756882332 Chr.22: 31105701-31105701 on Build GRCh38
rs763610803 Chr.22: 31105910-31105910 on Build GRCh38
rs765168425 Chr.22: 31105675-31105675 on Build GRCh38
rs769624443 Chr.22: 31105983-31105983 on Build GRCh38
rs757452970 Chr.22: 31105969-31105969 on Build GRCh38
rs374036477 Chr.22: 31105755-31105755 on Build GRCh38
rs773764383 Chr.22: 31105925-31105925 on Build GRCh38
rs921614685 Chr.22: 31105589-31105589 on Build GRCh38
rs935102998 Chr.22: 31105806-31105806 on Build GRCh38
rs555629509 Chr.22: 31105780-31105780 on Build GRCh38
rs781457609 Chr.22: 31105975-31105975 on Build GRCh38
rs745718217 Chr.22: 31105887-31105887 on Build GRCh38
rs113723658 Chr.22: 31105695-31105695 on Build GRCh38
rs1027980752 Chr.22: 31105959-31105959 on Build GRCh38
rs773554197 Chr.22: 31105642-31105642 on Build GRCh38
rs756384126 Chr.22: 31105648-31105650 on Build GRCh38
rs568098713 Chr.22: 31105602-31105602 on Build GRCh38
rs371089151 Chr.22: 31106068-31106068 on Build GRCh38
rs893266897 Chr.22: 31105858-31105858 on Build GRCh38
rs997421604 Chr.22: 31105865-31105865 on Build GRCh38
rs753938731 Chr.22: 31105954-31105955 on Build GRCh38
rs1028487560 Chr.22: 31106055-31106055 on Build GRCh38
rs1028525132 Chr.22: 31105866-31105866 on Build GRCh38
rs1045689967 Chr.22: 31105804-31105804 on Build GRCh38
rs200447643 Chr.22: 31105728-31105728 on Build GRCh38
rs767290809 Chr.22: 31105939-31105939 on Build GRCh38
rs747209827 Chr.22: 31105881-31105881 on Build GRCh38
rs145240749 Chr.22: 31105722-31105722 on Build GRCh38

View Full Product Details