Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202726041-202726356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM117B
Entrez Gene ID
150864
Gene Name
family with sequence similarity 117 member B
Gene Aliases
ALS2CR13
UniGene
Hs.471130
Gene Chromosome Location
Chr.2: 202635178-202769757 on Build GRCh38
Transcripts
NM_173511.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3372607 c.860G>T p.R287L Chr.2: 202726263-202726263 on Build GRCh38
719733 c.196C>T p.H66Y Chr.2: 202726331-202726331 on Build GRCh38
2908070 c.893G>A p.R298Q Chr.2: 202726296-202726296 on Build GRCh38
6658144 c.853A>G p.K285E Chr.2: 202726256-202726256 on Build GRCh38
7165858 c.115-1G>C p.? Chr.2: 202726249-202726249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61740247 Chr.2: 202726304-202726304 on Build GRCh38
rs906631743 Chr.2: 202726198-202726198 on Build GRCh38
rs113030263 Chr.2: 202726252-202726252 on Build GRCh38
rs192421016 Chr.2: 202726217-202726217 on Build GRCh38
rs763129785 Chr.2: 202726244-202726244 on Build GRCh38
rs753503653 Chr.2: 202726282-202726282 on Build GRCh38
rs973668580 Chr.2: 202726115-202726115 on Build GRCh38
rs937752517 Chr.2: 202726124-202726124 on Build GRCh38
rs764636553 Chr.2: 202726250-202726250 on Build GRCh38
rs772060239 Chr.2: 202726201-202726201 on Build GRCh38
rs777641751 Chr.2: 202726318-202726318 on Build GRCh38
rs764833158 Chr.2: 202726234-202726234 on Build GRCh38
rs377125931 Chr.2: 202726280-202726280 on Build GRCh38
rs770730408 Chr.2: 202726321-202726321 on Build GRCh38
rs771838646 Chr.2: 202726100-202726100 on Build GRCh38
rs987306265 Chr.2: 202726092-202726092 on Build GRCh38
rs36094769 Chr.2: 202726158-202726159 on Build GRCh38
rs778395003 Chr.2: 202726295-202726295 on Build GRCh38
rs370316883 Chr.2: 202726327-202726327 on Build GRCh38
rs775779695 Chr.2: 202726232-202726232 on Build GRCh38
rs752133206 Chr.2: 202726267-202726267 on Build GRCh38
rs926413497 Chr.2: 202726117-202726117 on Build GRCh38
rs200658071 Chr.2: 202726236-202726236 on Build GRCh38
rs907678302 Chr.2: 202726109-202726109 on Build GRCh38
rs546088745 Chr.2: 202726079-202726079 on Build GRCh38
rs920631669 Chr.2: 202726127-202726127 on Build GRCh38
rs774211714 Chr.2: 202726149-202726149 on Build GRCh38
rs746763648 Chr.2: 202726216-202726216 on Build GRCh38
rs954411582 Chr.2: 202726086-202726086 on Build GRCh38
rs775569250 Chr.2: 202726138-202726138 on Build GRCh38
rs1041980878 Chr.2: 202726209-202726209 on Build GRCh38
rs754640787 Chr.2: 202726294-202726294 on Build GRCh38
rs532358298 Chr.2: 202726173-202726173 on Build GRCh38
rs746090581 Chr.2: 202726332-202726332 on Build GRCh38
rs1011185138 Chr.2: 202726262-202726262 on Build GRCh38
rs562887167 Chr.2: 202726223-202726223 on Build GRCh38
rs372869909 Chr.2: 202726220-202726220 on Build GRCh38
rs187144021 Chr.2: 202726151-202726151 on Build GRCh38
rs746952864 Chr.2: 202726320-202726320 on Build GRCh38
rs899946344 Chr.2: 202726305-202726305 on Build GRCh38
rs770668447 Chr.2: 202726292-202726293 on Build GRCh38
rs572406805 Chr.2: 202726069-202726069 on Build GRCh38
rs763752705 Chr.2: 202726207-202726207 on Build GRCh38
rs757599870 Chr.2: 202726273-202726273 on Build GRCh38
rs563023873 Chr.2: 202726122-202726123 on Build GRCh38

View Full Product Details