Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7243577-7244133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDNEP1
Entrez Gene ID
23399
Gene Name
CTD nuclear envelope phosphatase 1
Gene Aliases
DULLARD, HSA011916, NET56
UniGene
Hs.513913
Gene Chromosome Location
Chr.17: 7243587-7251940 on Build GRCh38
Transcripts
NM_001143775.1 NM_015343.4

SNP Detail

SNP ID Chromosome Location
rs911106065 Chr.17: 7243613-7243613 on Build GRCh38
rs868635572 Chr.17: 7244108-7244108 on Build GRCh38
rs906915803 Chr.17: 7243977-7243977 on Build GRCh38
rs1051925532 Chr.17: 7243868-7243868 on Build GRCh38
rs16956630 Chr.17: 7243709-7243709 on Build GRCh38
rs551507969 Chr.17: 7243966-7243966 on Build GRCh38
rs767548427 Chr.17: 7243688-7243691 on Build GRCh38
rs1024983030 Chr.17: 7243807-7243808 on Build GRCh38
rs968153551 Chr.17: 7243808-7243808 on Build GRCh38
rs942539497 Chr.17: 7243647-7243647 on Build GRCh38
rs911146302 Chr.17: 7243873-7243873 on Build GRCh38
rs1010168127 Chr.17: 7243659-7243659 on Build GRCh38
rs549190536 Chr.17: 7243814-7243814 on Build GRCh38
rs575100595 Chr.17: 7243938-7243949 on Build GRCh38
rs1022455810 Chr.17: 7243799-7243799 on Build GRCh38
rs371312829 Chr.17: 7243994-7243994 on Build GRCh38
rs942614215 Chr.17: 7243902-7243902 on Build GRCh38
rs904389168 Chr.17: 7243674-7243674 on Build GRCh38
rs973906013 Chr.17: 7243937-7243937 on Build GRCh38
rs760186192 Chr.17: 7243699-7243699 on Build GRCh38
rs986789403 Chr.17: 7243858-7243858 on Build GRCh38
rs929764675 Chr.17: 7243964-7243964 on Build GRCh38
rs16956625 Chr.17: 7243691-7243691 on Build GRCh38
rs375061566 Chr.17: 7244082-7244082 on Build GRCh38
rs397815162 Chr.17: 7244033-7244034 on Build GRCh38
rs1046747200 Chr.17: 7243735-7243735 on Build GRCh38
rs529161062 Chr.17: 7243813-7243813 on Build GRCh38
rs16956628 Chr.17: 7243703-7243703 on Build GRCh38
rs945685685 Chr.17: 7243899-7243899 on Build GRCh38
rs397782425 Chr.17: 7244032-7244033 on Build GRCh38
rs1012441615 Chr.17: 7243776-7243776 on Build GRCh38
rs898273643 Chr.17: 7243707-7243707 on Build GRCh38
rs1002502621 Chr.17: 7243759-7243759 on Build GRCh38
rs1001311160 Chr.17: 7243687-7243687 on Build GRCh38
rs372251830 Chr.17: 7243730-7243730 on Build GRCh38
rs534752723 Chr.17: 7243938-7243938 on Build GRCh38
rs1012492151 Chr.17: 7244057-7244057 on Build GRCh38
rs571313836 Chr.17: 7244039-7244039 on Build GRCh38
rs549880804 Chr.17: 7243602-7243602 on Build GRCh38
rs933067531 Chr.17: 7243866-7243867 on Build GRCh38
rs955560012 Chr.17: 7243846-7243846 on Build GRCh38
rs922058056 Chr.17: 7243854-7243854 on Build GRCh38
rs1055443410 Chr.17: 7244002-7244002 on Build GRCh38
rs1046800297 Chr.17: 7243965-7243965 on Build GRCh38
rs562519076 Chr.17: 7243681-7243681 on Build GRCh38
rs909434086 Chr.17: 7243814-7243814 on Build GRCh38
rs978163004 Chr.17: 7243812-7243812 on Build GRCh38
rs974906995 Chr.17: 7243824-7243824 on Build GRCh38
rs1035691036 Chr.17: 7243761-7243761 on Build GRCh38
rs992164013 Chr.17: 7243778-7243778 on Build GRCh38
rs993863007 Chr.17: 7243708-7243708 on Build GRCh38
rs1051449 Chr.17: 7243785-7243785 on Build GRCh38
rs1013683808 Chr.17: 7244110-7244110 on Build GRCh38
rs753337024 Chr.17: 7243889-7243889 on Build GRCh38
rs757142685 Chr.17: 7244059-7244060 on Build GRCh38
rs768030065 Chr.17: 7243741-7243741 on Build GRCh38
rs569950308 Chr.17: 7243789-7243789 on Build GRCh38
rs8930 Chr.17: 7243916-7243916 on Build GRCh38
rs565881349 Chr.17: 7243905-7243905 on Build GRCh38
rs1022505153 Chr.17: 7244064-7244064 on Build GRCh38
rs776322228 Chr.17: 7243862-7243862 on Build GRCh38
rs983736448 Chr.17: 7243813-7243814 on Build GRCh38
rs906859622 Chr.17: 7243746-7243746 on Build GRCh38
rs1043008610 Chr.17: 7243950-7243950 on Build GRCh38
rs895372793 Chr.17: 7243771-7243771 on Build GRCh38
rs762347491 Chr.17: 7243978-7243978 on Build GRCh38
rs34926505 Chr.17: 7244031-7244032 on Build GRCh38
rs895413415 Chr.17: 7244055-7244055 on Build GRCh38
rs775038886 Chr.17: 7243667-7243667 on Build GRCh38
rs938361829 Chr.17: 7243989-7243989 on Build GRCh38

View Full Product Details