Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78692317-78692737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH1
Entrez Gene ID
9267
Gene Name
cytohesin 1
Gene Aliases
B2-1, CYTOHESIN-1, D17S811E, PSCD1, SEC7
UniGene
Hs.191215
Gene Chromosome Location
Chr.17: 78674047-78782342 on Build GRCh38
Transcripts
NM_001292018.1 NM_001292019.1 NM_004762.3 NM_017456.3 XM_011525475.2 XM_011525476.2 XM_011525477.1 XM_011525478.1 XM_017025336.1 XM_017025337.1 XM_017025338.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5513372 c.841C>T p.R281C Chr.17: 78692467-78692467 on Build GRCh38
7003782 c.890C>T p.T297M Chr.17: 78692418-78692418 on Build GRCh38
7245697 c.875A>G p.Y292C Chr.17: 78692433-78692433 on Build GRCh38
6639542 c.891G>A p.T297T Chr.17: 78692417-78692417 on Build GRCh38
7003783 c.890C>T p.T297M Chr.17: 78692418-78692418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754286384 Chr.17: 78692508-78692508 on Build GRCh38
rs902203552 Chr.17: 78692586-78692586 on Build GRCh38
rs557334889 Chr.17: 78692708-78692708 on Build GRCh38
rs759134866 Chr.17: 78692524-78692526 on Build GRCh38
rs758521237 Chr.17: 78692384-78692384 on Build GRCh38
rs373955119 Chr.17: 78692541-78692541 on Build GRCh38
rs749021541 Chr.17: 78692553-78692553 on Build GRCh38
rs762842090 Chr.17: 78692445-78692445 on Build GRCh38
rs759233925 Chr.17: 78692500-78692500 on Build GRCh38
rs770451515 Chr.17: 78692450-78692450 on Build GRCh38
rs371453580 Chr.17: 78692453-78692453 on Build GRCh38
rs1043799516 Chr.17: 78692562-78692562 on Build GRCh38
rs747884437 Chr.17: 78692417-78692417 on Build GRCh38
rs201654081 Chr.17: 78692489-78692489 on Build GRCh38
rs776487502 Chr.17: 78692403-78692403 on Build GRCh38
rs367619543 Chr.17: 78692374-78692374 on Build GRCh38
rs142053234 Chr.17: 78692610-78692610 on Build GRCh38
rs999692953 Chr.17: 78692583-78692583 on Build GRCh38
rs565847103 Chr.17: 78692563-78692563 on Build GRCh38
rs769822835 Chr.17: 78692428-78692428 on Build GRCh38
rs973343714 Chr.17: 78692396-78692396 on Build GRCh38
rs536933234 Chr.17: 78692707-78692707 on Build GRCh38
rs372380664 Chr.17: 78692521-78692521 on Build GRCh38
rs773106656 Chr.17: 78692429-78692429 on Build GRCh38
rs762060933 Chr.17: 78692511-78692511 on Build GRCh38
rs758923238 Chr.17: 78692520-78692520 on Build GRCh38
rs768700248 Chr.17: 78692400-78692400 on Build GRCh38
rs930437140 Chr.17: 78692336-78692336 on Build GRCh38
rs757847315 Chr.17: 78692367-78692367 on Build GRCh38
rs969724998 Chr.17: 78692659-78692659 on Build GRCh38
rs751273354 Chr.17: 78692500-78692500 on Build GRCh38
rs200585265 Chr.17: 78692501-78692501 on Build GRCh38
rs896085567 Chr.17: 78692626-78692626 on Build GRCh38
rs746148655 Chr.17: 78692399-78692399 on Build GRCh38
rs148622796 Chr.17: 78692595-78692595 on Build GRCh38
rs368086085 Chr.17: 78692516-78692516 on Build GRCh38
rs568472664 Chr.17: 78692611-78692611 on Build GRCh38
rs373997238 Chr.17: 78692370-78692370 on Build GRCh38
rs1052638966 Chr.17: 78692589-78692589 on Build GRCh38
rs999102478 Chr.17: 78692592-78692592 on Build GRCh38
rs191432612 Chr.17: 78692352-78692352 on Build GRCh38
rs200353991 Chr.17: 78692490-78692490 on Build GRCh38
rs374612374 Chr.17: 78692368-78692368 on Build GRCh38
rs780238056 Chr.17: 78692391-78692391 on Build GRCh38
rs750995087 Chr.17: 78692519-78692519 on Build GRCh38
rs769874503 Chr.17: 78692517-78692517 on Build GRCh38
rs8065203 Chr.17: 78692497-78692497 on Build GRCh38

View Full Product Details