Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6760648-6761054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2D3A
Entrez Gene ID
10045
Gene Name
SH2 domain containing 3A
Gene Aliases
NSP1
UniGene
Hs.439645
Gene Chromosome Location
Chr.19: 6752160-6767513 on Build GRCh38
Transcripts
NM_005490.2 XM_005259467.1 XM_005259472.2 XM_006722602.1 XM_011527608.2 XM_011527609.2 XM_017026123.1 XM_017026124.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5456789 c.362G>A p.R121Q Chr.19: 6760695-6760695 on Build GRCh38
5493789 c.113G>A p.R38H Chr.19: 6760944-6760944 on Build GRCh38
2728142 c.266C>T p.P89L Chr.19: 6760791-6760791 on Build GRCh38
6868875 c.161G>T p.R54L Chr.19: 6760896-6760896 on Build GRCh38
5000111 c.155G>A p.R52H Chr.19: 6760902-6760902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780377852 Chr.19: 6760755-6760755 on Build GRCh38
rs762593452 Chr.19: 6760910-6760910 on Build GRCh38
rs201030472 Chr.19: 6760905-6760905 on Build GRCh38
rs1013793715 Chr.19: 6760833-6760833 on Build GRCh38
rs776442591 Chr.19: 6760901-6760901 on Build GRCh38
rs985784380 Chr.19: 6760846-6760846 on Build GRCh38
rs143102735 Chr.19: 6760790-6760790 on Build GRCh38
rs781586587 Chr.19: 6760804-6760804 on Build GRCh38
rs765407222 Chr.19: 6760681-6760681 on Build GRCh38
rs756817738 Chr.19: 6760727-6760727 on Build GRCh38
rs976659800 Chr.19: 6760911-6760911 on Build GRCh38
rs568486135 Chr.19: 6760673-6760673 on Build GRCh38
rs764885044 Chr.19: 6760903-6760903 on Build GRCh38
rs760456128 Chr.19: 6760907-6760907 on Build GRCh38
rs761602238 Chr.19: 6760970-6760970 on Build GRCh38
rs893802890 Chr.19: 6760729-6760729 on Build GRCh38
rs562186406 Chr.19: 6761009-6761009 on Build GRCh38
rs988182316 Chr.19: 6760885-6760885 on Build GRCh38
rs965672727 Chr.19: 6761016-6761016 on Build GRCh38
rs138522995 Chr.19: 6760791-6760791 on Build GRCh38
rs778826830 Chr.19: 6760948-6760948 on Build GRCh38
rs932509223 Chr.19: 6760770-6760770 on Build GRCh38
rs7258236 Chr.19: 6760963-6760963 on Build GRCh38
rs200497909 Chr.19: 6760850-6760850 on Build GRCh38
rs199503315 Chr.19: 6760979-6760980 on Build GRCh38
rs749972880 Chr.19: 6760844-6760844 on Build GRCh38
rs745908479 Chr.19: 6760951-6760951 on Build GRCh38
rs777064873 Chr.19: 6760759-6760759 on Build GRCh38
rs185333277 Chr.19: 6760854-6760854 on Build GRCh38
rs201681732 Chr.19: 6760974-6760974 on Build GRCh38
rs7258083 Chr.19: 6760997-6760997 on Build GRCh38
rs562996882 Chr.19: 6760866-6760866 on Build GRCh38
rs747824684 Chr.19: 6760714-6760715 on Build GRCh38
rs758978010 Chr.19: 6760748-6760748 on Build GRCh38
rs371836476 Chr.19: 6760881-6760881 on Build GRCh38
rs753537539 Chr.19: 6760830-6760830 on Build GRCh38
rs753122669 Chr.19: 6760935-6760935 on Build GRCh38
rs764830130 Chr.19: 6760839-6760839 on Build GRCh38
rs770030908 Chr.19: 6760781-6760781 on Build GRCh38
rs547835903 Chr.19: 6760713-6760713 on Build GRCh38
rs371648594 Chr.19: 6760943-6760943 on Build GRCh38
rs1032487660 Chr.19: 6760860-6760860 on Build GRCh38
rs781176336 Chr.19: 6761025-6761025 on Build GRCh38
rs7254471 Chr.19: 6760942-6760942 on Build GRCh38
rs141564856 Chr.19: 6760813-6760813 on Build GRCh38
rs761547112 Chr.19: 6760902-6760902 on Build GRCh38
rs774491633 Chr.19: 6760689-6760689 on Build GRCh38
rs535033168 Chr.19: 6760867-6760867 on Build GRCh38
rs754129420 Chr.19: 6761005-6761005 on Build GRCh38
rs781676135 Chr.19: 6760865-6760865 on Build GRCh38
rs745960619 Chr.19: 6761012-6761012 on Build GRCh38
rs375581861 Chr.19: 6760787-6760787 on Build GRCh38
rs762496110 Chr.19: 6760977-6760977 on Build GRCh38
rs973571252 Chr.19: 6760816-6760816 on Build GRCh38
rs776494865 Chr.19: 6760966-6760966 on Build GRCh38
rs368368775 Chr.19: 6760828-6760828 on Build GRCh38
rs548598782 Chr.19: 6760976-6760976 on Build GRCh38
rs925089461 Chr.19: 6760691-6760691 on Build GRCh38
rs767313865 Chr.19: 6760693-6760693 on Build GRCh38
rs768478372 Chr.19: 6760897-6760897 on Build GRCh38
rs201397651 Chr.19: 6760945-6760945 on Build GRCh38
rs138101388 Chr.19: 6760916-6760916 on Build GRCh38
rs748066373 Chr.19: 6761032-6761032 on Build GRCh38
rs772992363 Chr.19: 6760769-6760770 on Build GRCh38
rs749830561 Chr.19: 6760855-6760855 on Build GRCh38
rs571112823 Chr.19: 6760747-6760747 on Build GRCh38
rs755569404 Chr.19: 6760928-6760928 on Build GRCh38
rs76024146 Chr.19: 6760773-6760773 on Build GRCh38
rs566329999 Chr.19: 6760848-6760848 on Build GRCh38
rs367560553 Chr.19: 6760864-6760864 on Build GRCh38
rs759054803 Chr.19: 6760996-6760996 on Build GRCh38
rs769078843 Chr.19: 6760758-6760758 on Build GRCh38
rs753482424 Chr.19: 6760721-6760721 on Build GRCh38
rs370407162 Chr.19: 6760772-6760772 on Build GRCh38
rs771676114 Chr.19: 6760770-6760771 on Build GRCh38
rs111881825 Chr.19: 6760933-6760933 on Build GRCh38
rs149620409 Chr.19: 6760696-6760696 on Build GRCh38
rs749455704 Chr.19: 6760944-6760944 on Build GRCh38
rs866094093 Chr.19: 6760929-6760929 on Build GRCh38
rs775222907 Chr.19: 6760805-6760805 on Build GRCh38
rs372429785 Chr.19: 6760692-6760692 on Build GRCh38
rs146265436 Chr.19: 6760896-6760896 on Build GRCh38
rs777797779 Chr.19: 6760869-6760869 on Build GRCh38
rs775553958 Chr.19: 6760893-6760893 on Build GRCh38
rs893883445 Chr.19: 6760915-6760915 on Build GRCh38
rs755804645 Chr.19: 6760712-6760712 on Build GRCh38
rs770909990 Chr.19: 6760890-6760890 on Build GRCh38
rs985815478 Chr.19: 6761006-6761006 on Build GRCh38
rs761425623 Chr.19: 6760831-6760831 on Build GRCh38
rs375700170 Chr.19: 6760849-6760849 on Build GRCh38
rs572061070 Chr.19: 6760930-6760930 on Build GRCh38
rs758408390 Chr.19: 6761014-6761014 on Build GRCh38
rs765957448 Chr.19: 6760981-6760981 on Build GRCh38
rs542474671 Chr.19: 6761007-6761007 on Build GRCh38
rs747416409 Chr.19: 6760756-6760756 on Build GRCh38
rs970941342 Chr.19: 6760771-6760771 on Build GRCh38
rs778372376 Chr.19: 6760746-6760746 on Build GRCh38
rs7246122 Chr.19: 6761000-6761000 on Build GRCh38
rs774016930 Chr.19: 6760993-6760993 on Build GRCh38
rs752160908 Chr.19: 6760926-6760926 on Build GRCh38
rs751284302 Chr.19: 6760838-6760839 on Build GRCh38
rs953200485 Chr.19: 6760842-6760842 on Build GRCh38

View Full Product Details