Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105589319-105589726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF38
Entrez Gene ID
54848
Gene Name
Rho guanine nucleotide exchange factor 38
Gene Aliases
-
UniGene
Hs.585224
Gene Chromosome Location
Chr.4: 105552603-105682539 on Build GRCh38
Transcripts
NM_001242729.1 NM_017700.1 XM_011532050.2 XM_011532051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1309597 c.330C>G p.L110L Chr.4: 105589381-105589381 on Build GRCh38
1183532 c.283C>T p.R95W Chr.4: 105589334-105589334 on Build GRCh38
3824900 c.330C>T p.L110L Chr.4: 105589381-105589381 on Build GRCh38
133396 c.286_287GA>TT p.E96L Chr.4: 105589337-105589338 on Build GRCh38
3824901 c.330C>T p.L110L Chr.4: 105589381-105589381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014685962 Chr.4: 105589678-105589678 on Build GRCh38
rs79342312 Chr.4: 105589403-105589403 on Build GRCh38
rs111829609 Chr.4: 105589366-105589366 on Build GRCh38
rs887320942 Chr.4: 105589649-105589649 on Build GRCh38
rs375499919 Chr.4: 105589436-105589436 on Build GRCh38
rs376809428 Chr.4: 105589469-105589469 on Build GRCh38
rs369544620 Chr.4: 105589704-105589704 on Build GRCh38
rs754001818 Chr.4: 105589474-105589474 on Build GRCh38
rs925912550 Chr.4: 105589546-105589546 on Build GRCh38
rs781159008 Chr.4: 105589447-105589448 on Build GRCh38
rs780008075 Chr.4: 105589383-105589384 on Build GRCh38
rs752845523 Chr.4: 105589393-105589393 on Build GRCh38
rs769590354 Chr.4: 105589361-105589361 on Build GRCh38
rs373844781 Chr.4: 105589536-105589536 on Build GRCh38
rs894741402 Chr.4: 105589572-105589572 on Build GRCh38
rs759344809 Chr.4: 105589459-105589459 on Build GRCh38
rs754124541 Chr.4: 105589404-105589404 on Build GRCh38
rs765574141 Chr.4: 105589383-105589383 on Build GRCh38
rs780516720 Chr.4: 105589430-105589430 on Build GRCh38
rs751443596 Chr.4: 105589387-105589388 on Build GRCh38
rs757602737 Chr.4: 105589410-105589410 on Build GRCh38
rs897747068 Chr.4: 105589702-105589702 on Build GRCh38
rs774233475 Chr.4: 105589454-105589454 on Build GRCh38
rs987724514 Chr.4: 105589486-105589486 on Build GRCh38
rs539154608 Chr.4: 105589468-105589468 on Build GRCh38
rs10018352 Chr.4: 105589472-105589472 on Build GRCh38
rs769611964 Chr.4: 105589453-105589454 on Build GRCh38
rs140625801 Chr.4: 105589386-105589386 on Build GRCh38
rs1005034002 Chr.4: 105589654-105589654 on Build GRCh38
rs185075193 Chr.4: 105589445-105589445 on Build GRCh38
rs750638877 Chr.4: 105589382-105589384 on Build GRCh38
rs767468964 Chr.4: 105589467-105589467 on Build GRCh38
rs1000440779 Chr.4: 105589340-105589340 on Build GRCh38
rs759526452 Chr.4: 105589389-105589389 on Build GRCh38
rs775632952 Chr.4: 105589418-105589418 on Build GRCh38
rs949510376 Chr.4: 105589609-105589609 on Build GRCh38
rs762027198 Chr.4: 105589481-105589481 on Build GRCh38
rs181691627 Chr.4: 105589565-105589565 on Build GRCh38
rs747637780 Chr.4: 105589434-105589434 on Build GRCh38
rs17035881 Chr.4: 105589525-105589525 on Build GRCh38
rs146153509 Chr.4: 105589387-105589387 on Build GRCh38
rs764271968 Chr.4: 105589398-105589398 on Build GRCh38
rs917970732 Chr.4: 105589577-105589577 on Build GRCh38
rs953192861 Chr.4: 105589475-105589475 on Build GRCh38
rs898798315 Chr.4: 105589686-105589686 on Build GRCh38
rs147453356 Chr.4: 105589441-105589441 on Build GRCh38
rs139274936 Chr.4: 105589573-105589573 on Build GRCh38
rs756237394 Chr.4: 105589394-105589394 on Build GRCh38
rs377228216 Chr.4: 105589390-105589390 on Build GRCh38
rs762317046 Chr.4: 105589360-105589363 on Build GRCh38
rs1056033263 Chr.4: 105589566-105589566 on Build GRCh38
rs189458754 Chr.4: 105589537-105589537 on Build GRCh38
rs562293060 Chr.4: 105589527-105589527 on Build GRCh38
rs905064310 Chr.4: 105589640-105589640 on Build GRCh38
rs1044914265 Chr.4: 105589638-105589638 on Build GRCh38
rs754899735 Chr.4: 105589389-105589390 on Build GRCh38
rs779347709 Chr.4: 105589412-105589412 on Build GRCh38
rs144590203 Chr.4: 105589425-105589425 on Build GRCh38
rs765524129 Chr.4: 105589378-105589379 on Build GRCh38
rs765474555 Chr.4: 105589487-105589487 on Build GRCh38
rs758450908 Chr.4: 105589382-105589383 on Build GRCh38
rs1056064658 Chr.4: 105589345-105589345 on Build GRCh38
rs370449378 Chr.4: 105589415-105589415 on Build GRCh38
rs941905805 Chr.4: 105589648-105589648 on Build GRCh38
rs35284621 Chr.4: 105589524-105589525 on Build GRCh38
rs1046298311 Chr.4: 105589611-105589611 on Build GRCh38
rs994482695 Chr.4: 105589691-105589691 on Build GRCh38
rs747759960 Chr.4: 105589354-105589354 on Build GRCh38
rs377313480 Chr.4: 105589657-105589657 on Build GRCh38
rs770568838 Chr.4: 105589447-105589447 on Build GRCh38
rs894607782 Chr.4: 105589360-105589360 on Build GRCh38
rs35054816 Chr.4: 105589363-105589364 on Build GRCh38
rs576769258 Chr.4: 105589454-105589454 on Build GRCh38

View Full Product Details