Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13469073-13469611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFOD1
Entrez Gene ID
54438
Gene Name
glucose-fructose oxidoreductase domain containing 1
Gene Aliases
ADG-90, C6orf114
UniGene
Hs.484686
Gene Chromosome Location
Chr.6: 13357830-13487637 on Build GRCh38
Transcripts
NM_001242628.1 NM_001242629.1 NM_018988.3 NR_038459.1

SNP Detail

SNP ID Chromosome Location
rs527703116 Chr.6: 13469398-13469398 on Build GRCh38
rs879285465 Chr.6: 13469186-13469186 on Build GRCh38
rs567764240 Chr.6: 13469471-13469471 on Build GRCh38
rs146815336 Chr.6: 13469233-13469233 on Build GRCh38
rs563089995 Chr.6: 13469257-13469257 on Build GRCh38
rs943713236 Chr.6: 13469147-13469147 on Build GRCh38
rs547774115 Chr.6: 13469469-13469469 on Build GRCh38
rs961780612 Chr.6: 13469151-13469151 on Build GRCh38
rs945351065 Chr.6: 13469563-13469563 on Build GRCh38
rs375271016 Chr.6: 13469343-13469343 on Build GRCh38
rs764948816 Chr.6: 13469266-13469266 on Build GRCh38
rs1041292993 Chr.6: 13469353-13469353 on Build GRCh38
rs559829149 Chr.6: 13469137-13469137 on Build GRCh38
rs910465400 Chr.6: 13469214-13469214 on Build GRCh38
rs1016370091 Chr.6: 13469407-13469407 on Build GRCh38
rs993243076 Chr.6: 13469466-13469466 on Build GRCh38
rs956726134 Chr.6: 13469099-13469099 on Build GRCh38
rs114079615 Chr.6: 13469264-13469264 on Build GRCh38
rs1015905971 Chr.6: 13469148-13469148 on Build GRCh38
rs917989164 Chr.6: 13469115-13469115 on Build GRCh38
rs980815411 Chr.6: 13469274-13469274 on Build GRCh38
rs776620767 Chr.6: 13469255-13469255 on Build GRCh38
rs904315715 Chr.6: 13469232-13469232 on Build GRCh38
rs958224467 Chr.6: 13469514-13469514 on Build GRCh38
rs971773074 Chr.6: 13469155-13469155 on Build GRCh38
rs747081075 Chr.6: 13469161-13469161 on Build GRCh38
rs191038964 Chr.6: 13469380-13469380 on Build GRCh38
rs372054463 Chr.6: 13469221-13469221 on Build GRCh38
rs908773857 Chr.6: 13469345-13469345 on Build GRCh38
rs1021348103 Chr.6: 13469227-13469227 on Build GRCh38
rs572403800 Chr.6: 13469438-13469438 on Build GRCh38
rs778929778 Chr.6: 13469445-13469445 on Build GRCh38
rs768522539 Chr.6: 13469254-13469254 on Build GRCh38
rs998681834 Chr.6: 13469434-13469434 on Build GRCh38
rs989395923 Chr.6: 13469555-13469555 on Build GRCh38
rs1024950952 Chr.6: 13469262-13469262 on Build GRCh38
rs536745889 Chr.6: 13469520-13469520 on Build GRCh38
rs910357242 Chr.6: 13469498-13469498 on Build GRCh38
rs1002240690 Chr.6: 13469560-13469560 on Build GRCh38
rs1047277627 Chr.6: 13469132-13469132 on Build GRCh38
rs948147153 Chr.6: 13469117-13469117 on Build GRCh38
rs761612556 Chr.6: 13469265-13469265 on Build GRCh38
rs954425154 Chr.6: 13469484-13469484 on Build GRCh38
rs960253612 Chr.6: 13469247-13469247 on Build GRCh38
rs1014159272 Chr.6: 13469501-13469501 on Build GRCh38
rs920015205 Chr.6: 13469280-13469280 on Build GRCh38
rs944319373 Chr.6: 13469347-13469347 on Build GRCh38
rs952958647 Chr.6: 13469142-13469142 on Build GRCh38
rs35753962 Chr.6: 13469464-13469465 on Build GRCh38
rs772309120 Chr.6: 13469118-13469118 on Build GRCh38
rs920085157 Chr.6: 13469125-13469125 on Build GRCh38
rs913931560 Chr.6: 13469562-13469562 on Build GRCh38
rs992199863 Chr.6: 13469105-13469105 on Build GRCh38
rs933148935 Chr.6: 13469177-13469177 on Build GRCh38
rs762540577 Chr.6: 13469394-13469394 on Build GRCh38
rs920905433 Chr.6: 13469385-13469385 on Build GRCh38
rs904305015 Chr.6: 13469417-13469417 on Build GRCh38
rs986390182 Chr.6: 13469492-13469492 on Build GRCh38
rs772887085 Chr.6: 13469284-13469284 on Build GRCh38
rs932322321 Chr.6: 13469396-13469396 on Build GRCh38
rs878874290 Chr.6: 13469420-13469420 on Build GRCh38
rs539812503 Chr.6: 13469130-13469130 on Build GRCh38
rs892453250 Chr.6: 13469243-13469243 on Build GRCh38
rs984866554 Chr.6: 13469143-13469143 on Build GRCh38
rs748772638 Chr.6: 13469094-13469094 on Build GRCh38

View Full Product Details