Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161347979-161348552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARK2
Entrez Gene ID
5071
Gene Name
parkin RBR E3 ubiquitin protein ligase
Gene Aliases
AR-JP, LPRS2, PDJ, PRKN
UniGene
Hs.132954
Gene Chromosome Location
Chr.6: 161347417-162727802 on Build GRCh38
Transcripts
NM_004562.2 NM_013987.2 NM_013988.2 XM_011535863.1 XM_017010908.1 XM_017010909.1

SNP Detail

SNP ID Chromosome Location
rs886180554 Chr.6: 161348051-161348051 on Build GRCh38
rs564009047 Chr.6: 161348030-161348030 on Build GRCh38
rs779851186 Chr.6: 161348498-161348498 on Build GRCh38
rs1122470 Chr.6: 161348078-161348078 on Build GRCh38
rs987980881 Chr.6: 161348141-161348141 on Build GRCh38
rs753448476 Chr.6: 161348331-161348331 on Build GRCh38
rs369401074 Chr.6: 161348294-161348294 on Build GRCh38
rs1018991074 Chr.6: 161348379-161348379 on Build GRCh38
rs557087876 Chr.6: 161348336-161348336 on Build GRCh38
rs903812654 Chr.6: 161348100-161348100 on Build GRCh38
rs187134044 Chr.6: 161348414-161348414 on Build GRCh38
rs1056630162 Chr.6: 161348264-161348264 on Build GRCh38
rs890969338 Chr.6: 161348332-161348332 on Build GRCh38
rs956131835 Chr.6: 161348117-161348117 on Build GRCh38
rs563061113 Chr.6: 161348018-161348018 on Build GRCh38
rs939090729 Chr.6: 161348257-161348257 on Build GRCh38
rs908057327 Chr.6: 161348203-161348203 on Build GRCh38
rs114587790 Chr.6: 161348281-161348281 on Build GRCh38
rs901933135 Chr.6: 161348365-161348365 on Build GRCh38
rs144460207 Chr.6: 161348142-161348142 on Build GRCh38
rs945120623 Chr.6: 161348424-161348424 on Build GRCh38
rs182252890 Chr.6: 161348350-161348350 on Build GRCh38
rs757383204 Chr.6: 161348275-161348275 on Build GRCh38
rs983622342 Chr.6: 161348196-161348196 on Build GRCh38
rs768016642 Chr.6: 161348065-161348065 on Build GRCh38
rs974705253 Chr.6: 161348398-161348398 on Build GRCh38
rs939124630 Chr.6: 161348068-161348068 on Build GRCh38
rs937856017 Chr.6: 161348163-161348163 on Build GRCh38
rs894777651 Chr.6: 161348272-161348272 on Build GRCh38
rs552155916 Chr.6: 161348162-161348162 on Build GRCh38
rs535107932 Chr.6: 161348150-161348150 on Build GRCh38
rs930929078 Chr.6: 161348195-161348195 on Build GRCh38
rs1018979909 Chr.6: 161348354-161348354 on Build GRCh38
rs903465113 Chr.6: 161348042-161348042 on Build GRCh38
rs537494414 Chr.6: 161348297-161348297 on Build GRCh38
rs991889545 Chr.6: 161348173-161348173 on Build GRCh38
rs1026751725 Chr.6: 161348373-161348373 on Build GRCh38
rs1031281346 Chr.6: 161348103-161348103 on Build GRCh38
rs917638739 Chr.6: 161348423-161348423 on Build GRCh38
rs1022228804 Chr.6: 161348085-161348085 on Build GRCh38
rs752358219 Chr.6: 161348076-161348076 on Build GRCh38
rs1043447655 Chr.6: 161348301-161348301 on Build GRCh38
rs1048399777 Chr.6: 161348035-161348035 on Build GRCh38
rs977759553 Chr.6: 161348486-161348486 on Build GRCh38
rs77283740 Chr.6: 161348404-161348404 on Build GRCh38
rs148426411 Chr.6: 161348190-161348190 on Build GRCh38
rs886061230 Chr.6: 161348077-161348077 on Build GRCh38
rs75529362 Chr.6: 161348102-161348102 on Build GRCh38
rs543363389 Chr.6: 161348496-161348496 on Build GRCh38
rs964806864 Chr.6: 161348384-161348384 on Build GRCh38
rs903760515 Chr.6: 161348309-161348309 on Build GRCh38
rs924952261 Chr.6: 161348489-161348489 on Build GRCh38
rs577554278 Chr.6: 161348487-161348487 on Build GRCh38
rs564250931 Chr.6: 161348477-161348477 on Build GRCh38
rs748008270 Chr.6: 161348517-161348517 on Build GRCh38
rs1030811289 Chr.6: 161348327-161348327 on Build GRCh38
rs758136049 Chr.6: 161348422-161348422 on Build GRCh38
rs1056115186 Chr.6: 161348069-161348069 on Build GRCh38
rs999873726 Chr.6: 161348314-161348314 on Build GRCh38
rs889401575 Chr.6: 161348499-161348499 on Build GRCh38
rs763492565 Chr.6: 161348174-161348174 on Build GRCh38
rs953437558 Chr.6: 161348076-161348076 on Build GRCh38
rs376011629 Chr.6: 161348150-161348158 on Build GRCh38
rs563296835 Chr.6: 161348521-161348521 on Build GRCh38
rs903393231 Chr.6: 161348191-161348191 on Build GRCh38
rs41268559 Chr.6: 161348418-161348418 on Build GRCh38
rs16892481 Chr.6: 161348405-161348405 on Build GRCh38
rs972509346 Chr.6: 161348101-161348101 on Build GRCh38
rs991883389 Chr.6: 161348518-161348518 on Build GRCh38
rs779959263 Chr.6: 161348403-161348403 on Build GRCh38
rs959188620 Chr.6: 161348413-161348413 on Build GRCh38
rs993474624 Chr.6: 161348366-161348366 on Build GRCh38

View Full Product Details