Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 45648427-45649008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY1
Entrez Gene ID
107
Gene Name
adenylate cyclase 1
Gene Aliases
AC1, DFNB44
UniGene
Hs.192215
Gene Chromosome Location
Chr.7: 45574140-45723116 on Build GRCh38
Transcripts
NM_001281768.1 NM_021116.2 XM_005249584.3 XM_005249585.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426264 c.1037G>A p.R346H Chr.7: 45648686-45648686 on Build GRCh38
4164358 c.1111T>A p.C371S Chr.7: 45648760-45648760 on Build GRCh38
1090069 c.1148+1G>A p.? Chr.7: 45648798-45648798 on Build GRCh38
1090068 c.1148C>T p.T383I Chr.7: 45648797-45648797 on Build GRCh38
1182003 c.1075G>A p.G359S Chr.7: 45648724-45648724 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961855504 Chr.7: 45648514-45648514 on Build GRCh38
rs765590357 Chr.7: 45648729-45648729 on Build GRCh38
rs367905122 Chr.7: 45648518-45648518 on Build GRCh38
rs1009843791 Chr.7: 45648533-45648533 on Build GRCh38
rs573413960 Chr.7: 45648615-45648615 on Build GRCh38
rs771986782 Chr.7: 45648792-45648792 on Build GRCh38
rs781561820 Chr.7: 45648943-45648943 on Build GRCh38
rs973353735 Chr.7: 45648650-45648650 on Build GRCh38
rs747202140 Chr.7: 45648741-45648741 on Build GRCh38
rs577535651 Chr.7: 45648913-45648913 on Build GRCh38
rs763616744 Chr.7: 45648844-45648844 on Build GRCh38
rs374138021 Chr.7: 45648806-45648806 on Build GRCh38
rs377495931 Chr.7: 45648717-45648717 on Build GRCh38
rs972317411 Chr.7: 45648841-45648841 on Build GRCh38
rs777521054 Chr.7: 45648748-45648749 on Build GRCh38
rs755619189 Chr.7: 45648750-45648750 on Build GRCh38
rs367633641 Chr.7: 45648845-45648845 on Build GRCh38
rs375178707 Chr.7: 45648738-45648738 on Build GRCh38
rs550997034 Chr.7: 45648500-45648500 on Build GRCh38
rs1007494119 Chr.7: 45648460-45648460 on Build GRCh38
rs766853068 Chr.7: 45648831-45648831 on Build GRCh38
rs751536724 Chr.7: 45648750-45648751 on Build GRCh38
rs534112934 Chr.7: 45648616-45648616 on Build GRCh38
rs748918186 Chr.7: 45648754-45648754 on Build GRCh38
rs78870608 Chr.7: 45648809-45648809 on Build GRCh38
rs755563183 Chr.7: 45648843-45648843 on Build GRCh38
rs756142009 Chr.7: 45648448-45648448 on Build GRCh38
rs777635281 Chr.7: 45648648-45648648 on Build GRCh38
rs961935018 Chr.7: 45648940-45648950 on Build GRCh38
rs931399283 Chr.7: 45648707-45648707 on Build GRCh38
rs903651120 Chr.7: 45648829-45648829 on Build GRCh38
rs36092155 Chr.7: 45648474-45648474 on Build GRCh38
rs753777637 Chr.7: 45648553-45648553 on Build GRCh38
rs757415759 Chr.7: 45648744-45648744 on Build GRCh38
rs540142646 Chr.7: 45648556-45648556 on Build GRCh38
rs6946621 Chr.7: 45648976-45648976 on Build GRCh38
rs769555896 Chr.7: 45648647-45648647 on Build GRCh38
rs1020778556 Chr.7: 45648544-45648544 on Build GRCh38
rs866935862 Chr.7: 45648501-45648501 on Build GRCh38
rs752055362 Chr.7: 45648833-45648833 on Build GRCh38
rs927319109 Chr.7: 45648890-45648890 on Build GRCh38
rs901740750 Chr.7: 45648513-45648513 on Build GRCh38
rs373262043 Chr.7: 45648672-45648672 on Build GRCh38
rs569116222 Chr.7: 45648541-45648541 on Build GRCh38
rs770517415 Chr.7: 45648755-45648755 on Build GRCh38
rs151091562 Chr.7: 45648747-45648747 on Build GRCh38
rs199602800 Chr.7: 45648812-45648812 on Build GRCh38
rs141023985 Chr.7: 45648780-45648780 on Build GRCh38
rs763477299 Chr.7: 45648817-45648817 on Build GRCh38
rs775475672 Chr.7: 45648798-45648798 on Build GRCh38
rs770700155 Chr.7: 45648663-45648663 on Build GRCh38
rs776597667 Chr.7: 45648808-45648808 on Build GRCh38
rs202040047 Chr.7: 45648665-45648665 on Build GRCh38
rs754027303 Chr.7: 45648723-45648723 on Build GRCh38
rs371662831 Chr.7: 45648633-45648633 on Build GRCh38
rs961644475 Chr.7: 45648636-45648636 on Build GRCh38
rs761956492 Chr.7: 45648726-45648726 on Build GRCh38
rs748896419 Chr.7: 45648655-45648655 on Build GRCh38
rs3735672 Chr.7: 45648634-45648634 on Build GRCh38
rs952066733 Chr.7: 45648860-45648860 on Build GRCh38
rs200391923 Chr.7: 45648819-45648819 on Build GRCh38
rs775522653 Chr.7: 45648685-45648685 on Build GRCh38
rs140092857 Chr.7: 45648571-45648571 on Build GRCh38
rs780810358 Chr.7: 45648636-45648636 on Build GRCh38
rs757710192 Chr.7: 45648632-45648632 on Build GRCh38
rs777438847 Chr.7: 45648753-45648753 on Build GRCh38
rs764300669 Chr.7: 45648722-45648722 on Build GRCh38
rs937338376 Chr.7: 45648899-45648899 on Build GRCh38
rs747701119 Chr.7: 45648639-45648639 on Build GRCh38
rs772033654 Chr.7: 45648684-45648684 on Build GRCh38
rs371172219 Chr.7: 45648756-45648756 on Build GRCh38
rs1024596570 Chr.7: 45648903-45648903 on Build GRCh38
rs868399531 Chr.7: 45648847-45648847 on Build GRCh38
rs980698799 Chr.7: 45648861-45648861 on Build GRCh38

View Full Product Details