Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101479168-101479646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTRC
Entrez Gene ID
8945
Gene Name
beta-transducin repeat containing E3 ubiquitin protein ligase
Gene Aliases
BETA-TRCP, FBW1A, FBXW1, FBXW1A, FWD1, bTrCP, bTrCP1, betaTrCP
UniGene
Hs.643802
Gene Chromosome Location
Chr.10: 101353808-101557321 on Build GRCh38
Transcripts
NM_001256856.1 NM_003939.4 NM_033637.3 XM_006718054.2 XM_017016870.1 XM_017016871.1 XM_017016872.1 XM_017016873.1 XM_017016874.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6279433 c.296C>T p.A99V Chr.10: 101479429-101479429 on Build GRCh38
914025 c.303G>T p.K101N Chr.10: 101479436-101479436 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1054009341 Chr.10: 101479242-101479242 on Build GRCh38
rs569018789 Chr.10: 101479365-101479365 on Build GRCh38
rs1016165869 Chr.10: 101479291-101479291 on Build GRCh38
rs187181852 Chr.10: 101479257-101479257 on Build GRCh38
rs755998630 Chr.10: 101479419-101479419 on Build GRCh38
rs144139485 Chr.10: 101479389-101479389 on Build GRCh38
rs369641092 Chr.10: 101479408-101479408 on Build GRCh38
rs769838165 Chr.10: 101479318-101479318 on Build GRCh38
rs987526933 Chr.10: 101479579-101479579 on Build GRCh38
rs772532571 Chr.10: 101479478-101479478 on Build GRCh38
rs915053297 Chr.10: 101479252-101479252 on Build GRCh38
rs975800063 Chr.10: 101479305-101479305 on Build GRCh38
rs201198184 Chr.10: 101479216-101479218 on Build GRCh38
rs768990158 Chr.10: 101479473-101479473 on Build GRCh38
rs61011895 Chr.10: 101479209-101479211 on Build GRCh38
rs534544666 Chr.10: 101479482-101479482 on Build GRCh38
rs987191570 Chr.10: 101479212-101479212 on Build GRCh38
rs954493636 Chr.10: 101479578-101479578 on Build GRCh38
rs757028314 Chr.10: 101479409-101479409 on Build GRCh38
rs574520092 Chr.10: 101479597-101479597 on Build GRCh38
rs780198232 Chr.10: 101479437-101479437 on Build GRCh38
rs749262230 Chr.10: 101479445-101479445 on Build GRCh38
rs1016948346 Chr.10: 101479432-101479432 on Build GRCh38
rs200334099 Chr.10: 101479215-101479216 on Build GRCh38
rs759156895 Chr.10: 101479383-101479383 on Build GRCh38
rs1004410146 Chr.10: 101479264-101479264 on Build GRCh38
rs961802074 Chr.10: 101479303-101479303 on Build GRCh38
rs935291370 Chr.10: 101479225-101479225 on Build GRCh38
rs563965708 Chr.10: 101479217-101479217 on Build GRCh38
rs768704689 Chr.10: 101479342-101479342 on Build GRCh38
rs894496354 Chr.10: 101479508-101479508 on Build GRCh38
rs947881122 Chr.10: 101479283-101479283 on Build GRCh38
rs977317311 Chr.10: 101479468-101479468 on Build GRCh38
rs764869582 Chr.10: 101479385-101479385 on Build GRCh38
rs202081619 Chr.10: 101479417-101479417 on Build GRCh38
rs948678604 Chr.10: 101479531-101479531 on Build GRCh38
rs772765182 Chr.10: 101479361-101479361 on Build GRCh38
rs763583896 Chr.10: 101479405-101479405 on Build GRCh38
rs923843685 Chr.10: 101479214-101479214 on Build GRCh38
rs748572060 Chr.10: 101479476-101479476 on Build GRCh38
rs571133807 Chr.10: 101479533-101479533 on Build GRCh38
rs567132261 Chr.10: 101479577-101479580 on Build GRCh38
rs953012713 Chr.10: 101479235-101479235 on Build GRCh38
rs1027212759 Chr.10: 101479230-101479230 on Build GRCh38
rs371635822 Chr.10: 101479329-101479329 on Build GRCh38
rs762265700 Chr.10: 101479360-101479360 on Build GRCh38
rs963486376 Chr.10: 101479464-101479464 on Build GRCh38
rs139115116 Chr.10: 101479474-101479474 on Build GRCh38
rs774784817 Chr.10: 101479351-101479351 on Build GRCh38
rs781151619 Chr.10: 101479416-101479416 on Build GRCh38
rs192056444 Chr.10: 101479562-101479562 on Build GRCh38
rs398046209 Chr.10: 101479215-101479217 on Build GRCh38
rs546480626 Chr.10: 101479480-101479480 on Build GRCh38
rs559273998 Chr.10: 101479530-101479530 on Build GRCh38
rs762677464 Chr.10: 101479402-101479402 on Build GRCh38
rs901308378 Chr.10: 101479610-101479610 on Build GRCh38
rs556756983 Chr.10: 101479585-101479585 on Build GRCh38
rs1044465864 Chr.10: 101479296-101479296 on Build GRCh38
rs376577588 Chr.10: 101479341-101479341 on Build GRCh38

View Full Product Details