Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134382546-134383091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GAT1
Entrez Gene ID
27087
Gene Name
beta-1,3-glucuronyltransferase 1
Gene Aliases
CD57, GLCATP, GLCUATP, HNK1, LEU7, NK-1, NK1
UniGene
Hs.381050
Gene Chromosome Location
Chr.11: 134378504-134411986 on Build GRCh38
Transcripts
NM_018644.3 NM_054025.2 XM_005271506.3 XM_011542751.2 XM_011542753.2 XM_017017550.1 XM_017017551.1 XM_017017552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5985832 c.849G>A p.Q283Q Chr.11: 134382779-134382779 on Build GRCh38
6351173 c.957+3G>T p.? Chr.11: 134382707-134382707 on Build GRCh38
5182261 c.841G>T p.G281C Chr.11: 134382787-134382787 on Build GRCh38
5527224 c.925G>A p.D309N Chr.11: 134382742-134382742 on Build GRCh38
5970096 c.740C>T p.P247L Chr.11: 134382888-134382888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981705570 Chr.11: 134382934-134382934 on Build GRCh38
rs376439912 Chr.11: 134382698-134382698 on Build GRCh38
rs773055144 Chr.11: 134382802-134382802 on Build GRCh38
rs769534324 Chr.11: 134382801-134382801 on Build GRCh38
rs767123521 Chr.11: 134383013-134383013 on Build GRCh38
rs752788516 Chr.11: 134382697-134382697 on Build GRCh38
rs1049406648 Chr.11: 134383035-134383035 on Build GRCh38
rs1001170365 Chr.11: 134382705-134382705 on Build GRCh38
rs563406434 Chr.11: 134383056-134383056 on Build GRCh38
rs558795192 Chr.11: 134382851-134382851 on Build GRCh38
rs1020447818 Chr.11: 134383061-134383061 on Build GRCh38
rs397762096 Chr.11: 134383020-134383021 on Build GRCh38
rs754424869 Chr.11: 134382857-134382857 on Build GRCh38
rs1048301900 Chr.11: 134382685-134382685 on Build GRCh38
rs760771426 Chr.11: 134382822-134382822 on Build GRCh38
rs372009493 Chr.11: 134382836-134382836 on Build GRCh38
rs890782473 Chr.11: 134383050-134383050 on Build GRCh38
rs775744958 Chr.11: 134382809-134382809 on Build GRCh38
rs769737442 Chr.11: 134382882-134382882 on Build GRCh38
rs760392401 Chr.11: 134383018-134383019 on Build GRCh38
rs924786631 Chr.11: 134382593-134382593 on Build GRCh38
rs897055981 Chr.11: 134382643-134382643 on Build GRCh38
rs758006182 Chr.11: 134382844-134382844 on Build GRCh38
rs749052053 Chr.11: 134382996-134382996 on Build GRCh38
rs747278356 Chr.11: 134383014-134383014 on Build GRCh38
rs751889272 Chr.11: 134383037-134383037 on Build GRCh38
rs773707460 Chr.11: 134383019-134383019 on Build GRCh38
rs576517113 Chr.11: 134382623-134382623 on Build GRCh38
rs780666900 Chr.11: 134382862-134382862 on Build GRCh38
rs754455175 Chr.11: 134382835-134382835 on Build GRCh38
rs979643937 Chr.11: 134382837-134382837 on Build GRCh38
rs749073170 Chr.11: 134382892-134382892 on Build GRCh38
rs150095434 Chr.11: 134382899-134382899 on Build GRCh38
rs374245735 Chr.11: 134382749-134382749 on Build GRCh38
rs1035380375 Chr.11: 134382763-134382763 on Build GRCh38
rs145736695 Chr.11: 134382610-134382610 on Build GRCh38
rs112888938 Chr.11: 134382966-134382966 on Build GRCh38
rs1036895932 Chr.11: 134382637-134382637 on Build GRCh38
rs777065682 Chr.11: 134382700-134382700 on Build GRCh38
rs751267144 Chr.11: 134382974-134382974 on Build GRCh38
rs759789693 Chr.11: 134382687-134382687 on Build GRCh38
rs896978571 Chr.11: 134382727-134382727 on Build GRCh38
rs754260857 Chr.11: 134382702-134382702 on Build GRCh38
rs892845262 Chr.11: 134382621-134382621 on Build GRCh38
rs762279455 Chr.11: 134382929-134382929 on Build GRCh38
rs761915526 Chr.11: 134383016-134383016 on Build GRCh38
rs765956817 Chr.11: 134382965-134382965 on Build GRCh38
rs779482078 Chr.11: 134382743-134382743 on Build GRCh38
rs758041241 Chr.11: 134382739-134382739 on Build GRCh38
rs752444545 Chr.11: 134382845-134382845 on Build GRCh38
rs200357434 Chr.11: 134382707-134382707 on Build GRCh38
rs778848224 Chr.11: 134383012-134383012 on Build GRCh38
rs759715133 Chr.11: 134382803-134382803 on Build GRCh38
rs535028435 Chr.11: 134383015-134383015 on Build GRCh38
rs867499391 Chr.11: 134383007-134383007 on Build GRCh38
rs776825420 Chr.11: 134382928-134382928 on Build GRCh38
rs934435086 Chr.11: 134382609-134382609 on Build GRCh38
rs541357496 Chr.11: 134382982-134382982 on Build GRCh38
rs768609394 Chr.11: 134382752-134382752 on Build GRCh38
rs768927813 Chr.11: 134382912-134382912 on Build GRCh38
rs891010422 Chr.11: 134382654-134382654 on Build GRCh38
rs776480442 Chr.11: 134382791-134382791 on Build GRCh38
rs777476819 Chr.11: 134382887-134382887 on Build GRCh38
rs764657042 Chr.11: 134382701-134382701 on Build GRCh38
rs750780574 Chr.11: 134382840-134382840 on Build GRCh38
rs368413334 Chr.11: 134382825-134382825 on Build GRCh38
rs766662276 Chr.11: 134383011-134383011 on Build GRCh38
rs866851601 Chr.11: 134382631-134382631 on Build GRCh38
rs566101176 Chr.11: 134382571-134382571 on Build GRCh38
rs748150106 Chr.11: 134382881-134382881 on Build GRCh38
rs995305132 Chr.11: 134383064-134383064 on Build GRCh38
rs374034968 Chr.11: 134382839-134382839 on Build GRCh38
rs115362398 Chr.11: 134382947-134382947 on Build GRCh38
rs1036946841 Chr.11: 134383059-134383059 on Build GRCh38
rs767755219 Chr.11: 134382807-134382807 on Build GRCh38
rs763590055 Chr.11: 134383023-134383024 on Build GRCh38
rs759233252 Chr.11: 134383038-134383038 on Build GRCh38
rs775798071 Chr.11: 134382911-134382911 on Build GRCh38
rs544708919 Chr.11: 134383033-134383033 on Build GRCh38
rs772820151 Chr.11: 134382700-134382701 on Build GRCh38
rs766795411 Chr.11: 134383032-134383032 on Build GRCh38
rs112164035 Chr.11: 134382574-134382574 on Build GRCh38
rs145216044 Chr.11: 134383012-134383013 on Build GRCh38
rs1049642276 Chr.11: 134382640-134382640 on Build GRCh38
rs763308287 Chr.11: 134382964-134382964 on Build GRCh38
rs770818268 Chr.11: 134383002-134383002 on Build GRCh38
rs752179560 Chr.11: 134382990-134382990 on Build GRCh38
rs747947460 Chr.11: 134382799-134382799 on Build GRCh38
rs529740294 Chr.11: 134383018-134383018 on Build GRCh38
rs780720219 Chr.11: 134382983-134382983 on Build GRCh38
rs138392079 Chr.11: 134383055-134383055 on Build GRCh38
rs149652609 Chr.11: 134382992-134382992 on Build GRCh38
rs577584567 Chr.11: 134383060-134383060 on Build GRCh38
rs756026134 Chr.11: 134382991-134382991 on Build GRCh38
rs371152055 Chr.11: 134382731-134382731 on Build GRCh38
rs752137819 Chr.11: 134383014-134383015 on Build GRCh38
rs765729898 Chr.11: 134382937-134382937 on Build GRCh38
rs943850062 Chr.11: 134382635-134382635 on Build GRCh38
rs1046296304 Chr.11: 134382581-134382581 on Build GRCh38

View Full Product Details