Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36751345-36751897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
UniGene
Hs.371350
Gene Chromosome Location
Chr.21: 36750888-36990254 on Build GRCh38
Transcripts
NM_000411.6 NM_001242784.1 NM_001242785.1 XM_005260953.3 XM_005260955.3 XM_005260956.3 XM_006723994.2 XM_011529541.2 XM_017028330.1

SNP Detail

SNP ID Chromosome Location
rs1046038242 Chr.21: 36751704-36751704 on Build GRCh38
rs1044902669 Chr.21: 36751431-36751431 on Build GRCh38
rs904798819 Chr.21: 36751790-36751790 on Build GRCh38
rs778163309 Chr.21: 36751370-36751370 on Build GRCh38
rs1043018475 Chr.21: 36751782-36751782 on Build GRCh38
rs1012109625 Chr.21: 36751825-36751825 on Build GRCh38
rs904769299 Chr.21: 36751536-36751536 on Build GRCh38
rs765847266 Chr.21: 36751866-36751866 on Build GRCh38
rs768743975 Chr.21: 36751570-36751570 on Build GRCh38
rs180866987 Chr.21: 36751701-36751701 on Build GRCh38
rs965167208 Chr.21: 36751576-36751576 on Build GRCh38
rs750036993 Chr.21: 36751652-36751652 on Build GRCh38
rs184935954 Chr.21: 36751705-36751705 on Build GRCh38
rs1010233753 Chr.21: 36751512-36751512 on Build GRCh38
rs781277606 Chr.21: 36751466-36751466 on Build GRCh38
rs73210780 Chr.21: 36751622-36751622 on Build GRCh38
rs948682000 Chr.21: 36751478-36751478 on Build GRCh38
rs1045659692 Chr.21: 36751479-36751479 on Build GRCh38
rs535197739 Chr.21: 36751843-36751843 on Build GRCh38
rs796464077 Chr.21: 36751435-36751435 on Build GRCh38
rs767127723 Chr.21: 36751638-36751638 on Build GRCh38
rs571952016 Chr.21: 36751736-36751736 on Build GRCh38
rs973759174 Chr.21: 36751658-36751658 on Build GRCh38
rs934482639 Chr.21: 36751708-36751708 on Build GRCh38
rs946079045 Chr.21: 36751747-36751747 on Build GRCh38
rs927752811 Chr.21: 36751709-36751709 on Build GRCh38
rs557043296 Chr.21: 36751559-36751559 on Build GRCh38
rs142191610 Chr.21: 36751617-36751617 on Build GRCh38
rs997543600 Chr.21: 36751845-36751845 on Build GRCh38
rs1018371826 Chr.21: 36751571-36751571 on Build GRCh38
rs1025845070 Chr.21: 36751858-36751858 on Build GRCh38
rs1039126152 Chr.21: 36751828-36751828 on Build GRCh38
rs998665120 Chr.21: 36751538-36751538 on Build GRCh38
rs527966859 Chr.21: 36751753-36751753 on Build GRCh38
rs907008203 Chr.21: 36751516-36751516 on Build GRCh38
rs546201018 Chr.21: 36751802-36751802 on Build GRCh38
rs566088233 Chr.21: 36751830-36751830 on Build GRCh38
rs747359583 Chr.21: 36751419-36751419 on Build GRCh38
rs563443241 Chr.21: 36751816-36751816 on Build GRCh38
rs140135359 Chr.21: 36751546-36751546 on Build GRCh38
rs14407 Chr.21: 36751624-36751624 on Build GRCh38
rs993358580 Chr.21: 36751721-36751721 on Build GRCh38
rs1018060824 Chr.21: 36751595-36751595 on Build GRCh38
rs372670716 Chr.21: 36751523-36751523 on Build GRCh38
rs929467921 Chr.21: 36751383-36751383 on Build GRCh38
rs529410036 Chr.21: 36751679-36751679 on Build GRCh38
rs551660081 Chr.21: 36751713-36751713 on Build GRCh38
rs891751607 Chr.21: 36751509-36751513 on Build GRCh38
rs561055554 Chr.21: 36751803-36751803 on Build GRCh38
rs867039512 Chr.21: 36751657-36751657 on Build GRCh38
rs964981859 Chr.21: 36751596-36751596 on Build GRCh38
rs886057062 Chr.21: 36751812-36751812 on Build GRCh38
rs79868396 Chr.21: 36751691-36751691 on Build GRCh38
rs901379935 Chr.21: 36751826-36751826 on Build GRCh38
rs544647675 Chr.21: 36751524-36751524 on Build GRCh38
rs762952630 Chr.21: 36751552-36751552 on Build GRCh38
rs146199781 Chr.21: 36751432-36751432 on Build GRCh38
rs998141307 Chr.21: 36751604-36751604 on Build GRCh38
rs771127234 Chr.21: 36751428-36751428 on Build GRCh38
rs746169122 Chr.21: 36751475-36751475 on Build GRCh38

View Full Product Details