Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50093592-50093921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM5
Entrez Gene ID
10181
Gene Name
RNA binding motif protein 5
Gene Aliases
G15, H37, LUCA15, RMB5
UniGene
Hs.439480
Gene Chromosome Location
Chr.3: 50088908-50118964 on Build GRCh38
Transcripts
NM_005778.3 NR_036627.2 XM_006712917.1 XM_011533261.1 XM_011533262.1 XM_017005503.1 XM_017005504.1 XM_017005505.1 XM_017005506.1 XM_017005507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6992360 c.282G>C p.E94D Chr.3: 50093818-50093818 on Build GRCh38
6047107 c.267T>C p.H89H Chr.3: 50093803-50093803 on Build GRCh38
6887308 c.337G>A p.D113N Chr.3: 50093873-50093873 on Build GRCh38
6764182 c.190C>T p.R64C Chr.3: 50093726-50093726 on Build GRCh38
4625136 c.308G>A p.R103H Chr.3: 50093844-50093844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929400667 Chr.3: 50093671-50093671 on Build GRCh38
rs779433688 Chr.3: 50093800-50093800 on Build GRCh38
rs764037672 Chr.3: 50093667-50093667 on Build GRCh38
rs564500126 Chr.3: 50093693-50093693 on Build GRCh38
rs76635723 Chr.3: 50093672-50093672 on Build GRCh38
rs1057063084 Chr.3: 50093792-50093792 on Build GRCh38
rs867277191 Chr.3: 50093746-50093746 on Build GRCh38
rs370702681 Chr.3: 50093698-50093698 on Build GRCh38
rs769341522 Chr.3: 50093785-50093785 on Build GRCh38
rs766280891 Chr.3: 50093891-50093891 on Build GRCh38
rs760665020 Chr.3: 50093625-50093625 on Build GRCh38
rs370431662 Chr.3: 50093757-50093757 on Build GRCh38
rs747668290 Chr.3: 50093783-50093783 on Build GRCh38
rs142145196 Chr.3: 50093725-50093725 on Build GRCh38
rs768235838 Chr.3: 50093685-50093685 on Build GRCh38
rs746638558 Chr.3: 50093679-50093679 on Build GRCh38
rs759558165 Chr.3: 50093678-50093678 on Build GRCh38
rs751253922 Chr.3: 50093892-50093892 on Build GRCh38
rs776910364 Chr.3: 50093843-50093843 on Build GRCh38
rs140669787 Chr.3: 50093836-50093836 on Build GRCh38
rs779947411 Chr.3: 50093763-50093763 on Build GRCh38
rs143299571 Chr.3: 50093796-50093796 on Build GRCh38
rs919240816 Chr.3: 50093848-50093848 on Build GRCh38
rs762944410 Chr.3: 50093873-50093873 on Build GRCh38
rs760756173 Chr.3: 50093733-50093733 on Build GRCh38
rs950777638 Chr.3: 50093871-50093871 on Build GRCh38
rs201825095 Chr.3: 50093802-50093802 on Build GRCh38
rs144164147 Chr.3: 50093791-50093791 on Build GRCh38
rs562253492 Chr.3: 50093782-50093782 on Build GRCh38
rs764242527 Chr.3: 50093748-50093748 on Build GRCh38
rs775788411 Chr.3: 50093815-50093815 on Build GRCh38
rs17225749 Chr.3: 50093707-50093707 on Build GRCh38
rs767572155 Chr.3: 50093790-50093792 on Build GRCh38
rs758773562 Chr.3: 50093680-50093680 on Build GRCh38
rs190899757 Chr.3: 50093656-50093656 on Build GRCh38
rs575650873 Chr.3: 50093618-50093618 on Build GRCh38
rs754787036 Chr.3: 50093895-50093895 on Build GRCh38
rs775735149 Chr.3: 50093728-50093728 on Build GRCh38
rs562012691 Chr.3: 50093724-50093724 on Build GRCh38
rs746349284 Chr.3: 50093801-50093801 on Build GRCh38
rs751289041 Chr.3: 50093781-50093781 on Build GRCh38
rs145486184 Chr.3: 50093872-50093872 on Build GRCh38
rs550716644 Chr.3: 50093750-50093750 on Build GRCh38
rs201956265 Chr.3: 50093753-50093753 on Build GRCh38
rs566441117 Chr.3: 50093845-50093845 on Build GRCh38
rs772392655 Chr.3: 50093727-50093727 on Build GRCh38
rs760969950 Chr.3: 50093820-50093820 on Build GRCh38
rs529469481 Chr.3: 50093726-50093726 on Build GRCh38
rs752361115 Chr.3: 50093898-50093898 on Build GRCh38
rs765156684 Chr.3: 50093752-50093752 on Build GRCh38
rs765388451 Chr.3: 50093861-50093861 on Build GRCh38
rs540511916 Chr.3: 50093714-50093714 on Build GRCh38
rs1024741519 Chr.3: 50093664-50093664 on Build GRCh38

View Full Product Details