Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7669260-7669825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TP53
Entrez Gene ID
7157
Gene Name
tumor protein p53
Gene Aliases
BCC7, LFS1, P53, TRP53
UniGene
Hs.437460
Gene Chromosome Location
Chr.17: 7668402-7687550 on Build GRCh38
Transcripts
NM_000546.5 NM_001126112.2 NM_001126113.2 NM_001126114.2 NM_001126115.1 NM_001126116.1 NM_001126117.1 NM_001126118.1 NM_001276695.1 NM_001276696.1 NM_001276697.1 NM_001276698.1 NM_001276699.1 NM_001276760.1 NM_001276761.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5028203 c.1158G>C p.K386N Chr.17: 7669633-7669633 on Build GRCh38
7347279 c.1169delC p.P390fs*>4 Chr.17: 7669622-7669622 on Build GRCh38
6474196 c.1127C>T p.S376F Chr.17: 7669664-7669664 on Build GRCh38
6503572 c.1135_1136insA p.R379fs*3 Chr.17: 7669655-7669656 on Build GRCh38
12663 c.1101-2A>C p.? Chr.17: 7669692-7669692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036818198 Chr.17: 7669302-7669302 on Build GRCh38
rs371002418 Chr.17: 7669465-7669465 on Build GRCh38
rs545818827 Chr.17: 7669575-7669575 on Build GRCh38
rs749817236 Chr.17: 7669624-7669624 on Build GRCh38
rs863224682 Chr.17: 7669655-7669655 on Build GRCh38
rs562130162 Chr.17: 7669794-7669794 on Build GRCh38
rs1042526 Chr.17: 7669572-7669572 on Build GRCh38
rs986431055 Chr.17: 7669736-7669736 on Build GRCh38
rs958423177 Chr.17: 7669464-7669464 on Build GRCh38
rs80184930 Chr.17: 7669659-7669659 on Build GRCh38
rs1018513029 Chr.17: 7669410-7669410 on Build GRCh38
rs140346117 Chr.17: 7669414-7669415 on Build GRCh38
rs587781858 Chr.17: 7669671-7669671 on Build GRCh38
rs876658876 Chr.17: 7669676-7669676 on Build GRCh38
rs16956880 Chr.17: 7669404-7669404 on Build GRCh38
rs1024927068 Chr.17: 7669374-7669374 on Build GRCh38
rs771579509 Chr.17: 7669334-7669334 on Build GRCh38
rs922919883 Chr.17: 7669535-7669535 on Build GRCh38
rs876658982 Chr.17: 7669691-7669691 on Build GRCh38
rs953090502 Chr.17: 7669339-7669339 on Build GRCh38
rs759018180 Chr.17: 7669571-7669571 on Build GRCh38
rs1032606977 Chr.17: 7669458-7669458 on Build GRCh38
rs1027267843 Chr.17: 7669334-7669334 on Build GRCh38
rs730882009 Chr.17: 7669641-7669641 on Build GRCh38
rs145723145 Chr.17: 7669778-7669778 on Build GRCh38
rs749061599 Chr.17: 7669656-7669656 on Build GRCh38
rs778913915 Chr.17: 7669767-7669767 on Build GRCh38
rs35919705 Chr.17: 7669504-7669504 on Build GRCh38
rs786202368 Chr.17: 7669654-7669654 on Build GRCh38
rs1002854255 Chr.17: 7669419-7669419 on Build GRCh38
rs746581308 Chr.17: 7669579-7669579 on Build GRCh38
rs764011631 Chr.17: 7669666-7669666 on Build GRCh38
rs927888647 Chr.17: 7669631-7669631 on Build GRCh38
rs897689379 Chr.17: 7669703-7669703 on Build GRCh38
rs17883043 Chr.17: 7669763-7669763 on Build GRCh38
rs879254030 Chr.17: 7669593-7669593 on Build GRCh38
rs767047551 Chr.17: 7669741-7669741 on Build GRCh38
rs8065799 Chr.17: 7669600-7669600 on Build GRCh38
rs760698953 Chr.17: 7669558-7669558 on Build GRCh38
rs34486624 Chr.17: 7669295-7669295 on Build GRCh38
rs1031046075 Chr.17: 7669489-7669489 on Build GRCh38
rs897704706 Chr.17: 7669769-7669769 on Build GRCh38
rs374294340 Chr.17: 7669594-7669594 on Build GRCh38
rs948064327 Chr.17: 7669570-7669570 on Build GRCh38
rs560862596 Chr.17: 7669415-7669415 on Build GRCh38
rs769664911 Chr.17: 7669616-7669616 on Build GRCh38
rs774269719 Chr.17: 7669662-7669662 on Build GRCh38
rs915225600 Chr.17: 7669569-7669569 on Build GRCh38
rs765530090 Chr.17: 7669678-7669678 on Build GRCh38
rs150842067 Chr.17: 7669644-7669644 on Build GRCh38
rs587781736 Chr.17: 7669628-7669628 on Build GRCh38
rs587781664 Chr.17: 7669692-7669692 on Build GRCh38
rs897923545 Chr.17: 7669336-7669336 on Build GRCh38
rs1013659649 Chr.17: 7669369-7669369 on Build GRCh38
rs931650342 Chr.17: 7669588-7669588 on Build GRCh38
rs994478116 Chr.17: 7669314-7669314 on Build GRCh38
rs771458013 Chr.17: 7669467-7669467 on Build GRCh38
rs17881850 Chr.17: 7669739-7669739 on Build GRCh38
rs730882017 Chr.17: 7669666-7669666 on Build GRCh38
rs886053513 Chr.17: 7669510-7669510 on Build GRCh38
rs369567704 Chr.17: 7669603-7669603 on Build GRCh38
rs878854063 Chr.17: 7669670-7669670 on Build GRCh38
rs373710656 Chr.17: 7669642-7669642 on Build GRCh38
rs770970987 Chr.17: 7669662-7669662 on Build GRCh38
rs977343345 Chr.17: 7669437-7669437 on Build GRCh38
rs587783064 Chr.17: 7669626-7669626 on Build GRCh38
rs760820708 Chr.17: 7669663-7669663 on Build GRCh38
rs786204227 Chr.17: 7669689-7669689 on Build GRCh38
rs79516338 Chr.17: 7669300-7669300 on Build GRCh38
rs750853458 Chr.17: 7669733-7669733 on Build GRCh38
rs375913211 Chr.17: 7669503-7669503 on Build GRCh38

View Full Product Details