Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185418623-185419089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf47
Entrez Gene ID
441054
Gene Name
chromosome 4 open reading frame 47
Gene Aliases
-
UniGene
Hs.549398
Gene Chromosome Location
Chr.4: 185405227-185449828 on Build GRCh38
Transcripts
XM_017008236.1 XM_017008237.1 XM_017008238.1 XM_017008239.1 XM_017008241.1 XM_017008242.1 NM_018359.3 NR_028085.1

Gene Symbol
UFSP2
Entrez Gene ID
55325
Gene Name
UFM1 specific peptidase 2
Gene Aliases
BHD, C4orf20
UniGene
Hs.713548
Gene Chromosome Location
Chr.4: 185399540-185425985 on Build GRCh38
Transcripts
XM_017008236.1 XM_017008237.1 XM_017008238.1 XM_017008239.1 XM_017008241.1 XM_017008242.1 NM_018359.3 NR_028085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1053804 c.145A>G p.N49D Chr.4: 185418708-185418708 on Build GRCh38
4441500 c.109C>A p.H37N Chr.4: 185418744-185418744 on Build GRCh38
1053803 c.209C>T p.T70I Chr.4: 185418644-185418644 on Build GRCh38
5491861 c.85A>T p.I29F Chr.4: 185418768-185418768 on Build GRCh38
3825607 c.189G>A p.W63* Chr.4: 185418664-185418664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781397551 Chr.4: 185418769-185418769 on Build GRCh38
rs1018830770 Chr.4: 185418921-185418921 on Build GRCh38
rs1045574868 Chr.4: 185418975-185418975 on Build GRCh38
rs751595382 Chr.4: 185418759-185418759 on Build GRCh38
rs1014355758 Chr.4: 185418664-185418664 on Build GRCh38
rs755443922 Chr.4: 185418680-185418680 on Build GRCh38
rs977084252 Chr.4: 185418903-185418903 on Build GRCh38
rs142706569 Chr.4: 185418667-185418667 on Build GRCh38
rs373332194 Chr.4: 185418706-185418706 on Build GRCh38
rs915126533 Chr.4: 185418958-185418958 on Build GRCh38
rs922332164 Chr.4: 185418861-185418861 on Build GRCh38
rs773298065 Chr.4: 185418657-185418657 on Build GRCh38
rs760647426 Chr.4: 185418744-185418744 on Build GRCh38
rs778417875 Chr.4: 185418800-185418800 on Build GRCh38
rs1008948141 Chr.4: 185418909-185418909 on Build GRCh38
rs753659775 Chr.4: 185418749-185418749 on Build GRCh38
rs953454413 Chr.4: 185418891-185418891 on Build GRCh38
rs753621426 Chr.4: 185418794-185418798 on Build GRCh38
rs766511674 Chr.4: 185418758-185418758 on Build GRCh38
rs935936813 Chr.4: 185418920-185418920 on Build GRCh38
rs771672837 Chr.4: 185418734-185418734 on Build GRCh38
rs773610804 Chr.4: 185419033-185419033 on Build GRCh38
rs967969002 Chr.4: 185419067-185419067 on Build GRCh38
rs1028973696 Chr.4: 185418859-185418859 on Build GRCh38
rs758982893 Chr.4: 185418727-185418727 on Build GRCh38
rs993574859 Chr.4: 185419064-185419064 on Build GRCh38
rs764862996 Chr.4: 185418820-185418823 on Build GRCh38
rs760594879 Chr.4: 185418742-185418743 on Build GRCh38
rs751624574 Chr.4: 185418674-185418674 on Build GRCh38
rs772422498 Chr.4: 185418704-185418704 on Build GRCh38
rs376629977 Chr.4: 185418723-185418723 on Build GRCh38
rs756732713 Chr.4: 185418777-185418777 on Build GRCh38
rs116637007 Chr.4: 185418841-185418841 on Build GRCh38
rs770560758 Chr.4: 185418785-185418785 on Build GRCh38
rs535517408 Chr.4: 185418862-185418862 on Build GRCh38
rs988105130 Chr.4: 185418937-185418937 on Build GRCh38
rs753222930 Chr.4: 185418770-185418770 on Build GRCh38
rs745602824 Chr.4: 185418808-185418808 on Build GRCh38
rs778321208 Chr.4: 185418705-185418705 on Build GRCh38
rs966146313 Chr.4: 185418793-185418793 on Build GRCh38
rs773871032 Chr.4: 185418714-185418714 on Build GRCh38
rs763858649 Chr.4: 185418745-185418745 on Build GRCh38
rs547373836 Chr.4: 185418668-185418668 on Build GRCh38
rs754997381 Chr.4: 185418768-185418768 on Build GRCh38
rs956738728 Chr.4: 185418935-185418935 on Build GRCh38
rs778234928 Chr.4: 185418780-185418780 on Build GRCh38
rs770511201 Chr.4: 185418712-185418712 on Build GRCh38
rs949841670 Chr.4: 185418952-185418952 on Build GRCh38
rs763944488 Chr.4: 185418655-185418655 on Build GRCh38
rs757083211 Chr.4: 185418802-185418802 on Build GRCh38
rs370588942 Chr.4: 185418998-185418998 on Build GRCh38
rs748719187 Chr.4: 185418700-185418700 on Build GRCh38
rs549916890 Chr.4: 185418754-185418754 on Build GRCh38
rs781562054 Chr.4: 185418699-185418699 on Build GRCh38
rs967082283 Chr.4: 185418887-185418889 on Build GRCh38
rs749700475 Chr.4: 185418783-185418783 on Build GRCh38
rs760642634 Chr.4: 185418742-185418742 on Build GRCh38
rs993522568 Chr.4: 185418684-185418684 on Build GRCh38
rs995227882 Chr.4: 185418854-185418854 on Build GRCh38
rs151018748 Chr.4: 185418670-185418670 on Build GRCh38
rs763972449 Chr.4: 185418760-185418760 on Build GRCh38
rs897943642 Chr.4: 185419006-185419006 on Build GRCh38
rs915759386 Chr.4: 185418944-185418944 on Build GRCh38

View Full Product Details