Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89932180-89932763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH2
Entrez Gene ID
60468
Gene Name
BTB domain and CNC homolog 2
Gene Aliases
BTBD25
UniGene
Hs.269764
Gene Chromosome Location
Chr.6: 89926528-90296912 on Build GRCh38
Transcripts
NM_001170794.1 NM_021813.3 XM_005248758.4 XM_005248759.4 XM_011536039.2 XM_011536040.2 XM_017011156.1 XM_017011157.1 XM_017011158.1 XM_017011159.1 XM_017011160.1 XM_017011161.1 XM_017011162.1 XM_017011163.1 XM_017011164.1 XM_017011165.1 XM_017011166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7135770 c.2482G>A p.D828N Chr.6: 89932452-89932452 on Build GRCh38
5871242 c.2311G>A p.E771K Chr.6: 89932623-89932623 on Build GRCh38
6495735 c.2272T>C p.S758P Chr.6: 89932662-89932662 on Build GRCh38
5424492 c.2264T>C p.I755T Chr.6: 89932670-89932670 on Build GRCh38
5886450 c.2338C>T p.P780S Chr.6: 89932596-89932596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754280254 Chr.6: 89932470-89932470 on Build GRCh38
rs368416831 Chr.6: 89932619-89932619 on Build GRCh38
rs202088643 Chr.6: 89932402-89932402 on Build GRCh38
rs754299207 Chr.6: 89932568-89932568 on Build GRCh38
rs776040980 Chr.6: 89932666-89932666 on Build GRCh38
rs140270440 Chr.6: 89932694-89932694 on Build GRCh38
rs979952034 Chr.6: 89932268-89932268 on Build GRCh38
rs756574688 Chr.6: 89932444-89932444 on Build GRCh38
rs200514428 Chr.6: 89932322-89932322 on Build GRCh38
rs1053918340 Chr.6: 89932300-89932300 on Build GRCh38
rs867357454 Chr.6: 89932263-89932263 on Build GRCh38
rs878885096 Chr.6: 89932715-89932715 on Build GRCh38
rs767049051 Chr.6: 89932541-89932541 on Build GRCh38
rs73752855 Chr.6: 89932388-89932388 on Build GRCh38
rs764721711 Chr.6: 89932550-89932550 on Build GRCh38
rs757652995 Chr.6: 89932572-89932572 on Build GRCh38
rs1003204059 Chr.6: 89932337-89932337 on Build GRCh38
rs768610688 Chr.6: 89932602-89932602 on Build GRCh38
rs970250578 Chr.6: 89932264-89932264 on Build GRCh38
rs373437222 Chr.6: 89932531-89932531 on Build GRCh38
rs988665256 Chr.6: 89932291-89932291 on Build GRCh38
rs147787294 Chr.6: 89932743-89932743 on Build GRCh38
rs374620073 Chr.6: 89932653-89932653 on Build GRCh38
rs915321630 Chr.6: 89932657-89932657 on Build GRCh38
rs943484662 Chr.6: 89932334-89932334 on Build GRCh38
rs757186928 Chr.6: 89932648-89932648 on Build GRCh38
rs372214373 Chr.6: 89932727-89932727 on Build GRCh38
rs747070206 Chr.6: 89932445-89932445 on Build GRCh38
rs761793535 Chr.6: 89932391-89932391 on Build GRCh38
rs139170041 Chr.6: 89932635-89932635 on Build GRCh38
rs760300804 Chr.6: 89932614-89932614 on Build GRCh38
rs944032756 Chr.6: 89932259-89932259 on Build GRCh38
rs968647499 Chr.6: 89932523-89932523 on Build GRCh38
rs945346652 Chr.6: 89932314-89932314 on Build GRCh38
rs886691640 Chr.6: 89932390-89932390 on Build GRCh38
rs145487290 Chr.6: 89932247-89932248 on Build GRCh38
rs899906226 Chr.6: 89932277-89932277 on Build GRCh38
rs763412318 Chr.6: 89932670-89932670 on Build GRCh38
rs141231678 Chr.6: 89932612-89932612 on Build GRCh38
rs759143305 Chr.6: 89932534-89932534 on Build GRCh38
rs866820400 Chr.6: 89932738-89932738 on Build GRCh38
rs147649326 Chr.6: 89932693-89932693 on Build GRCh38
rs746028382 Chr.6: 89932483-89932483 on Build GRCh38
rs188927020 Chr.6: 89932597-89932597 on Build GRCh38
rs755611605 Chr.6: 89932602-89932603 on Build GRCh38
rs201308047 Chr.6: 89932380-89932380 on Build GRCh38
rs768240230 Chr.6: 89932665-89932665 on Build GRCh38
rs879226832 Chr.6: 89932593-89932593 on Build GRCh38
rs996940769 Chr.6: 89932297-89932297 on Build GRCh38
rs750863282 Chr.6: 89932621-89932621 on Build GRCh38
rs201359587 Chr.6: 89932532-89932532 on Build GRCh38
rs139627462 Chr.6: 89932603-89932603 on Build GRCh38
rs903936235 Chr.6: 89932332-89932332 on Build GRCh38
rs200166094 Chr.6: 89932398-89932398 on Build GRCh38
rs754468247 Chr.6: 89932362-89932362 on Build GRCh38
rs150771521 Chr.6: 89932615-89932615 on Build GRCh38
rs773007535 Chr.6: 89932530-89932530 on Build GRCh38
rs375103380 Chr.6: 89932613-89932613 on Build GRCh38
rs1021189022 Chr.6: 89932333-89932333 on Build GRCh38
rs746784202 Chr.6: 89932744-89932744 on Build GRCh38
rs766772225 Chr.6: 89932626-89932626 on Build GRCh38
rs765807351 Chr.6: 89932620-89932620 on Build GRCh38
rs147908828 Chr.6: 89932714-89932714 on Build GRCh38
rs746009967 Chr.6: 89932384-89932384 on Build GRCh38
rs767126356 Chr.6: 89932371-89932371 on Build GRCh38
rs980283280 Chr.6: 89932449-89932449 on Build GRCh38
rs750705861 Chr.6: 89932596-89932596 on Build GRCh38
rs1020109921 Chr.6: 89932656-89932656 on Build GRCh38
rs777216582 Chr.6: 89932616-89932616 on Build GRCh38
rs142254303 Chr.6: 89932476-89932476 on Build GRCh38
rs748305367 Chr.6: 89932690-89932690 on Build GRCh38
rs758209574 Chr.6: 89932732-89932732 on Build GRCh38
rs772300752 Chr.6: 89932489-89932489 on Build GRCh38
rs536262907 Chr.6: 89932435-89932435 on Build GRCh38
rs895424140 Chr.6: 89932729-89932729 on Build GRCh38
rs745724297 Chr.6: 89932651-89932651 on Build GRCh38
rs747141129 Chr.6: 89932601-89932601 on Build GRCh38
rs372285524 Chr.6: 89932624-89932624 on Build GRCh38
rs752773442 Chr.6: 89932705-89932705 on Build GRCh38
rs780242149 Chr.6: 89932496-89932496 on Build GRCh38
rs771225359 Chr.6: 89932383-89932383 on Build GRCh38
rs781715671 Chr.6: 89932363-89932363 on Build GRCh38
rs767748883 Chr.6: 89932699-89932699 on Build GRCh38
rs925306997 Chr.6: 89932536-89932536 on Build GRCh38
rs984461186 Chr.6: 89932242-89932242 on Build GRCh38
rs749282554 Chr.6: 89932645-89932645 on Build GRCh38
rs1033773955 Chr.6: 89932345-89932345 on Build GRCh38
rs148726175 Chr.6: 89932378-89932378 on Build GRCh38
rs761363627 Chr.6: 89932667-89932667 on Build GRCh38
rs748591360 Chr.6: 89932377-89932377 on Build GRCh38
rs577012189 Chr.6: 89932561-89932561 on Build GRCh38
rs1044120934 Chr.6: 89932323-89932323 on Build GRCh38
rs1042523147 Chr.6: 89932681-89932681 on Build GRCh38
rs751211841 Chr.6: 89932412-89932412 on Build GRCh38
rs369688322 Chr.6: 89932401-89932401 on Build GRCh38
rs778154893 Chr.6: 89932379-89932379 on Build GRCh38
rs745740912 Chr.6: 89932728-89932728 on Build GRCh38
rs368652670 Chr.6: 89932654-89932654 on Build GRCh38
rs371078442 Chr.6: 89932548-89932548 on Build GRCh38
rs1047650464 Chr.6: 89932249-89932249 on Build GRCh38
rs769816700 Chr.6: 89932526-89932526 on Build GRCh38
rs531936987 Chr.6: 89932327-89932327 on Build GRCh38
rs866952396 Chr.6: 89932287-89932287 on Build GRCh38
rs561647724 Chr.6: 89932222-89932222 on Build GRCh38
rs938583026 Chr.6: 89932288-89932288 on Build GRCh38
rs766794727 Chr.6: 89932688-89932688 on Build GRCh38
rs199646582 Chr.6: 89932607-89932607 on Build GRCh38
rs778111753 Chr.6: 89932450-89932450 on Build GRCh38
rs769503769 Chr.6: 89932396-89932396 on Build GRCh38
rs932699219 Chr.6: 89932422-89932422 on Build GRCh38
rs141976854 Chr.6: 89932516-89932516 on Build GRCh38
rs772255557 Chr.6: 89932256-89932256 on Build GRCh38
rs913165030 Chr.6: 89932309-89932309 on Build GRCh38
rs766960741 Chr.6: 89932434-89932434 on Build GRCh38
rs965034801 Chr.6: 89932707-89932707 on Build GRCh38
rs3798789 Chr.6: 89932639-89932639 on Build GRCh38
rs773849871 Chr.6: 89932669-89932669 on Build GRCh38
rs755100835 Chr.6: 89932638-89932638 on Build GRCh38
rs138800850 Chr.6: 89932477-89932477 on Build GRCh38
rs759055183 Chr.6: 89932425-89932425 on Build GRCh38
rs200505776 Chr.6: 89932600-89932600 on Build GRCh38
rs867412754 Chr.6: 89932605-89932605 on Build GRCh38
rs973178409 Chr.6: 89932239-89932239 on Build GRCh38
rs749989388 Chr.6: 89932453-89932454 on Build GRCh38
rs78688236 Chr.6: 89932573-89932573 on Build GRCh38
rs200671083 Chr.6: 89932608-89932608 on Build GRCh38

View Full Product Details