Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138062099-138062697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZIP1L
Entrez Gene ID
199221
Gene Name
DAZ interacting zinc finger protein 1 like
Gene Aliases
DZIP2
UniGene
Hs.351403
Gene Chromosome Location
Chr.3: 138061985-138115862 on Build GRCh38
Transcripts
NM_173543.2 XM_005247198.3

SNP Detail

SNP ID Chromosome Location
rs980254312 Chr.3: 138062249-138062249 on Build GRCh38
rs555189321 Chr.3: 138062300-138062300 on Build GRCh38
rs541738097 Chr.3: 138062450-138062450 on Build GRCh38
rs955629984 Chr.3: 138062562-138062562 on Build GRCh38
rs907870722 Chr.3: 138062343-138062343 on Build GRCh38
rs1056654643 Chr.3: 138062365-138062365 on Build GRCh38
rs1008427068 Chr.3: 138062151-138062151 on Build GRCh38
rs866868521 Chr.3: 138062547-138062547 on Build GRCh38
rs768765853 Chr.3: 138062196-138062196 on Build GRCh38
rs1019287639 Chr.3: 138062166-138062166 on Build GRCh38
rs561953016 Chr.3: 138062478-138062478 on Build GRCh38
rs775426386 Chr.3: 138062513-138062513 on Build GRCh38
rs1021301245 Chr.3: 138062635-138062635 on Build GRCh38
rs772247063 Chr.3: 138062295-138062295 on Build GRCh38
rs189849921 Chr.3: 138062182-138062182 on Build GRCh38
rs908577793 Chr.3: 138062305-138062305 on Build GRCh38
rs566039419 Chr.3: 138062253-138062253 on Build GRCh38
rs964177242 Chr.3: 138062191-138062191 on Build GRCh38
rs557495548 Chr.3: 138062652-138062653 on Build GRCh38
rs974059538 Chr.3: 138062636-138062636 on Build GRCh38
rs999864189 Chr.3: 138062456-138062456 on Build GRCh38
rs748203600 Chr.3: 138062246-138062246 on Build GRCh38
rs1007958915 Chr.3: 138062530-138062530 on Build GRCh38
rs532429109 Chr.3: 138062674-138062674 on Build GRCh38
rs749454852 Chr.3: 138062136-138062136 on Build GRCh38
rs964006767 Chr.3: 138062494-138062494 on Build GRCh38
rs982727681 Chr.3: 138062302-138062302 on Build GRCh38
rs938779211 Chr.3: 138062311-138062311 on Build GRCh38
rs114147197 Chr.3: 138062172-138062172 on Build GRCh38
rs982869796 Chr.3: 138062564-138062564 on Build GRCh38
rs1014035837 Chr.3: 138062395-138062395 on Build GRCh38
rs151311173 Chr.3: 138062599-138062599 on Build GRCh38
rs931231429 Chr.3: 138062427-138062427 on Build GRCh38
rs140556026 Chr.3: 138062602-138062602 on Build GRCh38
rs574910856 Chr.3: 138062338-138062338 on Build GRCh38
rs2291195 Chr.3: 138062407-138062407 on Build GRCh38
rs141049999 Chr.3: 138062364-138062364 on Build GRCh38
rs34429764 Chr.3: 138062315-138062315 on Build GRCh38
rs997059929 Chr.3: 138062503-138062503 on Build GRCh38
rs147741948 Chr.3: 138062339-138062339 on Build GRCh38
rs941613405 Chr.3: 138062321-138062321 on Build GRCh38
rs569818046 Chr.3: 138062663-138062663 on Build GRCh38
rs184591456 Chr.3: 138062130-138062130 on Build GRCh38
rs72991157 Chr.3: 138062199-138062199 on Build GRCh38
rs377568656 Chr.3: 138062382-138062382 on Build GRCh38
rs538670414 Chr.3: 138062194-138062194 on Build GRCh38
rs889510876 Chr.3: 138062150-138062150 on Build GRCh38
rs762651521 Chr.3: 138062558-138062558 on Build GRCh38
rs534974012 Chr.3: 138062255-138062255 on Build GRCh38
rs926846335 Chr.3: 138062661-138062661 on Build GRCh38
rs59771921 Chr.3: 138062351-138062351 on Build GRCh38
rs570106296 Chr.3: 138062429-138062429 on Build GRCh38
rs528442336 Chr.3: 138062440-138062440 on Build GRCh38
rs759616017 Chr.3: 138062441-138062441 on Build GRCh38

View Full Product Details