Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5863423-5864012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf196
Entrez Gene ID
149840
Gene Name
chromosome 20 open reading frame 196
Gene Aliases
-
UniGene
Hs.529340
Gene Chromosome Location
Chr.20: 5750286-5864407 on Build GRCh38
Transcripts
NM_001303477.1 NM_001303478.1 NM_152504.3 XM_006723549.3 XM_011529177.2 XM_017027684.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
362116 c.607A>T p.K203* Chr.20: 5863452-5863452 on Build GRCh38
4919700 c.613C>G p.L205V Chr.20: 5863458-5863458 on Build GRCh38
4426944 c.588C>T p.L196L Chr.20: 5863433-5863433 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1002828392 Chr.20: 5863898-5863898 on Build GRCh38
rs749674516 Chr.20: 5863485-5863485 on Build GRCh38
rs912696976 Chr.20: 5863566-5863566 on Build GRCh38
rs535029440 Chr.20: 5863764-5863764 on Build GRCh38
rs918630302 Chr.20: 5863606-5863606 on Build GRCh38
rs769936729 Chr.20: 5863926-5863926 on Build GRCh38
rs376423049 Chr.20: 5863666-5863666 on Build GRCh38
rs371846885 Chr.20: 5863962-5863962 on Build GRCh38
rs965126322 Chr.20: 5863619-5863619 on Build GRCh38
rs939902556 Chr.20: 5863701-5863707 on Build GRCh38
rs965534636 Chr.20: 5863930-5863930 on Build GRCh38
rs1021267539 Chr.20: 5863982-5863982 on Build GRCh38
rs932344887 Chr.20: 5863669-5863669 on Build GRCh38
rs755925999 Chr.20: 5863985-5863985 on Build GRCh38
rs560882274 Chr.20: 5863680-5863680 on Build GRCh38
rs972956443 Chr.20: 5863585-5863585 on Build GRCh38
rs752733611 Chr.20: 5863577-5863577 on Build GRCh38
rs1699232 Chr.20: 5863475-5863475 on Build GRCh38
rs906993226 Chr.20: 5863883-5863883 on Build GRCh38
rs144897306 Chr.20: 5863706-5863706 on Build GRCh38
rs1011639796 Chr.20: 5863971-5863971 on Build GRCh38
rs374958074 Chr.20: 5863472-5863472 on Build GRCh38
rs781155387 Chr.20: 5863721-5863721 on Build GRCh38
rs954259585 Chr.20: 5863523-5863523 on Build GRCh38
rs1777353 Chr.20: 5863476-5863476 on Build GRCh38
rs1036895266 Chr.20: 5863702-5863702 on Build GRCh38
rs141147152 Chr.20: 5863471-5863471 on Build GRCh38
rs1053016028 Chr.20: 5863916-5863916 on Build GRCh38
rs386812125 Chr.20: 5863475-5863476 on Build GRCh38
rs948131707 Chr.20: 5863776-5863776 on Build GRCh38
rs377580419 Chr.20: 5863459-5863459 on Build GRCh38
rs921058549 Chr.20: 5863961-5863961 on Build GRCh38
rs895665786 Chr.20: 5863705-5863705 on Build GRCh38
rs909842508 Chr.20: 5863687-5863687 on Build GRCh38
rs1046479051 Chr.20: 5863716-5863716 on Build GRCh38
rs772812366 Chr.20: 5863451-5863451 on Build GRCh38
rs372204786 Chr.20: 5863505-5863505 on Build GRCh38
rs536559610 Chr.20: 5863670-5863670 on Build GRCh38
rs779084191 Chr.20: 5863492-5863492 on Build GRCh38
rs766103830 Chr.20: 5863457-5863457 on Build GRCh38
rs985610723 Chr.20: 5863528-5863528 on Build GRCh38
rs760429954 Chr.20: 5863455-5863455 on Build GRCh38
rs1044257493 Chr.20: 5863851-5863851 on Build GRCh38
rs920988023 Chr.20: 5863655-5863655 on Build GRCh38
rs988307653 Chr.20: 5863525-5863525 on Build GRCh38
rs1013659039 Chr.20: 5863458-5863458 on Build GRCh38
rs1002242781 Chr.20: 5863880-5863880 on Build GRCh38
rs769080325 Chr.20: 5863489-5863489 on Build GRCh38
rs781280643 Chr.20: 5863474-5863474 on Build GRCh38
rs1032301782 Chr.20: 5863898-5863898 on Build GRCh38
rs765142480 Chr.20: 5863470-5863470 on Build GRCh38
rs144988084 Chr.20: 5863467-5863467 on Build GRCh38
rs1009759928 Chr.20: 5863908-5863908 on Build GRCh38
rs564881956 Chr.20: 5863775-5863775 on Build GRCh38
rs1699231 Chr.20: 5863947-5863947 on Build GRCh38
rs905240987 Chr.20: 5863791-5863791 on Build GRCh38
rs370101802 Chr.20: 5863620-5863620 on Build GRCh38

View Full Product Details