Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 35683780-35684358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
UniGene
Hs.475902
Gene Chromosome Location
Chr.3: 35638981-35798933 on Build GRCh38
Transcripts
NM_001025068.1 NM_001025069.1 NM_001267616.1 NM_001267617.1 NM_001267618.1 NM_001267619.1 NM_016300.4 NM_198399.1 XM_005264811.2 XM_006712943.2 XM_006712944.2 XM_011533298.2 XM_011533299.2 XM_011533300.2 XM_011533301.2 XM_011533302.2 XM_011533303.2 XM_017005574.1 XM_017005575.1 XM_017005576.1 XM_017005577.1 XM_017005578.1 XM_017005579.1 XM_017005580.1 XM_017005581.1 XM_017005582.1 XM_017005583.1 XM_017005584.1 XM_017005585.1 XM_017005586.1 XM_017005587.1 XM_017005588.1 XM_017005589.1 XM_017005590.1 XM_017005591.1 XM_017005592.1 XM_017005593.1 XM_017005594.1 XM_017005595.1 XM_017005596.1 XM_017005597.1 XM_017005598.1 XM_017005599.1 XM_017005600.1 XM_017005601.1 XM_017005602.1 XM_017005603.1 XM_017005604.1 XM_017005605.1 XM_017005606.1 XM_017005607.1 XM_017005608.1 XM_017005609.1 XM_017005610.1 XM_017005611.1 XM_017005612.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5179547 c.233G>T p.R78I Chr.3: 35683787-35683787 on Build GRCh38
4323821 c.250C>T p.L84F Chr.3: 35683804-35683804 on Build GRCh38
221990 c.244G>A p.E82K Chr.3: 35683798-35683798 on Build GRCh38
3592560 c.227C>T p.S76F Chr.3: 35683781-35683781 on Build GRCh38
3660493 c.247A>G p.S83G Chr.3: 35683801-35683801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779599344 Chr.3: 35683896-35683896 on Build GRCh38
rs1026010579 Chr.3: 35684271-35684271 on Build GRCh38
rs768926671 Chr.3: 35683983-35683983 on Build GRCh38
rs1008270792 Chr.3: 35684326-35684326 on Build GRCh38
rs984737521 Chr.3: 35684084-35684084 on Build GRCh38
rs770912517 Chr.3: 35683830-35683830 on Build GRCh38
rs762730939 Chr.3: 35683821-35683821 on Build GRCh38
rs774237174 Chr.3: 35684003-35684003 on Build GRCh38
rs1053140740 Chr.3: 35684136-35684136 on Build GRCh38
rs922376294 Chr.3: 35684159-35684159 on Build GRCh38
rs1045565316 Chr.3: 35684307-35684307 on Build GRCh38
rs1034198686 Chr.3: 35684220-35684220 on Build GRCh38
rs753405443 Chr.3: 35683885-35683885 on Build GRCh38
rs894338649 Chr.3: 35684267-35684267 on Build GRCh38
rs906965839 Chr.3: 35684168-35684168 on Build GRCh38
rs575577283 Chr.3: 35683877-35683877 on Build GRCh38
rs951669212 Chr.3: 35684061-35684061 on Build GRCh38
rs962664228 Chr.3: 35683902-35683902 on Build GRCh38
rs774083637 Chr.3: 35683838-35683838 on Build GRCh38
rs370988378 Chr.3: 35683872-35683872 on Build GRCh38
rs1045372142 Chr.3: 35684162-35684162 on Build GRCh38
rs974365012 Chr.3: 35683909-35683909 on Build GRCh38
rs750952897 Chr.3: 35684076-35684076 on Build GRCh38
rs975160323 Chr.3: 35684146-35684146 on Build GRCh38
rs945752739 Chr.3: 35684152-35684152 on Build GRCh38
rs201010760 Chr.3: 35683833-35683833 on Build GRCh38
rs771839344 Chr.3: 35684022-35684022 on Build GRCh38
rs986785468 Chr.3: 35683949-35683949 on Build GRCh38
rs1001204445 Chr.3: 35684217-35684217 on Build GRCh38
rs906955786 Chr.3: 35683862-35683862 on Build GRCh38
rs1025660011 Chr.3: 35683913-35683913 on Build GRCh38
rs757658724 Chr.3: 35683891-35683891 on Build GRCh38
rs563559929 Chr.3: 35683874-35683874 on Build GRCh38
rs568777665 Chr.3: 35684279-35684279 on Build GRCh38
rs551995518 Chr.3: 35684268-35684268 on Build GRCh38
rs150230446 Chr.3: 35684230-35684230 on Build GRCh38
rs747284341 Chr.3: 35683802-35683802 on Build GRCh38
rs371102133 Chr.3: 35683988-35683988 on Build GRCh38
rs542975560 Chr.3: 35683939-35683939 on Build GRCh38
rs906628064 Chr.3: 35684310-35684310 on Build GRCh38
rs183848724 Chr.3: 35683985-35683985 on Build GRCh38
rs1041393013 Chr.3: 35684229-35684229 on Build GRCh38
rs775328452 Chr.3: 35684042-35684042 on Build GRCh38
rs532139265 Chr.3: 35684169-35684169 on Build GRCh38
rs759357576 Chr.3: 35683842-35683842 on Build GRCh38
rs1054773507 Chr.3: 35684261-35684261 on Build GRCh38
rs565138148 Chr.3: 35684088-35684088 on Build GRCh38
rs34709978 Chr.3: 35684319-35684320 on Build GRCh38
rs760191172 Chr.3: 35684048-35684048 on Build GRCh38
rs200173509 Chr.3: 35683972-35683972 on Build GRCh38
rs759515161 Chr.3: 35684013-35684013 on Build GRCh38
rs764889992 Chr.3: 35684071-35684071 on Build GRCh38
rs777420967 Chr.3: 35683936-35683937 on Build GRCh38
rs747193852 Chr.3: 35683982-35683982 on Build GRCh38
rs936957054 Chr.3: 35684116-35684116 on Build GRCh38
rs1001364478 Chr.3: 35683865-35683865 on Build GRCh38
rs780624748 Chr.3: 35683981-35683981 on Build GRCh38
rs776838616 Chr.3: 35683810-35683810 on Build GRCh38
rs1023713874 Chr.3: 35684043-35684043 on Build GRCh38
rs951483064 Chr.3: 35683935-35683935 on Build GRCh38
rs763832567 Chr.3: 35684054-35684054 on Build GRCh38
rs767336974 Chr.3: 35683860-35683860 on Build GRCh38
rs753251836 Chr.3: 35684056-35684056 on Build GRCh38
rs951322474 Chr.3: 35684312-35684312 on Build GRCh38
rs369367281 Chr.3: 35684094-35684094 on Build GRCh38
rs912684687 Chr.3: 35683954-35683954 on Build GRCh38
rs1012372746 Chr.3: 35683946-35683946 on Build GRCh38
rs898297287 Chr.3: 35684249-35684249 on Build GRCh38
rs187954667 Chr.3: 35684295-35684295 on Build GRCh38
rs892707327 Chr.3: 35684138-35684138 on Build GRCh38
rs935972724 Chr.3: 35684237-35684237 on Build GRCh38

View Full Product Details