Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27329044-27329610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C2
Entrez Gene ID
2976
Gene Name
general transcription factor IIIC subunit 2
Gene Aliases
TFIIIC-BETA, TFIIIC110
UniGene
Hs.745168
Gene Chromosome Location
Chr.2: 27325849-27357034 on Build GRCh38
Transcripts
NM_001035521.2 NM_001318909.1 NM_001521.3 XM_005264272.4 XM_011532801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
721437 c.1790C>G p.S597C Chr.2: 27329466-27329466 on Build GRCh38
109676 c.1788C>T p.L596L Chr.2: 27329468-27329468 on Build GRCh38
6334120 c.1941C>A p.Y647* Chr.2: 27329219-27329219 on Build GRCh38
3839276 c.1774C>T p.Q592* Chr.2: 27329482-27329482 on Build GRCh38
6298477 c.1769C>A p.P590H Chr.2: 27329487-27329487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896537055 Chr.2: 27329207-27329207 on Build GRCh38
rs747775019 Chr.2: 27329227-27329227 on Build GRCh38
rs778809914 Chr.2: 27329342-27329342 on Build GRCh38
rs775287186 Chr.2: 27329383-27329383 on Build GRCh38
rs551596395 Chr.2: 27329149-27329149 on Build GRCh38
rs776853502 Chr.2: 27329299-27329299 on Build GRCh38
rs984477002 Chr.2: 27329586-27329586 on Build GRCh38
rs763682355 Chr.2: 27329571-27329571 on Build GRCh38
rs764005582 Chr.2: 27329403-27329403 on Build GRCh38
rs144790568 Chr.2: 27329478-27329478 on Build GRCh38
rs572927438 Chr.2: 27329349-27329349 on Build GRCh38
rs190251496 Chr.2: 27329444-27329444 on Build GRCh38
rs770660635 Chr.2: 27329132-27329132 on Build GRCh38
rs765304322 Chr.2: 27329199-27329199 on Build GRCh38
rs763423156 Chr.2: 27329315-27329315 on Build GRCh38
rs533881871 Chr.2: 27329356-27329356 on Build GRCh38
rs763797102 Chr.2: 27329458-27329458 on Build GRCh38
rs111964097 Chr.2: 27329363-27329363 on Build GRCh38
rs140039030 Chr.2: 27329504-27329504 on Build GRCh38
rs758285292 Chr.2: 27329479-27329479 on Build GRCh38
rs750901561 Chr.2: 27329203-27329203 on Build GRCh38
rs773208162 Chr.2: 27329372-27329372 on Build GRCh38
rs749203416 Chr.2: 27329276-27329276 on Build GRCh38
rs568136490 Chr.2: 27329246-27329246 on Build GRCh38
rs780902662 Chr.2: 27329076-27329076 on Build GRCh38
rs550971239 Chr.2: 27329294-27329294 on Build GRCh38
rs201499584 Chr.2: 27329278-27329278 on Build GRCh38
rs964522837 Chr.2: 27329347-27329347 on Build GRCh38
rs753985250 Chr.2: 27329160-27329160 on Build GRCh38
rs747652439 Chr.2: 27329098-27329098 on Build GRCh38
rs138720572 Chr.2: 27329219-27329219 on Build GRCh38
rs780974234 Chr.2: 27329226-27329226 on Build GRCh38
rs374911992 Chr.2: 27329083-27329083 on Build GRCh38
rs768378393 Chr.2: 27329070-27329070 on Build GRCh38
rs766504805 Chr.2: 27329414-27329414 on Build GRCh38
rs769824966 Chr.2: 27329115-27329115 on Build GRCh38
rs768035725 Chr.2: 27329557-27329557 on Build GRCh38
rs757252797 Chr.2: 27329329-27329329 on Build GRCh38
rs142984034 Chr.2: 27329480-27329480 on Build GRCh38
rs777195510 Chr.2: 27329324-27329324 on Build GRCh38
rs753347705 Chr.2: 27329464-27329464 on Build GRCh38
rs771162565 Chr.2: 27329516-27329516 on Build GRCh38
rs573699100 Chr.2: 27329472-27329472 on Build GRCh38
rs983210961 Chr.2: 27329456-27329456 on Build GRCh38
rs752023228 Chr.2: 27329286-27329287 on Build GRCh38
rs774521994 Chr.2: 27329532-27329532 on Build GRCh38
rs187061145 Chr.2: 27329322-27329322 on Build GRCh38
rs775000342 Chr.2: 27329426-27329426 on Build GRCh38
rs769981496 Chr.2: 27329500-27329500 on Build GRCh38
rs762029935 Chr.2: 27329305-27329305 on Build GRCh38
rs752023144 Chr.2: 27329317-27329317 on Build GRCh38
rs750470401 Chr.2: 27329313-27329313 on Build GRCh38
rs544832548 Chr.2: 27329561-27329561 on Build GRCh38
rs753082569 Chr.2: 27329446-27329446 on Build GRCh38
rs143400550 Chr.2: 27329204-27329204 on Build GRCh38
rs755761840 Chr.2: 27329455-27329455 on Build GRCh38
rs760078139 Chr.2: 27329428-27329428 on Build GRCh38
rs748799655 Chr.2: 27329327-27329327 on Build GRCh38
rs749451862 Chr.2: 27329510-27329510 on Build GRCh38
rs191702515 Chr.2: 27329150-27329150 on Build GRCh38
rs774171667 Chr.2: 27329287-27329287 on Build GRCh38
rs375695294 Chr.2: 27329528-27329528 on Build GRCh38
rs764269890 Chr.2: 27329157-27329157 on Build GRCh38
rs773034060 Chr.2: 27329117-27329117 on Build GRCh38
rs367917139 Chr.2: 27329288-27329288 on Build GRCh38
rs200063514 Chr.2: 27329126-27329126 on Build GRCh38
rs775317006 Chr.2: 27329355-27329355 on Build GRCh38
rs777296231 Chr.2: 27329258-27329258 on Build GRCh38
rs770037918 Chr.2: 27329365-27329365 on Build GRCh38
rs142646778 Chr.2: 27329468-27329468 on Build GRCh38
rs183210129 Chr.2: 27329196-27329196 on Build GRCh38
rs569568286 Chr.2: 27329321-27329321 on Build GRCh38
rs558376671 Chr.2: 27329323-27329323 on Build GRCh38
rs544220164 Chr.2: 27329473-27329473 on Build GRCh38
rs769189061 Chr.2: 27329234-27329234 on Build GRCh38
rs368162172 Chr.2: 27329541-27329541 on Build GRCh38
rs751953553 Chr.2: 27329218-27329218 on Build GRCh38
rs763088522 Chr.2: 27329406-27329406 on Build GRCh38

View Full Product Details