Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27326444-27327002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C2
Entrez Gene ID
2976
Gene Name
general transcription factor IIIC subunit 2
Gene Aliases
TFIIIC-BETA, TFIIIC110
UniGene
Hs.745168
Gene Chromosome Location
Chr.2: 27325849-27357034 on Build GRCh38
Transcripts
NM_001035521.2 NM_001318909.1 NM_001521.3 XM_005264272.4 XM_011532801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5497612 c.2600G>A p.R867H Chr.2: 27326811-27326811 on Build GRCh38
1019637 c.2649C>T p.A883A Chr.2: 27326762-27326762 on Build GRCh38
1019636 c.2690G>A p.R897Q Chr.2: 27326721-27326721 on Build GRCh38
3939052 c.2559A>G p.V853V Chr.2: 27326852-27326852 on Build GRCh38
5994184 c.2557G>A p.V853I Chr.2: 27326854-27326854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778908231 Chr.2: 27326666-27326666 on Build GRCh38
rs879013313 Chr.2: 27326595-27326595 on Build GRCh38
rs551930166 Chr.2: 27326482-27326482 on Build GRCh38
rs750397337 Chr.2: 27326604-27326604 on Build GRCh38
rs776421272 Chr.2: 27326699-27326699 on Build GRCh38
rs35978802 Chr.2: 27326879-27326880 on Build GRCh38
rs758996734 Chr.2: 27326735-27326735 on Build GRCh38
rs113469548 Chr.2: 27326966-27326967 on Build GRCh38
rs376505721 Chr.2: 27326700-27326700 on Build GRCh38
rs758792957 Chr.2: 27326683-27326683 on Build GRCh38
rs777015032 Chr.2: 27326915-27326916 on Build GRCh38
rs373604869 Chr.2: 27326922-27326922 on Build GRCh38
rs370776889 Chr.2: 27326914-27326914 on Build GRCh38
rs961551574 Chr.2: 27326686-27326686 on Build GRCh38
rs185853433 Chr.2: 27326968-27326968 on Build GRCh38
rs764268008 Chr.2: 27326655-27326655 on Build GRCh38
rs771956711 Chr.2: 27326945-27326945 on Build GRCh38
rs765321012 Chr.2: 27326601-27326601 on Build GRCh38
rs1049784839 Chr.2: 27326596-27326596 on Build GRCh38
rs35615796 Chr.2: 27326973-27326974 on Build GRCh38
rs878984154 Chr.2: 27326567-27326567 on Build GRCh38
rs397868275 Chr.2: 27326978-27326978 on Build GRCh38
rs747354601 Chr.2: 27326473-27326473 on Build GRCh38
rs766970414 Chr.2: 27326811-27326811 on Build GRCh38
rs193107013 Chr.2: 27326939-27326939 on Build GRCh38
rs187695337 Chr.2: 27326695-27326695 on Build GRCh38
rs944023169 Chr.2: 27326476-27326476 on Build GRCh38
rs397871054 Chr.2: 27326978-27326979 on Build GRCh38
rs770965884 Chr.2: 27326785-27326785 on Build GRCh38
rs756408181 Chr.2: 27326646-27326646 on Build GRCh38
rs34211481 Chr.2: 27326555-27326555 on Build GRCh38
rs138644187 Chr.2: 27326722-27326722 on Build GRCh38
rs759461209 Chr.2: 27326793-27326793 on Build GRCh38
rs753085863 Chr.2: 27326868-27326868 on Build GRCh38
rs373008992 Chr.2: 27326815-27326815 on Build GRCh38
rs976738421 Chr.2: 27326812-27326812 on Build GRCh38
rs775225951 Chr.2: 27326797-27326797 on Build GRCh38
rs1055946207 Chr.2: 27326959-27326959 on Build GRCh38
rs778420883 Chr.2: 27326930-27326930 on Build GRCh38
rs755871058 Chr.2: 27326890-27326890 on Build GRCh38
rs777287054 Chr.2: 27326903-27326903 on Build GRCh38
rs189095327 Chr.2: 27326977-27326977 on Build GRCh38
rs76943914 Chr.2: 27326967-27326967 on Build GRCh38
rs889823364 Chr.2: 27326643-27326643 on Build GRCh38
rs911830931 Chr.2: 27326784-27326784 on Build GRCh38
rs780396358 Chr.2: 27326748-27326748 on Build GRCh38
rs1011648741 Chr.2: 27326543-27326543 on Build GRCh38
rs534621889 Chr.2: 27326634-27326634 on Build GRCh38
rs752645430 Chr.2: 27326902-27326902 on Build GRCh38
rs911192675 Chr.2: 27326475-27326475 on Build GRCh38
rs762728750 Chr.2: 27326620-27326620 on Build GRCh38
rs781703987 Chr.2: 27326865-27326865 on Build GRCh38
rs780144842 Chr.2: 27326530-27326530 on Build GRCh38
rs767306443 Chr.2: 27326794-27326794 on Build GRCh38
rs771948035 Chr.2: 27326936-27326936 on Build GRCh38
rs767433437 Chr.2: 27326929-27326929 on Build GRCh38
rs1014524755 Chr.2: 27326973-27326973 on Build GRCh38
rs537451431 Chr.2: 27326969-27326969 on Build GRCh38
rs1049836 Chr.2: 27326467-27326467 on Build GRCh38
rs772804342 Chr.2: 27326701-27326701 on Build GRCh38
rs755500154 Chr.2: 27326855-27326855 on Build GRCh38
rs757317712 Chr.2: 27326664-27326664 on Build GRCh38
rs35921404 Chr.2: 27326496-27326496 on Build GRCh38
rs762940062 Chr.2: 27326806-27326806 on Build GRCh38
rs199644340 Chr.2: 27326846-27326846 on Build GRCh38
rs1041127572 Chr.2: 27326495-27326495 on Build GRCh38
rs774241650 Chr.2: 27326718-27326718 on Build GRCh38
rs766202890 Chr.2: 27326716-27326716 on Build GRCh38
rs181281564 Chr.2: 27326966-27326966 on Build GRCh38
rs368948527 Chr.2: 27326804-27326804 on Build GRCh38
rs377223288 Chr.2: 27326729-27326729 on Build GRCh38
rs116010390 Chr.2: 27326931-27326931 on Build GRCh38
rs374655370 Chr.2: 27326721-27326721 on Build GRCh38
rs747477056 Chr.2: 27326749-27326749 on Build GRCh38
rs369412955 Chr.2: 27326704-27326704 on Build GRCh38
rs781633359 Chr.2: 27326762-27326762 on Build GRCh38
rs1049831 Chr.2: 27326518-27326518 on Build GRCh38
rs1053675127 Chr.2: 27326498-27326498 on Build GRCh38
rs891900791 Chr.2: 27326499-27326499 on Build GRCh38
rs568205864 Chr.2: 27326914-27326915 on Build GRCh38
rs371595443 Chr.2: 27326698-27326698 on Build GRCh38
rs746391591 Chr.2: 27326679-27326679 on Build GRCh38
rs772827211 Chr.2: 27326776-27326776 on Build GRCh38
rs144448178 Chr.2: 27326754-27326754 on Build GRCh38
rs953343182 Chr.2: 27326732-27326732 on Build GRCh38
rs762045776 Chr.2: 27326801-27326801 on Build GRCh38
rs774140717 Chr.2: 27326792-27326792 on Build GRCh38
rs769518542 Chr.2: 27326766-27326766 on Build GRCh38
rs35568684 Chr.2: 27326967-27326967 on Build GRCh38
rs780492879 Chr.2: 27326693-27326693 on Build GRCh38
rs763681410 Chr.2: 27326914-27326917 on Build GRCh38
rs904730376 Chr.2: 27326653-27326653 on Build GRCh38
rs1043213966 Chr.2: 27326627-27326627 on Build GRCh38
rs147156644 Chr.2: 27326777-27326777 on Build GRCh38
rs750421174 Chr.2: 27326734-27326734 on Build GRCh38
rs765427783 Chr.2: 27326803-27326803 on Build GRCh38
rs768397410 Chr.2: 27326697-27326697 on Build GRCh38

View Full Product Details