Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134952474-134953010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT2
Entrez Gene ID
905
Gene Name
cyclin T2
Gene Aliases
CYCT2
UniGene
Hs.744115
Gene Chromosome Location
Chr.2: 134918810-134959345 on Build GRCh38
Transcripts
NM_001241.3 NM_001320748.1 NM_001320749.1 NM_058241.2 NR_037649.1 NR_135468.1 XM_017005227.1 XM_017005228.1 XM_017005229.1 XM_017005230.1 XM_017005231.1 XM_017005232.1 XM_017005233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203850 c.774+4A>C p.? Chr.2: 134952715-134952715 on Build GRCh38
256562 c.763C>T p.R255* Chr.2: 134952700-134952700 on Build GRCh38
4572366 c.722T>C p.L241P Chr.2: 134952659-134952659 on Build GRCh38
1669261 c.740C>T p.T247M Chr.2: 134952677-134952677 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758285647 Chr.2: 134952723-134952723 on Build GRCh38
rs928285531 Chr.2: 134952528-134952528 on Build GRCh38
rs777363438 Chr.2: 134952793-134952796 on Build GRCh38
rs999928092 Chr.2: 134952577-134952577 on Build GRCh38
rs147275367 Chr.2: 134952686-134952686 on Build GRCh38
rs752384251 Chr.2: 134952650-134952651 on Build GRCh38
rs45480696 Chr.2: 134952950-134952950 on Build GRCh38
rs889904735 Chr.2: 134952639-134952639 on Build GRCh38
rs752765528 Chr.2: 134952642-134952642 on Build GRCh38
rs749605663 Chr.2: 134952765-134952765 on Build GRCh38
rs182040038 Chr.2: 134952891-134952891 on Build GRCh38
rs1026466627 Chr.2: 134952850-134952850 on Build GRCh38
rs753614860 Chr.2: 134952863-134952864 on Build GRCh38
rs866242940 Chr.2: 134952504-134952504 on Build GRCh38
rs780095204 Chr.2: 134952727-134952727 on Build GRCh38
rs755157486 Chr.2: 134952848-134952848 on Build GRCh38
rs751549685 Chr.2: 134952729-134952729 on Build GRCh38
rs755696650 Chr.2: 134952748-134952748 on Build GRCh38
rs764350215 Chr.2: 134952806-134952806 on Build GRCh38
rs767291883 Chr.2: 134952616-134952616 on Build GRCh38
rs931099170 Chr.2: 134952583-134952583 on Build GRCh38
rs747672128 Chr.2: 134952737-134952737 on Build GRCh38
rs1042049503 Chr.2: 134952953-134952953 on Build GRCh38
rs1006965021 Chr.2: 134952684-134952684 on Build GRCh38
rs918540154 Chr.2: 134952943-134952943 on Build GRCh38
rs746362919 Chr.2: 134952781-134952781 on Build GRCh38
rs370012767 Chr.2: 134952736-134952736 on Build GRCh38
rs200922251 Chr.2: 134952750-134952750 on Build GRCh38
rs771369400 Chr.2: 134952591-134952591 on Build GRCh38
rs979852299 Chr.2: 134952907-134952907 on Build GRCh38
rs943333059 Chr.2: 134952900-134952900 on Build GRCh38
rs972767502 Chr.2: 134952842-134952842 on Build GRCh38
rs772509563 Chr.2: 134952791-134952791 on Build GRCh38
rs911042187 Chr.2: 134952978-134952978 on Build GRCh38
rs1036879706 Chr.2: 134952536-134952536 on Build GRCh38
rs1023791949 Chr.2: 134952828-134952828 on Build GRCh38
rs959664097 Chr.2: 134952914-134952914 on Build GRCh38
rs774617561 Chr.2: 134952771-134952771 on Build GRCh38
rs1041091638 Chr.2: 134952701-134952701 on Build GRCh38
rs767601859 Chr.2: 134952623-134952623 on Build GRCh38
rs938617554 Chr.2: 134952530-134952530 on Build GRCh38
rs774860077 Chr.2: 134952613-134952613 on Build GRCh38
rs189488016 Chr.2: 134952730-134952730 on Build GRCh38
rs752205781 Chr.2: 134952650-134952650 on Build GRCh38
rs759492346 Chr.2: 134952617-134952617 on Build GRCh38
rs201586820 Chr.2: 134952672-134952672 on Build GRCh38
rs1048219343 Chr.2: 134952620-134952620 on Build GRCh38
rs1012591753 Chr.2: 134952757-134952757 on Build GRCh38
rs976060687 Chr.2: 134952917-134952917 on Build GRCh38
rs935838972 Chr.2: 134952984-134952984 on Build GRCh38
rs952343775 Chr.2: 134952872-134952872 on Build GRCh38
rs760838432 Chr.2: 134952799-134952799 on Build GRCh38
rs896999230 Chr.2: 134952574-134952574 on Build GRCh38
rs984808816 Chr.2: 134952889-134952889 on Build GRCh38
rs901151981 Chr.2: 134952714-134952714 on Build GRCh38
rs760935418 Chr.2: 134952654-134952654 on Build GRCh38
rs530191070 Chr.2: 134952929-134952929 on Build GRCh38
rs10166142 Chr.2: 134952752-134952752 on Build GRCh38
rs764473101 Chr.2: 134952663-134952663 on Build GRCh38
rs777504580 Chr.2: 134952773-134952773 on Build GRCh38
rs570199708 Chr.2: 134952956-134952956 on Build GRCh38
rs563723884 Chr.2: 134952666-134952666 on Build GRCh38

View Full Product Details