Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23597014-23597551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC126
Entrez Gene ID
90693
Gene Name
coiled-coil domain containing 126
Gene Aliases
-
UniGene
Hs.232296
Gene Chromosome Location
Chr.7: 23597333-23644708 on Build GRCh38
Transcripts
NM_138771.3 XM_017012775.1

SNP Detail

SNP ID Chromosome Location
rs560099870 Chr.7: 23597253-23597253 on Build GRCh38
rs951213800 Chr.7: 23597402-23597402 on Build GRCh38
rs1002621309 Chr.7: 23597111-23597111 on Build GRCh38
rs915374867 Chr.7: 23597415-23597415 on Build GRCh38
rs1055410297 Chr.7: 23597461-23597461 on Build GRCh38
rs1019492026 Chr.7: 23597408-23597408 on Build GRCh38
rs990934791 Chr.7: 23597275-23597275 on Build GRCh38
rs565298504 Chr.7: 23597427-23597427 on Build GRCh38
rs1025684791 Chr.7: 23597081-23597081 on Build GRCh38
rs1046607297 Chr.7: 23597346-23597346 on Build GRCh38
rs899032971 Chr.7: 23597367-23597367 on Build GRCh38
rs190554533 Chr.7: 23597092-23597092 on Build GRCh38
rs886177060 Chr.7: 23597484-23597484 on Build GRCh38
rs567671973 Chr.7: 23597495-23597495 on Build GRCh38
rs919742966 Chr.7: 23597180-23597180 on Build GRCh38
rs920984186 Chr.7: 23597292-23597292 on Build GRCh38
rs1055111605 Chr.7: 23597072-23597072 on Build GRCh38
rs1047582191 Chr.7: 23597473-23597473 on Build GRCh38
rs1017876699 Chr.7: 23597220-23597220 on Build GRCh38
rs567635290 Chr.7: 23597053-23597053 on Build GRCh38
rs562888159 Chr.7: 23597300-23597300 on Build GRCh38
rs950364975 Chr.7: 23597390-23597390 on Build GRCh38
rs1033737634 Chr.7: 23597117-23597117 on Build GRCh38
rs994556235 Chr.7: 23597376-23597376 on Build GRCh38
rs1047017017 Chr.7: 23597429-23597429 on Build GRCh38
rs1026316407 Chr.7: 23597467-23597467 on Build GRCh38
rs965685513 Chr.7: 23597413-23597413 on Build GRCh38
rs994870281 Chr.7: 23597396-23597396 on Build GRCh38
rs1003280862 Chr.7: 23597485-23597485 on Build GRCh38
rs753639596 Chr.7: 23597118-23597118 on Build GRCh38
rs544423690 Chr.7: 23597166-23597166 on Build GRCh38
rs143275586 Chr.7: 23597449-23597449 on Build GRCh38
rs371455238 Chr.7: 23597212-23597212 on Build GRCh38
rs573263598 Chr.7: 23597271-23597271 on Build GRCh38
rs551981844 Chr.7: 23597421-23597421 on Build GRCh38
rs1007510560 Chr.7: 23597356-23597356 on Build GRCh38
rs965262763 Chr.7: 23597501-23597501 on Build GRCh38
rs1039365138 Chr.7: 23597462-23597462 on Build GRCh38
rs973853669 Chr.7: 23597179-23597179 on Build GRCh38
rs1038857732 Chr.7: 23597359-23597359 on Build GRCh38
rs899164231 Chr.7: 23597464-23597464 on Build GRCh38
rs1034647902 Chr.7: 23597490-23597490 on Build GRCh38
rs1003144552 Chr.7: 23597399-23597399 on Build GRCh38
rs1053556495 Chr.7: 23597301-23597301 on Build GRCh38
rs890262358 Chr.7: 23597211-23597211 on Build GRCh38
rs923932945 Chr.7: 23597303-23597303 on Build GRCh38
rs945533675 Chr.7: 23597428-23597428 on Build GRCh38
rs555659110 Chr.7: 23597137-23597137 on Build GRCh38
rs539065992 Chr.7: 23597221-23597221 on Build GRCh38
rs1002182108 Chr.7: 23597360-23597360 on Build GRCh38
rs1051655725 Chr.7: 23597354-23597355 on Build GRCh38
rs1009802923 Chr.7: 23597067-23597067 on Build GRCh38
rs899196729 Chr.7: 23597391-23597391 on Build GRCh38
rs539925583 Chr.7: 23597251-23597251 on Build GRCh38
rs547525924 Chr.7: 23597465-23597465 on Build GRCh38
rs945568299 Chr.7: 23597342-23597342 on Build GRCh38
rs749493593 Chr.7: 23597323-23597323 on Build GRCh38
rs542208250 Chr.7: 23597272-23597272 on Build GRCh38
rs936348389 Chr.7: 23597417-23597417 on Build GRCh38
rs557653615 Chr.7: 23597168-23597168 on Build GRCh38
rs892698008 Chr.7: 23597061-23597061 on Build GRCh38
rs928238381 Chr.7: 23597270-23597270 on Build GRCh38
rs890357167 Chr.7: 23597355-23597355 on Build GRCh38
rs920912458 Chr.7: 23597416-23597416 on Build GRCh38
rs938244119 Chr.7: 23597453-23597453 on Build GRCh38
rs951246648 Chr.7: 23597195-23597195 on Build GRCh38
rs531860512 Chr.7: 23597382-23597382 on Build GRCh38
rs575608311 Chr.7: 23597155-23597155 on Build GRCh38
rs886533708 Chr.7: 23597486-23597486 on Build GRCh38
rs1025977240 Chr.7: 23597224-23597224 on Build GRCh38
rs765111106 Chr.7: 23597422-23597422 on Build GRCh38
rs1026390437 Chr.7: 23597400-23597400 on Build GRCh38
rs768482215 Chr.7: 23597332-23597332 on Build GRCh38
rs775747766 Chr.7: 23597317-23597317 on Build GRCh38
rs975080231 Chr.7: 23597414-23597414 on Build GRCh38
rs1039784442 Chr.7: 23597290-23597290 on Build GRCh38
rs950389523 Chr.7: 23597240-23597240 on Build GRCh38
rs943264104 Chr.7: 23597459-23597459 on Build GRCh38
rs983024208 Chr.7: 23597405-23597405 on Build GRCh38
rs963443839 Chr.7: 23597119-23597119 on Build GRCh38
rs906789287 Chr.7: 23597349-23597349 on Build GRCh38
rs1004020297 Chr.7: 23597489-23597489 on Build GRCh38

View Full Product Details