Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 129748381-129748863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSDMC
Entrez Gene ID
56169
Gene Name
gasdermin C
Gene Aliases
MLZE
UniGene
Hs.133244
Gene Chromosome Location
Chr.8: 129705903-129786888 on Build GRCh38
Transcripts
NM_031415.2 XM_011517158.1 XM_011517159.1 XM_011517161.1 XM_011517162.1 XM_011517163.2 XM_011517165.1 XM_011517166.2 XM_017013644.1 XM_017013645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5591206 c.1377G>A p.E459E Chr.8: 129748651-129748651 on Build GRCh38
5798159 c.1320C>A p.Y440* Chr.8: 129748708-129748708 on Build GRCh38
4162669 c.1320C>T p.Y440Y Chr.8: 129748708-129748708 on Build GRCh38
150539 c.1424T>C p.M475T Chr.8: 129748604-129748604 on Build GRCh38
1488997 c.1442G>T p.R481M Chr.8: 129748586-129748586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767221256 Chr.8: 129748582-129748582 on Build GRCh38
rs760297354 Chr.8: 129748655-129748655 on Build GRCh38
rs754311687 Chr.8: 129748464-129748464 on Build GRCh38
rs778036535 Chr.8: 129748459-129748459 on Build GRCh38
rs4144738 Chr.8: 129748604-129748604 on Build GRCh38
rs1018262478 Chr.8: 129748764-129748764 on Build GRCh38
rs773424017 Chr.8: 129748546-129748546 on Build GRCh38
rs764585515 Chr.8: 129748642-129748642 on Build GRCh38
rs943573369 Chr.8: 129748656-129748656 on Build GRCh38
rs34385086 Chr.8: 129748708-129748708 on Build GRCh38
rs770936296 Chr.8: 129748548-129748548 on Build GRCh38
rs550396087 Chr.8: 129748733-129748733 on Build GRCh38
rs758152327 Chr.8: 129748701-129748701 on Build GRCh38
rs765875845 Chr.8: 129748760-129748760 on Build GRCh38
rs957108574 Chr.8: 129748806-129748806 on Build GRCh38
rs115019901 Chr.8: 129748556-129748556 on Build GRCh38
rs773282596 Chr.8: 129748554-129748554 on Build GRCh38
rs4144736 Chr.8: 129748784-129748784 on Build GRCh38
rs758381625 Chr.8: 129748783-129748783 on Build GRCh38
rs532245909 Chr.8: 129748730-129748730 on Build GRCh38
rs771010578 Chr.8: 129748629-129748629 on Build GRCh38
rs753360215 Chr.8: 129748668-129748668 on Build GRCh38
rs745314907 Chr.8: 129748484-129748484 on Build GRCh38
rs1047596087 Chr.8: 129748707-129748707 on Build GRCh38
rs757018901 Chr.8: 129748477-129748477 on Build GRCh38
rs780835315 Chr.8: 129748483-129748483 on Build GRCh38
rs765550055 Chr.8: 129748474-129748474 on Build GRCh38
rs562110298 Chr.8: 129748689-129748689 on Build GRCh38
rs116452429 Chr.8: 129748693-129748693 on Build GRCh38
rs373227533 Chr.8: 129748669-129748669 on Build GRCh38
rs776364654 Chr.8: 129748770-129748770 on Build GRCh38
rs766839217 Chr.8: 129748462-129748462 on Build GRCh38
rs909413827 Chr.8: 129748417-129748417 on Build GRCh38
rs779994855 Chr.8: 129748503-129748503 on Build GRCh38
rs750123614 Chr.8: 129748587-129748587 on Build GRCh38
rs777586767 Chr.8: 129748705-129748705 on Build GRCh38
rs926525412 Chr.8: 129748650-129748650 on Build GRCh38
rs777153525 Chr.8: 129748549-129748549 on Build GRCh38
rs554878884 Chr.8: 129748794-129748794 on Build GRCh38
rs774910688 Chr.8: 129748744-129748744 on Build GRCh38
rs979564302 Chr.8: 129748553-129748553 on Build GRCh38
rs747260455 Chr.8: 129748627-129748627 on Build GRCh38
rs1008178850 Chr.8: 129748749-129748749 on Build GRCh38
rs776622434 Chr.8: 129748631-129748631 on Build GRCh38
rs770270429 Chr.8: 129748634-129748634 on Build GRCh38
rs1032490508 Chr.8: 129748796-129748796 on Build GRCh38
rs919279559 Chr.8: 129748573-129748573 on Build GRCh38
rs374835768 Chr.8: 129748577-129748577 on Build GRCh38
rs778760906 Chr.8: 129748680-129748680 on Build GRCh38
rs376086270 Chr.8: 129748452-129748452 on Build GRCh38
rs767565999 Chr.8: 129748456-129748456 on Build GRCh38
rs1042471734 Chr.8: 129748479-129748479 on Build GRCh38
rs202184744 Chr.8: 129748641-129748641 on Build GRCh38
rs748360610 Chr.8: 129748532-129748532 on Build GRCh38
rs749217446 Chr.8: 129748508-129748508 on Build GRCh38
rs897671513 Chr.8: 129748703-129748703 on Build GRCh38
rs747867390 Chr.8: 129748605-129748605 on Build GRCh38
rs577181423 Chr.8: 129748779-129748779 on Build GRCh38
rs760686608 Chr.8: 129748547-129748547 on Build GRCh38
rs772199372 Chr.8: 129748529-129748530 on Build GRCh38
rs374274478 Chr.8: 129748509-129748509 on Build GRCh38
rs771864086 Chr.8: 129748606-129748606 on Build GRCh38
rs201817415 Chr.8: 129748557-129748557 on Build GRCh38
rs776098748 Chr.8: 129748728-129748728 on Build GRCh38
rs188881042 Chr.8: 129748781-129748781 on Build GRCh38
rs569568081 Chr.8: 129748711-129748711 on Build GRCh38
rs776155660 Chr.8: 129748637-129748637 on Build GRCh38
rs200964859 Chr.8: 129748700-129748700 on Build GRCh38
rs4733559 Chr.8: 129748528-129748528 on Build GRCh38
rs770014882 Chr.8: 129748717-129748717 on Build GRCh38
rs949080598 Chr.8: 129748639-129748639 on Build GRCh38
rs370955474 Chr.8: 129748552-129748552 on Build GRCh38
rs150468394 Chr.8: 129748615-129748615 on Build GRCh38
rs754932128 Chr.8: 129748603-129748603 on Build GRCh38
rs368419620 Chr.8: 129748524-129748524 on Build GRCh38
rs753795099 Chr.8: 129748593-129748593 on Build GRCh38
rs753169677 Chr.8: 129748575-129748575 on Build GRCh38
rs772342562 Chr.8: 129748543-129748543 on Build GRCh38
rs898565948 Chr.8: 129748453-129748453 on Build GRCh38
rs765903577 Chr.8: 129748662-129748662 on Build GRCh38
rs755594549 Chr.8: 129748487-129748487 on Build GRCh38
rs755612165 Chr.8: 129748590-129748590 on Build GRCh38
rs369201045 Chr.8: 129748632-129748632 on Build GRCh38
rs760367904 Chr.8: 129748751-129748751 on Build GRCh38
rs150668828 Chr.8: 129748583-129748583 on Build GRCh38
rs750072260 Chr.8: 129748644-129748644 on Build GRCh38
rs1001227177 Chr.8: 129748495-129748495 on Build GRCh38
rs987874846 Chr.8: 129748815-129748815 on Build GRCh38
rs200212993 Chr.8: 129748469-129748469 on Build GRCh38
rs778626448 Chr.8: 129748529-129748529 on Build GRCh38
rs1037572431 Chr.8: 129748416-129748416 on Build GRCh38
rs149748731 Chr.8: 129748706-129748706 on Build GRCh38
rs761162492 Chr.8: 129748460-129748460 on Build GRCh38

View Full Product Details