Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228097337-228097828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF1
Entrez Gene ID
375
Gene Name
ADP ribosylation factor 1
Gene Aliases
-
UniGene
Hs.286221
Gene Chromosome Location
Chr.1: 228082660-228099212 on Build GRCh38
Transcripts
NM_001024226.1 NM_001024227.1 NM_001024228.1 NM_001658.3 XM_017001233.1 NR_037415.1

Gene Symbol
MIR3620
Entrez Gene ID
100500810
Gene Name
microRNA 3620
Gene Aliases
mir-3620
UniGene
Gene Chromosome Location
Chr.1: 228097263-228097341 on Build GRCh38
Transcripts
NM_001024226.1 NM_001024227.1 NM_001024228.1 NM_001658.3 XM_017001233.1 NR_037415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5469978 c.166G>A p.V56M Chr.1: 228097359-228097359 on Build GRCh38
1317574 c.165C>T p.T55T Chr.1: 228097358-228097358 on Build GRCh38
5182008 c.377A>G p.N126S Chr.1: 228097708-228097708 on Build GRCh38
5949797 c.270C>T p.F90F Chr.1: 228097601-228097601 on Build GRCh38
1626862 c.182T>G p.I61S Chr.1: 228097375-228097375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117028801 Chr.1: 228097542-228097542 on Build GRCh38
rs754126054 Chr.1: 228097568-228097568 on Build GRCh38
rs1000671194 Chr.1: 228097498-228097498 on Build GRCh38
rs142418933 Chr.1: 228097673-228097673 on Build GRCh38
rs536352994 Chr.1: 228097552-228097552 on Build GRCh38
rs147354087 Chr.1: 228097631-228097631 on Build GRCh38
rs748857647 Chr.1: 228097717-228097718 on Build GRCh38
rs764690724 Chr.1: 228097424-228097424 on Build GRCh38
rs535462991 Chr.1: 228097535-228097535 on Build GRCh38
rs777847166 Chr.1: 228097751-228097751 on Build GRCh38
rs766906376 Chr.1: 228097723-228097723 on Build GRCh38
rs11541558 Chr.1: 228097385-228097385 on Build GRCh38
rs754506796 Chr.1: 228097397-228097397 on Build GRCh38
rs748152912 Chr.1: 228097486-228097486 on Build GRCh38
rs752069371 Chr.1: 228097428-228097428 on Build GRCh38
rs781034886 Chr.1: 228097765-228097765 on Build GRCh38
rs755237018 Chr.1: 228097574-228097574 on Build GRCh38
rs752583036 Chr.1: 228097681-228097681 on Build GRCh38
rs762769241 Chr.1: 228097454-228097455 on Build GRCh38
rs765917983 Chr.1: 228097376-228097376 on Build GRCh38
rs780442475 Chr.1: 228097472-228097472 on Build GRCh38
rs11541556 Chr.1: 228097661-228097661 on Build GRCh38
rs373715936 Chr.1: 228097703-228097703 on Build GRCh38
rs201067408 Chr.1: 228097546-228097546 on Build GRCh38
rs756404286 Chr.1: 228097469-228097469 on Build GRCh38
rs768860382 Chr.1: 228097481-228097481 on Build GRCh38
rs975692572 Chr.1: 228097725-228097725 on Build GRCh38
rs143545613 Chr.1: 228097667-228097667 on Build GRCh38
rs772867236 Chr.1: 228097370-228097370 on Build GRCh38
rs779199839 Chr.1: 228097576-228097576 on Build GRCh38
rs1032194642 Chr.1: 228097499-228097499 on Build GRCh38
rs202220531 Chr.1: 228097522-228097522 on Build GRCh38
rs200897192 Chr.1: 228097722-228097722 on Build GRCh38
rs372015059 Chr.1: 228097388-228097388 on Build GRCh38
rs552986915 Chr.1: 228097358-228097358 on Build GRCh38
rs760445481 Chr.1: 228097373-228097373 on Build GRCh38
rs147721417 Chr.1: 228097791-228097792 on Build GRCh38
rs1023852317 Chr.1: 228097534-228097534 on Build GRCh38
rs752138967 Chr.1: 228097657-228097681 on Build GRCh38
rs767885108 Chr.1: 228097550-228097550 on Build GRCh38
rs754248760 Chr.1: 228097724-228097724 on Build GRCh38
rs11541561 Chr.1: 228097694-228097694 on Build GRCh38
rs751420953 Chr.1: 228097498-228097499 on Build GRCh38
rs754859607 Chr.1: 228097545-228097545 on Build GRCh38
rs1010202646 Chr.1: 228097656-228097656 on Build GRCh38
rs752915360 Chr.1: 228097742-228097742 on Build GRCh38
rs750719848 Chr.1: 228097466-228097466 on Build GRCh38
rs201797905 Chr.1: 228097585-228097585 on Build GRCh38
rs990598292 Chr.1: 228097519-228097519 on Build GRCh38
rs370867199 Chr.1: 228097548-228097548 on Build GRCh38
rs373902191 Chr.1: 228097541-228097541 on Build GRCh38
rs200443433 Chr.1: 228097794-228097794 on Build GRCh38
rs760365227 Chr.1: 228097500-228097500 on Build GRCh38
rs375875865 Chr.1: 228097764-228097764 on Build GRCh38
rs766057583 Chr.1: 228097498-228097498 on Build GRCh38
rs765536091 Chr.1: 228097732-228097732 on Build GRCh38
rs762388073 Chr.1: 228097720-228097720 on Build GRCh38
rs763751551 Chr.1: 228097718-228097718 on Build GRCh38
rs901254200 Chr.1: 228097654-228097654 on Build GRCh38
rs367780730 Chr.1: 228097726-228097726 on Build GRCh38
rs750890801 Chr.1: 228097551-228097551 on Build GRCh38
rs934172552 Chr.1: 228097729-228097729 on Build GRCh38
rs781667395 Chr.1: 228097460-228097460 on Build GRCh38
rs780499813 Chr.1: 228097562-228097562 on Build GRCh38
rs755638275 Chr.1: 228097716-228097717 on Build GRCh38
rs753396348 Chr.1: 228097394-228097394 on Build GRCh38
rs374548874 Chr.1: 228097485-228097485 on Build GRCh38
rs771901253 Chr.1: 228097499-228097499 on Build GRCh38
rs775027460 Chr.1: 228097719-228097719 on Build GRCh38
rs112615324 Chr.1: 228097552-228097552 on Build GRCh38
rs369646198 Chr.1: 228097476-228097476 on Build GRCh38
rs560609119 Chr.1: 228097771-228097771 on Build GRCh38
rs758439811 Chr.1: 228097745-228097745 on Build GRCh38
rs369031105 Chr.1: 228097546-228097546 on Build GRCh38
rs1020829141 Chr.1: 228097495-228097495 on Build GRCh38
rs941366287 Chr.1: 228097790-228097790 on Build GRCh38
rs745638410 Chr.1: 228097806-228097806 on Build GRCh38
rs754134486 Chr.1: 228097453-228097454 on Build GRCh38
rs762419898 Chr.1: 228097549-228097549 on Build GRCh38
rs746936604 Chr.1: 228097754-228097754 on Build GRCh38
rs1047120728 Chr.1: 228097747-228097747 on Build GRCh38
rs375556877 Chr.1: 228097448-228097448 on Build GRCh38
rs763615109 Chr.1: 228097721-228097721 on Build GRCh38
rs371617800 Chr.1: 228097601-228097601 on Build GRCh38

View Full Product Details