Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50799153-50799712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf48
Entrez Gene ID
84798
Gene Name
chromosome 19 open reading frame 48
Gene Aliases
-
UniGene
Hs.256301
Gene Chromosome Location
Chr.19: 50797693-50804853 on Build GRCh38
Transcripts
NM_001290149.1 NM_001290150.1 NM_001290151.1 NM_001290152.1 NM_001290153.1 NM_001290154.1 NM_001290155.1 NM_032712.2 NM_199249.2 NM_199250.2 NR_003067.1

Gene Symbol
SNORD88A
Entrez Gene ID
692202
Gene Name
small nucleolar RNA, C/D box 88A
Gene Aliases
HBII-180A
UniGene
Gene Chromosome Location
Chr.19: 50799439-50799535 on Build GRCh38
Transcripts
NM_001290149.1 NM_001290150.1 NM_001290151.1 NM_001290152.1 NM_001290153.1 NM_001290154.1 NM_001290155.1 NM_032712.2 NM_199249.2 NM_199250.2 NR_003067.1

SNP Detail

SNP ID Chromosome Location
rs1031423214 Chr.19: 50799382-50799382 on Build GRCh38
rs886516143 Chr.19: 50799313-50799313 on Build GRCh38
rs143867891 Chr.19: 50799198-50799199 on Build GRCh38
rs759717139 Chr.19: 50799450-50799450 on Build GRCh38
rs570929046 Chr.19: 50799534-50799534 on Build GRCh38
rs770007971 Chr.19: 50799408-50799408 on Build GRCh38
rs761877676 Chr.19: 50799524-50799524 on Build GRCh38
rs552574811 Chr.19: 50799515-50799515 on Build GRCh38
rs58572527 Chr.19: 50799376-50799377 on Build GRCh38
rs369927981 Chr.19: 50799217-50799218 on Build GRCh38
rs567289034 Chr.19: 50799460-50799460 on Build GRCh38
rs771648149 Chr.19: 50799376-50799382 on Build GRCh38
rs1022538186 Chr.19: 50799678-50799678 on Build GRCh38
rs970085585 Chr.19: 50799281-50799281 on Build GRCh38
rs760547912 Chr.19: 50799457-50799457 on Build GRCh38
rs554929640 Chr.19: 50799533-50799533 on Build GRCh38
rs774400873 Chr.19: 50799444-50799444 on Build GRCh38
rs1039066493 Chr.19: 50799273-50799273 on Build GRCh38
rs985002412 Chr.19: 50799300-50799300 on Build GRCh38
rs181246022 Chr.19: 50799204-50799204 on Build GRCh38
rs781453979 Chr.19: 50799399-50799399 on Build GRCh38
rs1004156871 Chr.19: 50799342-50799343 on Build GRCh38
rs933848726 Chr.19: 50799333-50799333 on Build GRCh38
rs941013621 Chr.19: 50799303-50799303 on Build GRCh38
rs992705112 Chr.19: 50799241-50799241 on Build GRCh38
rs771013559 Chr.19: 50799432-50799432 on Build GRCh38
rs778055647 Chr.19: 50799485-50799485 on Build GRCh38
rs778722947 Chr.19: 50799474-50799475 on Build GRCh38
rs72528782 Chr.19: 50799218-50799219 on Build GRCh38
rs911055247 Chr.19: 50799346-50799346 on Build GRCh38
rs1043681572 Chr.19: 50799650-50799650 on Build GRCh38
rs775783043 Chr.19: 50799503-50799503 on Build GRCh38
rs773741172 Chr.19: 50799419-50799419 on Build GRCh38
rs918302497 Chr.19: 50799324-50799324 on Build GRCh38
rs934524789 Chr.19: 50799211-50799211 on Build GRCh38
rs866024501 Chr.19: 50799302-50799302 on Build GRCh38
rs972475394 Chr.19: 50799317-50799317 on Build GRCh38
rs942491019 Chr.19: 50799649-50799649 on Build GRCh38
rs760748247 Chr.19: 50799506-50799506 on Build GRCh38
rs3810285 Chr.19: 50799502-50799502 on Build GRCh38
rs994820087 Chr.19: 50799298-50799299 on Build GRCh38
rs763912779 Chr.19: 50799458-50799458 on Build GRCh38
rs999638804 Chr.19: 50799197-50799197 on Build GRCh38
rs895343008 Chr.19: 50799588-50799588 on Build GRCh38
rs745982194 Chr.19: 50799495-50799495 on Build GRCh38
rs955338727 Chr.19: 50799539-50799539 on Build GRCh38
rs986558062 Chr.19: 50799592-50799592 on Build GRCh38
rs542894082 Chr.19: 50799687-50799687 on Build GRCh38
rs774463302 Chr.19: 50799493-50799493 on Build GRCh38
rs763385519 Chr.19: 50799422-50799422 on Build GRCh38
rs1012478869 Chr.19: 50799365-50799365 on Build GRCh38
rs751239177 Chr.19: 50799579-50799579 on Build GRCh38
rs77396687 Chr.19: 50799648-50799648 on Build GRCh38
rs749927051 Chr.19: 50799466-50799466 on Build GRCh38
rs756508225 Chr.19: 50799484-50799484 on Build GRCh38
rs911801608 Chr.19: 50799237-50799237 on Build GRCh38
rs942626465 Chr.19: 50799363-50799363 on Build GRCh38
rs3810284 Chr.19: 50799459-50799459 on Build GRCh38
rs758252125 Chr.19: 50799454-50799454 on Build GRCh38
rs559664767 Chr.19: 50799203-50799203 on Build GRCh38
rs3833237 Chr.19: 50799216-50799217 on Build GRCh38
rs778629467 Chr.19: 50799461-50799461 on Build GRCh38
rs999501368 Chr.19: 50799388-50799388 on Build GRCh38
rs1023893660 Chr.19: 50799242-50799242 on Build GRCh38
rs749852420 Chr.19: 50799489-50799489 on Build GRCh38
rs189191071 Chr.19: 50799194-50799194 on Build GRCh38
rs764035508 Chr.19: 50799512-50799512 on Build GRCh38
rs915551133 Chr.19: 50799198-50799198 on Build GRCh38
rs925160343 Chr.19: 50799662-50799662 on Build GRCh38
rs771424345 Chr.19: 50799491-50799491 on Build GRCh38
rs1051332679 Chr.19: 50799343-50799343 on Build GRCh38
rs749148065 Chr.19: 50799401-50799402 on Build GRCh38
rs551714627 Chr.19: 50799246-50799246 on Build GRCh38
rs779782302 Chr.19: 50799475-50799475 on Build GRCh38
rs745610219 Chr.19: 50799337-50799337 on Build GRCh38
rs960921636 Chr.19: 50799220-50799220 on Build GRCh38
rs1006894604 Chr.19: 50799180-50799180 on Build GRCh38
rs758038966 Chr.19: 50799474-50799474 on Build GRCh38
rs1022343485 Chr.19: 50799193-50799193 on Build GRCh38
rs113992561 Chr.19: 50799293-50799293 on Build GRCh38
rs186507896 Chr.19: 50799294-50799294 on Build GRCh38
rs1052532227 Chr.19: 50799394-50799394 on Build GRCh38
rs903876739 Chr.19: 50799379-50799379 on Build GRCh38
rs978506754 Chr.19: 50799199-50799201 on Build GRCh38
rs143534987 Chr.19: 50799368-50799368 on Build GRCh38
rs557236479 Chr.19: 50799547-50799547 on Build GRCh38
rs397819575 Chr.19: 50799219-50799220 on Build GRCh38
rs538908593 Chr.19: 50799535-50799535 on Build GRCh38
rs754708361 Chr.19: 50799419-50799419 on Build GRCh38
rs767756978 Chr.19: 50799452-50799452 on Build GRCh38
rs748527471 Chr.19: 50799405-50799405 on Build GRCh38

View Full Product Details