Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 80496700-80497214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP128
Entrez Gene ID
145508
Gene Name
centrosomal protein 128
Gene Aliases
C14orf145, C14orf61, LEDP/132
UniGene
Hs.162889
Gene Chromosome Location
Chr.14: 80478321-80959501 on Build GRCh38
Transcripts
NM_152446.3 XM_006720056.3 XM_011536490.2 XM_011536491.2 XM_011536492.2 XM_011536493.2 XM_017021042.1 XM_017021043.1

SNP Detail

SNP ID Chromosome Location
rs1052698656 Chr.14: 80497027-80497027 on Build GRCh38
rs563448162 Chr.14: 80496876-80496876 on Build GRCh38
rs1017932424 Chr.14: 80497066-80497066 on Build GRCh38
rs796142383 Chr.14: 80497041-80497042 on Build GRCh38
rs765982721 Chr.14: 80497174-80497174 on Build GRCh38
rs542373262 Chr.14: 80497058-80497058 on Build GRCh38
rs749598601 Chr.14: 80496947-80496947 on Build GRCh38
rs964986611 Chr.14: 80497188-80497191 on Build GRCh38
rs72522774 Chr.14: 80497039-80497040 on Build GRCh38
rs926542796 Chr.14: 80497002-80497002 on Build GRCh38
rs576764338 Chr.14: 80496810-80496810 on Build GRCh38
rs374772583 Chr.14: 80497004-80497004 on Build GRCh38
rs56003521 Chr.14: 80497040-80497041 on Build GRCh38
rs937888140 Chr.14: 80497012-80497012 on Build GRCh38
rs913535615 Chr.14: 80496921-80496921 on Build GRCh38
rs907963011 Chr.14: 80496848-80496848 on Build GRCh38
rs558117861 Chr.14: 80497074-80497074 on Build GRCh38
rs771386932 Chr.14: 80496998-80496998 on Build GRCh38
rs993501731 Chr.14: 80497103-80497103 on Build GRCh38
rs577602712 Chr.14: 80496992-80496992 on Build GRCh38
rs998290084 Chr.14: 80497157-80497157 on Build GRCh38
rs182599464 Chr.14: 80497029-80497029 on Build GRCh38
rs544176289 Chr.14: 80496845-80496845 on Build GRCh38
rs200565867 Chr.14: 80497037-80497037 on Build GRCh38
rs1030575774 Chr.14: 80497176-80497176 on Build GRCh38
rs887744989 Chr.14: 80497154-80497154 on Build GRCh38
rs772973972 Chr.14: 80496933-80496933 on Build GRCh38
rs1017958261 Chr.14: 80496762-80496762 on Build GRCh38
rs964927333 Chr.14: 80496780-80496780 on Build GRCh38
rs115970124 Chr.14: 80496796-80496796 on Build GRCh38
rs1037636930 Chr.14: 80496914-80496914 on Build GRCh38
rs145531335 Chr.14: 80497181-80497181 on Build GRCh38
rs954888414 Chr.14: 80496831-80496831 on Build GRCh38
rs989109212 Chr.14: 80496751-80496751 on Build GRCh38
rs948078931 Chr.14: 80496979-80496979 on Build GRCh38
rs188066035 Chr.14: 80497101-80497101 on Build GRCh38
rs952331892 Chr.14: 80496752-80496752 on Build GRCh38
rs531497840 Chr.14: 80497159-80497159 on Build GRCh38
rs896584198 Chr.14: 80497049-80497049 on Build GRCh38
rs942122559 Chr.14: 80496869-80496869 on Build GRCh38
rs1006206747 Chr.14: 80497158-80497158 on Build GRCh38
rs999362115 Chr.14: 80497023-80497023 on Build GRCh38
rs916599143 Chr.14: 80496949-80496949 on Build GRCh38
rs527410196 Chr.14: 80497042-80497042 on Build GRCh38
rs965273509 Chr.14: 80496789-80496789 on Build GRCh38
rs33941291 Chr.14: 80497041-80497042 on Build GRCh38
rs189572040 Chr.14: 80496915-80496915 on Build GRCh38
rs560750419 Chr.14: 80497105-80497105 on Build GRCh38

View Full Product Details