Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4952758-4953255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO3
Entrez Gene ID
2027
Gene Name
enolase 3
Gene Aliases
GSD13, MSE
UniGene
Hs.224171
Gene Chromosome Location
Chr.17: 4949182-4957131 on Build GRCh38
Transcripts
NM_001193503.1 NM_001976.4 NM_053013.3 XM_005256521.2 XM_011523729.1 XM_017024346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5946842 c.89G>A p.R30Q Chr.17: 4952798-4952798 on Build GRCh38
7278965 c.100G>A p.A34T Chr.17: 4952809-4952809 on Build GRCh38
1684474 c.199delG p.E67fs*16 Chr.17: 4953068-4953068 on Build GRCh38
4829047 c.155G>A p.G52E Chr.17: 4952864-4952864 on Build GRCh38
436845 c.106C>G p.P36A Chr.17: 4952815-4952815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781666028 Chr.17: 4953152-4953152 on Build GRCh38
rs753323275 Chr.17: 4952928-4952928 on Build GRCh38
rs961179627 Chr.17: 4953079-4953079 on Build GRCh38
rs965347169 Chr.17: 4952834-4952834 on Build GRCh38
rs557068790 Chr.17: 4953035-4953035 on Build GRCh38
rs757769893 Chr.17: 4952863-4952863 on Build GRCh38
rs77592115 Chr.17: 4953171-4953171 on Build GRCh38
rs543203167 Chr.17: 4953074-4953074 on Build GRCh38
rs574181335 Chr.17: 4953055-4953055 on Build GRCh38
rs936041755 Chr.17: 4952950-4952950 on Build GRCh38
rs754637196 Chr.17: 4952825-4952825 on Build GRCh38
rs757716763 Chr.17: 4952930-4952930 on Build GRCh38
rs114341110 Chr.17: 4953017-4953017 on Build GRCh38
rs374519215 Chr.17: 4953027-4953027 on Build GRCh38
rs967756914 Chr.17: 4953137-4953137 on Build GRCh38
rs374581656 Chr.17: 4953159-4953159 on Build GRCh38
rs761512276 Chr.17: 4953129-4953129 on Build GRCh38
rs538519673 Chr.17: 4952924-4952924 on Build GRCh38
rs1049665944 Chr.17: 4953175-4953175 on Build GRCh38
rs367565420 Chr.17: 4952801-4952801 on Build GRCh38
rs775155235 Chr.17: 4953044-4953044 on Build GRCh38
rs749748461 Chr.17: 4952862-4952862 on Build GRCh38
rs886053165 Chr.17: 4952792-4952792 on Build GRCh38
rs763535870 Chr.17: 4952893-4952895 on Build GRCh38
rs201571802 Chr.17: 4952785-4952785 on Build GRCh38
rs963982168 Chr.17: 4952840-4952840 on Build GRCh38
rs927577100 Chr.17: 4952945-4952945 on Build GRCh38
rs201859879 Chr.17: 4952797-4952797 on Build GRCh38
rs779213108 Chr.17: 4952865-4952865 on Build GRCh38
rs935129522 Chr.17: 4952885-4952885 on Build GRCh38
rs200793919 Chr.17: 4952895-4952895 on Build GRCh38
rs868413211 Chr.17: 4953182-4953182 on Build GRCh38
rs778031259 Chr.17: 4952916-4952916 on Build GRCh38
rs373537923 Chr.17: 4953043-4953043 on Build GRCh38
rs771319383 Chr.17: 4952796-4952796 on Build GRCh38
rs111630522 Chr.17: 4953184-4953184 on Build GRCh38
rs553035991 Chr.17: 4952908-4952908 on Build GRCh38
rs865889960 Chr.17: 4953183-4953183 on Build GRCh38
rs774395644 Chr.17: 4953036-4953036 on Build GRCh38
rs746259097 Chr.17: 4952868-4952868 on Build GRCh38
rs371406227 Chr.17: 4953153-4953153 on Build GRCh38
rs778318241 Chr.17: 4952782-4952782 on Build GRCh38
rs777213473 Chr.17: 4952851-4952851 on Build GRCh38
rs930161930 Chr.17: 4953211-4953211 on Build GRCh38
rs755263055 Chr.17: 4953143-4953143 on Build GRCh38
rs566502858 Chr.17: 4953218-4953218 on Build GRCh38
rs775659728 Chr.17: 4952876-4952876 on Build GRCh38
rs768769464 Chr.17: 4952886-4952886 on Build GRCh38
rs917488664 Chr.17: 4953195-4953195 on Build GRCh38
rs370439421 Chr.17: 4952848-4952848 on Build GRCh38
rs189305036 Chr.17: 4953172-4953172 on Build GRCh38
rs753486120 Chr.17: 4953104-4953105 on Build GRCh38
rs766217572 Chr.17: 4952817-4952817 on Build GRCh38
rs143992803 Chr.17: 4952881-4952881 on Build GRCh38
rs543950776 Chr.17: 4952978-4952978 on Build GRCh38
rs760194274 Chr.17: 4952866-4952866 on Build GRCh38
rs374246241 Chr.17: 4953104-4953104 on Build GRCh38
rs774951960 Chr.17: 4952856-4952857 on Build GRCh38
rs141245336 Chr.17: 4952815-4952815 on Build GRCh38
rs376821730 Chr.17: 4952804-4952804 on Build GRCh38
rs546785438 Chr.17: 4953194-4953194 on Build GRCh38
rs748416893 Chr.17: 4953080-4953080 on Build GRCh38
rs759611531 Chr.17: 4953042-4953042 on Build GRCh38
rs375137643 Chr.17: 4952906-4952906 on Build GRCh38
rs560611328 Chr.17: 4952879-4952879 on Build GRCh38
rs372561412 Chr.17: 4952927-4952927 on Build GRCh38
rs776731834 Chr.17: 4953023-4953023 on Build GRCh38
rs752156818 Chr.17: 4953138-4953138 on Build GRCh38
rs578052442 Chr.17: 4952957-4952957 on Build GRCh38
rs543435370 Chr.17: 4953058-4953058 on Build GRCh38
rs141103742 Chr.17: 4952824-4952824 on Build GRCh38
rs760369089 Chr.17: 4953107-4953107 on Build GRCh38
rs202207445 Chr.17: 4953086-4953086 on Build GRCh38
rs535451602 Chr.17: 4952792-4952792 on Build GRCh38
rs776292024 Chr.17: 4953134-4953135 on Build GRCh38
rs970683373 Chr.17: 4953106-4953106 on Build GRCh38
rs143945974 Chr.17: 4953095-4953095 on Build GRCh38
rs980318534 Chr.17: 4953109-4953109 on Build GRCh38
rs111316621 Chr.17: 4953110-4953110 on Build GRCh38
rs749942573 Chr.17: 4953130-4953130 on Build GRCh38
rs1040901145 Chr.17: 4953011-4953011 on Build GRCh38
rs75279666 Chr.17: 4953126-4953126 on Build GRCh38
rs747724047 Chr.17: 4952779-4952779 on Build GRCh38
rs763679836 Chr.17: 4953052-4953052 on Build GRCh38
rs780396901 Chr.17: 4953062-4953062 on Build GRCh38
rs200352599 Chr.17: 4952917-4952917 on Build GRCh38
rs926240215 Chr.17: 4953125-4953125 on Build GRCh38
rs752338197 Chr.17: 4952832-4952832 on Build GRCh38
rs201048680 Chr.17: 4953135-4953135 on Build GRCh38
rs369407375 Chr.17: 4953015-4953015 on Build GRCh38
rs767296674 Chr.17: 4952831-4952831 on Build GRCh38
rs768540512 Chr.17: 4953117-4953117 on Build GRCh38
rs914950773 Chr.17: 4952905-4952905 on Build GRCh38
rs776135288 Chr.17: 4953227-4953227 on Build GRCh38
rs766817185 Chr.17: 4953136-4953136 on Build GRCh38
rs370041604 Chr.17: 4953019-4953019 on Build GRCh38
rs977014646 Chr.17: 4952844-4952844 on Build GRCh38
rs368381926 Chr.17: 4952907-4952907 on Build GRCh38
rs151153333 Chr.17: 4952790-4952790 on Build GRCh38
rs761281354 Chr.17: 4953056-4953056 on Build GRCh38
rs771937916 Chr.17: 4953097-4953097 on Build GRCh38
rs746414652 Chr.17: 4953012-4953012 on Build GRCh38
rs1041242459 Chr.17: 4953169-4953169 on Build GRCh38
rs945185772 Chr.17: 4953007-4953007 on Build GRCh38
rs201114068 Chr.17: 4952837-4952837 on Build GRCh38
rs764120380 Chr.17: 4952803-4952803 on Build GRCh38
rs752387577 Chr.17: 4952923-4952923 on Build GRCh38
rs770147328 Chr.17: 4952780-4952780 on Build GRCh38
rs777962361 Chr.17: 4953156-4953156 on Build GRCh38
rs238238 Chr.17: 4953081-4953081 on Build GRCh38
rs267604959 Chr.17: 4952798-4952798 on Build GRCh38
rs1053122313 Chr.17: 4952955-4952955 on Build GRCh38
rs139073445 Chr.17: 4952875-4952875 on Build GRCh38
rs762880151 Chr.17: 4952901-4952901 on Build GRCh38
rs146408284 Chr.17: 4953078-4953078 on Build GRCh38
rs984120559 Chr.17: 4952820-4952820 on Build GRCh38
rs771094653 Chr.17: 4953225-4953225 on Build GRCh38
rs377545954 Chr.17: 4953070-4953070 on Build GRCh38
rs112363205 Chr.17: 4953048-4953048 on Build GRCh38
rs779374700 Chr.17: 4952933-4952933 on Build GRCh38

View Full Product Details