Product Details
- Primer Sequences
- Log in to view
- Primer Chromosome Location
-
Chr.1:
145921951-145921392
on Build GRCh38
View in UCSC browser
- Format
- Each primer will be delivered in a single tube, dried-down
- Quick Reference Guide
- Primer Designer™ Tool Quick Reference
- Ampliseq ID's
Gene Details
- Gene Symbol
- GNRHR2
- Entrez Gene ID
-
114814
- Gene Name
- gonadotropin releasing hormone receptor 2 (pseudogene)
- Gene Aliases
-
GnRH-II-R
- UniGene
- Hs.356873
- Gene Chromosome Location
-
Chr.1:
145919013-145925341
on Build GRCh38
- Transcripts
-
NR_002328.4
NR_104033.1
NM_005105.4
- Gene Symbol
- RBM8A
- Entrez Gene ID
-
9939
- Gene Name
- RNA binding motif protein 8A
- Gene Aliases
-
BOV-1A, BOV-1B, BOV-1C, C1DELq21.1, DEL1q21.1, MDS014, RBM8, RBM8B, TAR, Y14, ZNRP, ZRNP1
- UniGene
- Hs.591455
- Gene Chromosome Location
-
Chr.1:
145921556-145927536
on Build GRCh38
- Transcripts
-
NR_002328.4
NR_104033.1
NM_005105.4
View Full Product Details