Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140802785-140803314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_031496.1 NM_018906.2 NM_031497.1

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_031496.1 NM_018906.2 NM_031497.1

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018905.2 NM_031496.1 NM_018906.2 NM_031497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3609593 c.1885G>A p.G629R Chr.5: 140803082-140803082 on Build GRCh38
6260136 c.1993G>T p.V665L Chr.5: 140803190-140803190 on Build GRCh38
5744259 c.2117C>T p.A706V Chr.5: 140803314-140803314 on Build GRCh38
382384 c.1942C>A p.L648M Chr.5: 140803139-140803139 on Build GRCh38
6842743 c.1719C>G p.I573M Chr.5: 140802916-140802916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782172629 Chr.5: 140803281-140803281 on Build GRCh38
rs782799537 Chr.5: 140802949-140802949 on Build GRCh38
rs782694897 Chr.5: 140802841-140802841 on Build GRCh38
rs782401655 Chr.5: 140802846-140802846 on Build GRCh38
rs782438161 Chr.5: 140803095-140803095 on Build GRCh38
rs17844262 Chr.5: 140803192-140803192 on Build GRCh38
rs781793277 Chr.5: 140803035-140803035 on Build GRCh38
rs782799500 Chr.5: 140803283-140803283 on Build GRCh38
rs375111738 Chr.5: 140802944-140802944 on Build GRCh38
rs372042203 Chr.5: 140803008-140803008 on Build GRCh38
rs781941927 Chr.5: 140803078-140803078 on Build GRCh38
rs200608373 Chr.5: 140802882-140802882 on Build GRCh38
rs782532560 Chr.5: 140803037-140803037 on Build GRCh38
rs782345607 Chr.5: 140803077-140803077 on Build GRCh38
rs782553261 Chr.5: 140802902-140802902 on Build GRCh38
rs780506250 Chr.5: 140802941-140802941 on Build GRCh38
rs758831845 Chr.5: 140802900-140802900 on Build GRCh38
rs138097279 Chr.5: 140803012-140803012 on Build GRCh38
rs782736507 Chr.5: 140803274-140803274 on Build GRCh38
rs782219037 Chr.5: 140802992-140802992 on Build GRCh38
rs782140638 Chr.5: 140803010-140803010 on Build GRCh38
rs782356178 Chr.5: 140802913-140802913 on Build GRCh38
rs781909871 Chr.5: 140803228-140803228 on Build GRCh38
rs781825644 Chr.5: 140803106-140803106 on Build GRCh38
rs368819593 Chr.5: 140803081-140803081 on Build GRCh38
rs781934902 Chr.5: 140802997-140802997 on Build GRCh38
rs782366845 Chr.5: 140803155-140803155 on Build GRCh38
rs782531842 Chr.5: 140803229-140803229 on Build GRCh38
rs782240036 Chr.5: 140803075-140803075 on Build GRCh38
rs782024220 Chr.5: 140802805-140802805 on Build GRCh38
rs982936754 Chr.5: 140803011-140803011 on Build GRCh38
rs148554584 Chr.5: 140803223-140803223 on Build GRCh38
rs781956675 Chr.5: 140802817-140802817 on Build GRCh38
rs564787047 Chr.5: 140803263-140803263 on Build GRCh38
rs782605739 Chr.5: 140802914-140802914 on Build GRCh38
rs782640007 Chr.5: 140802819-140802819 on Build GRCh38
rs782015955 Chr.5: 140802936-140802936 on Build GRCh38
rs139788338 Chr.5: 140803100-140803100 on Build GRCh38
rs781937723 Chr.5: 140802859-140802859 on Build GRCh38
rs34395025 Chr.5: 140803234-140803234 on Build GRCh38
rs782103564 Chr.5: 140802875-140802875 on Build GRCh38
rs782276594 Chr.5: 140802975-140802975 on Build GRCh38
rs782496418 Chr.5: 140803134-140803134 on Build GRCh38
rs142727818 Chr.5: 140803124-140803124 on Build GRCh38
rs934549073 Chr.5: 140802890-140802890 on Build GRCh38
rs149374718 Chr.5: 140803137-140803137 on Build GRCh38
rs782603015 Chr.5: 140803057-140803057 on Build GRCh38
rs781822960 Chr.5: 140803219-140803219 on Build GRCh38
rs782254841 Chr.5: 140803144-140803144 on Build GRCh38
rs782378272 Chr.5: 140802940-140802940 on Build GRCh38
rs544698058 Chr.5: 140802912-140802912 on Build GRCh38
rs782428262 Chr.5: 140803000-140803000 on Build GRCh38
rs782172179 Chr.5: 140802945-140802945 on Build GRCh38
rs190722768 Chr.5: 140802881-140802881 on Build GRCh38
rs368124157 Chr.5: 140802957-140802957 on Build GRCh38
rs782677776 Chr.5: 140803142-140803142 on Build GRCh38
rs782418479 Chr.5: 140803172-140803172 on Build GRCh38
rs782454635 Chr.5: 140802971-140802971 on Build GRCh38
rs781920190 Chr.5: 140803178-140803178 on Build GRCh38
rs782115337 Chr.5: 140803196-140803196 on Build GRCh38
rs782451626 Chr.5: 140802899-140802899 on Build GRCh38
rs781861660 Chr.5: 140803131-140803131 on Build GRCh38
rs782637283 Chr.5: 140803231-140803231 on Build GRCh38
rs782081759 Chr.5: 140803272-140803272 on Build GRCh38
rs782195436 Chr.5: 140802833-140802833 on Build GRCh38
rs782642335 Chr.5: 140803073-140803073 on Build GRCh38
rs781996568 Chr.5: 140803096-140803096 on Build GRCh38
rs782201569 Chr.5: 140803138-140803138 on Build GRCh38
rs376717092 Chr.5: 140803050-140803050 on Build GRCh38
rs782310533 Chr.5: 140802838-140802838 on Build GRCh38
rs587776956 Chr.5: 140802947-140802947 on Build GRCh38
rs200661884 Chr.5: 140803159-140803159 on Build GRCh38
rs781899440 Chr.5: 140803125-140803125 on Build GRCh38
rs577958641 Chr.5: 140802965-140802965 on Build GRCh38
rs782463768 Chr.5: 140803220-140803220 on Build GRCh38
rs782285589 Chr.5: 140803065-140803065 on Build GRCh38
rs909930689 Chr.5: 140802812-140802812 on Build GRCh38
rs782171071 Chr.5: 140802999-140802999 on Build GRCh38
rs782445556 Chr.5: 140803060-140803060 on Build GRCh38
rs782648396 Chr.5: 140802905-140802905 on Build GRCh38
rs782279925 Chr.5: 140802842-140802842 on Build GRCh38
rs782739618 Chr.5: 140803206-140803206 on Build GRCh38
rs782398516 Chr.5: 140803072-140803072 on Build GRCh38
rs149770195 Chr.5: 140803255-140803255 on Build GRCh38
rs950112713 Chr.5: 140803107-140803107 on Build GRCh38
rs781966718 Chr.5: 140803021-140803021 on Build GRCh38
rs781845951 Chr.5: 140802809-140802809 on Build GRCh38
rs140993239 Chr.5: 140803291-140803291 on Build GRCh38
rs368728288 Chr.5: 140802874-140802874 on Build GRCh38
rs782763796 Chr.5: 140802808-140802808 on Build GRCh38
rs782518156 Chr.5: 140802982-140802982 on Build GRCh38
rs781928066 Chr.5: 140803277-140803277 on Build GRCh38
rs781822296 Chr.5: 140802970-140802970 on Build GRCh38
rs782417260 Chr.5: 140803257-140803257 on Build GRCh38
rs782014177 Chr.5: 140803258-140803258 on Build GRCh38
rs781849256 Chr.5: 140802821-140802821 on Build GRCh38
rs892863974 Chr.5: 140802917-140802917 on Build GRCh38
rs782102274 Chr.5: 140803098-140803098 on Build GRCh38
rs782417048 Chr.5: 140802923-140802923 on Build GRCh38
rs373825257 Chr.5: 140803042-140803042 on Build GRCh38
rs782135685 Chr.5: 140802807-140802807 on Build GRCh38
rs901539492 Chr.5: 140802964-140802964 on Build GRCh38
rs924510716 Chr.5: 140803126-140803126 on Build GRCh38
rs782634798 Chr.5: 140802989-140802989 on Build GRCh38
rs782371333 Chr.5: 140803264-140803264 on Build GRCh38
rs782125982 Chr.5: 140802937-140802937 on Build GRCh38
rs782332200 Chr.5: 140802994-140802994 on Build GRCh38
rs141385563 Chr.5: 140802886-140802886 on Build GRCh38
rs782186555 Chr.5: 140803059-140803059 on Build GRCh38
rs375912217 Chr.5: 140803129-140803129 on Build GRCh38
rs782357905 Chr.5: 140803245-140803245 on Build GRCh38
rs148243447 Chr.5: 140802851-140802851 on Build GRCh38
rs386352345 Chr.5: 140803127-140803127 on Build GRCh38
rs782074820 Chr.5: 140802943-140802943 on Build GRCh38
rs782082974 Chr.5: 140803024-140803024 on Build GRCh38
rs981760111 Chr.5: 140803111-140803111 on Build GRCh38
rs567006708 Chr.5: 140802857-140802857 on Build GRCh38
rs199682432 Chr.5: 140802868-140802868 on Build GRCh38
rs782577100 Chr.5: 140803247-140803247 on Build GRCh38
rs17844261 Chr.5: 140803182-140803182 on Build GRCh38
rs782318997 Chr.5: 140803162-140803162 on Build GRCh38
rs144811750 Chr.5: 140803177-140803177 on Build GRCh38
rs781991461 Chr.5: 140803193-140803193 on Build GRCh38
rs781969076 Chr.5: 140803157-140803157 on Build GRCh38
rs374629500 Chr.5: 140802825-140802825 on Build GRCh38
rs782235300 Chr.5: 140802951-140802951 on Build GRCh38
rs991420459 Chr.5: 140802816-140802816 on Build GRCh38
rs781982895 Chr.5: 140802850-140802850 on Build GRCh38
rs782709763 Chr.5: 140803034-140803034 on Build GRCh38
rs782761119 Chr.5: 140802968-140802968 on Build GRCh38
rs374457025 Chr.5: 140803063-140803063 on Build GRCh38
rs782083603 Chr.5: 140803160-140803160 on Build GRCh38
rs782050127 Chr.5: 140803082-140803082 on Build GRCh38
rs145535421 Chr.5: 140802920-140802920 on Build GRCh38
rs915930472 Chr.5: 140802869-140802869 on Build GRCh38

View Full Product Details