Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38351590-38352099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4462882 c.1265C>T p.P422L Chr.15: 38351594-38351594 on Build GRCh38
7100456 c.1314G>C p.G438G Chr.15: 38351643-38351643 on Build GRCh38
2268639 c.1282C>T p.R428C Chr.15: 38351611-38351611 on Build GRCh38
346424 c.1304G>T p.C435F Chr.15: 38351633-38351633 on Build GRCh38
6210416 c.1331G>A p.G444E Chr.15: 38351660-38351660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982818134 Chr.15: 38351875-38351875 on Build GRCh38
rs555317480 Chr.15: 38352055-38352055 on Build GRCh38
rs891077662 Chr.15: 38351685-38351685 on Build GRCh38
rs994432500 Chr.15: 38351945-38351945 on Build GRCh38
rs1052988828 Chr.15: 38351853-38351853 on Build GRCh38
rs1025945380 Chr.15: 38351739-38351739 on Build GRCh38
rs908220961 Chr.15: 38351911-38351911 on Build GRCh38
rs776485386 Chr.15: 38351813-38351813 on Build GRCh38
rs932465161 Chr.15: 38352018-38352018 on Build GRCh38
rs779454951 Chr.15: 38351627-38351627 on Build GRCh38
rs1029867136 Chr.15: 38351957-38351957 on Build GRCh38
rs761401264 Chr.15: 38351653-38351653 on Build GRCh38
rs751117768 Chr.15: 38352024-38352024 on Build GRCh38
rs1044533508 Chr.15: 38351886-38351886 on Build GRCh38
rs547722924 Chr.15: 38351914-38351914 on Build GRCh38
rs145373657 Chr.15: 38351920-38351920 on Build GRCh38
rs746414913 Chr.15: 38351630-38351630 on Build GRCh38
rs567404158 Chr.15: 38351869-38351869 on Build GRCh38
rs1006156390 Chr.15: 38351913-38351913 on Build GRCh38
rs569990101 Chr.15: 38352022-38352022 on Build GRCh38
rs530541798 Chr.15: 38351847-38351847 on Build GRCh38
rs769445247 Chr.15: 38351666-38351666 on Build GRCh38
rs772421738 Chr.15: 38351636-38351636 on Build GRCh38
rs1009278466 Chr.15: 38351750-38351750 on Build GRCh38
rs954413934 Chr.15: 38352010-38352010 on Build GRCh38
rs544137820 Chr.15: 38351728-38351728 on Build GRCh38
rs533114849 Chr.15: 38351730-38351730 on Build GRCh38
rs545295358 Chr.15: 38351769-38351769 on Build GRCh38
rs749389617 Chr.15: 38351618-38351618 on Build GRCh38
rs539607759 Chr.15: 38351929-38351929 on Build GRCh38
rs759838497 Chr.15: 38351648-38351649 on Build GRCh38
rs377201217 Chr.15: 38351890-38351890 on Build GRCh38
rs373694127 Chr.15: 38351712-38351712 on Build GRCh38
rs765093939 Chr.15: 38351837-38351837 on Build GRCh38
rs1020286096 Chr.15: 38351758-38351758 on Build GRCh38
rs770816015 Chr.15: 38351807-38351807 on Build GRCh38
rs954339455 Chr.15: 38351764-38351764 on Build GRCh38
rs377091058 Chr.15: 38351854-38351854 on Build GRCh38
rs910296382 Chr.15: 38351784-38351784 on Build GRCh38
rs773453342 Chr.15: 38351745-38351745 on Build GRCh38
rs772254530 Chr.15: 38351697-38351698 on Build GRCh38
rs943227853 Chr.15: 38351795-38351795 on Build GRCh38
rs192826011 Chr.15: 38351832-38351832 on Build GRCh38
rs13329392 Chr.15: 38352046-38352046 on Build GRCh38
rs537079236 Chr.15: 38352023-38352023 on Build GRCh38
rs1033084809 Chr.15: 38351812-38351812 on Build GRCh38
rs16966842 Chr.15: 38351912-38351912 on Build GRCh38
rs934606444 Chr.15: 38351829-38351829 on Build GRCh38
rs369492789 Chr.15: 38351612-38351612 on Build GRCh38
rs573603750 Chr.15: 38351642-38351642 on Build GRCh38
rs772560499 Chr.15: 38351711-38351711 on Build GRCh38
rs200871227 Chr.15: 38351624-38351624 on Build GRCh38
rs973368328 Chr.15: 38351694-38351694 on Build GRCh38
rs137920898 Chr.15: 38351985-38351985 on Build GRCh38

View Full Product Details