Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173325770-173326278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STC2
Entrez Gene ID
8614
Gene Name
stanniocalcin 2
Gene Aliases
STC-2, STCRP
UniGene
Hs.233160
Gene Chromosome Location
Chr.5: 173314723-173329503 on Build GRCh38
Transcripts
NM_003714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1292094 c.249G>A p.G83G Chr.5: 173325913-173325913 on Build GRCh38
3853772 c.272A>G p.N91S Chr.5: 173325890-173325890 on Build GRCh38
3339877 c.184G>A p.D62N Chr.5: 173325978-173325978 on Build GRCh38
3339878 c.178G>A p.A60T Chr.5: 173325984-173325984 on Build GRCh38
3339881 c.152-8C>T p.? Chr.5: 173326018-173326018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145449570 Chr.5: 173326009-173326009 on Build GRCh38
rs201103640 Chr.5: 173325985-173325985 on Build GRCh38
rs113396561 Chr.5: 173325828-173325828 on Build GRCh38
rs758852559 Chr.5: 173326026-173326026 on Build GRCh38
rs1037098980 Chr.5: 173326239-173326239 on Build GRCh38
rs780297853 Chr.5: 173325944-173325944 on Build GRCh38
rs1018037294 Chr.5: 173325811-173325811 on Build GRCh38
rs773075267 Chr.5: 173325894-173325894 on Build GRCh38
rs777050582 Chr.5: 173325991-173325991 on Build GRCh38
rs375809330 Chr.5: 173325822-173325822 on Build GRCh38
rs781768761 Chr.5: 173326237-173326237 on Build GRCh38
rs780867329 Chr.5: 173325955-173325955 on Build GRCh38
rs753936878 Chr.5: 173325913-173325913 on Build GRCh38
rs758725089 Chr.5: 173325927-173325927 on Build GRCh38
rs377701281 Chr.5: 173326055-173326055 on Build GRCh38
rs762800064 Chr.5: 173325897-173325897 on Build GRCh38
rs773130884 Chr.5: 173325978-173325978 on Build GRCh38
rs13361431 Chr.5: 173326235-173326235 on Build GRCh38
rs373327101 Chr.5: 173325871-173325871 on Build GRCh38
rs559928257 Chr.5: 173326094-173326094 on Build GRCh38
rs542236985 Chr.5: 173326157-173326157 on Build GRCh38
rs146162200 Chr.5: 173326176-173326176 on Build GRCh38
rs984784775 Chr.5: 173326218-173326218 on Build GRCh38
rs780812376 Chr.5: 173326051-173326051 on Build GRCh38
rs916188948 Chr.5: 173325824-173325824 on Build GRCh38
rs970324869 Chr.5: 173326215-173326215 on Build GRCh38
rs748039896 Chr.5: 173325889-173325889 on Build GRCh38
rs1037532480 Chr.5: 173325945-173325945 on Build GRCh38
rs751893130 Chr.5: 173326047-173326047 on Build GRCh38
rs916156340 Chr.5: 173326179-173326179 on Build GRCh38
rs1031857642 Chr.5: 173326171-173326171 on Build GRCh38
rs937324656 Chr.5: 173326234-173326234 on Build GRCh38
rs750959444 Chr.5: 173325926-173325926 on Build GRCh38
rs779752047 Chr.5: 173325854-173325854 on Build GRCh38
rs779222160 Chr.5: 173325934-173325934 on Build GRCh38
rs1024272834 Chr.5: 173325919-173325919 on Build GRCh38
rs754772317 Chr.5: 173326005-173326006 on Build GRCh38
rs199859079 Chr.5: 173326010-173326010 on Build GRCh38
rs746670391 Chr.5: 173325856-173325856 on Build GRCh38
rs765762557 Chr.5: 173325925-173325925 on Build GRCh38
rs62636573 Chr.5: 173326007-173326007 on Build GRCh38
rs755589523 Chr.5: 173326057-173326057 on Build GRCh38
rs968797147 Chr.5: 173326116-173326116 on Build GRCh38
rs770708060 Chr.5: 173325984-173325984 on Build GRCh38
rs1057463631 Chr.5: 173326235-173326235 on Build GRCh38
rs371382926 Chr.5: 173325812-173325812 on Build GRCh38
rs10076089 Chr.5: 173326086-173326086 on Build GRCh38
rs147198518 Chr.5: 173325949-173325949 on Build GRCh38
rs146441603 Chr.5: 173325904-173325904 on Build GRCh38
rs758748245 Chr.5: 173325832-173325832 on Build GRCh38
rs143058338 Chr.5: 173325964-173325964 on Build GRCh38
rs988026115 Chr.5: 173325915-173325915 on Build GRCh38
rs909852008 Chr.5: 173326233-173326233 on Build GRCh38
rs369369541 Chr.5: 173325823-173325823 on Build GRCh38
rs36042025 Chr.5: 173325901-173325902 on Build GRCh38
rs766088063 Chr.5: 173325902-173325902 on Build GRCh38
rs779025975 Chr.5: 173326013-173326013 on Build GRCh38
rs374560214 Chr.5: 173325988-173325988 on Build GRCh38
rs376762243 Chr.5: 173325948-173325948 on Build GRCh38
rs960180024 Chr.5: 173326175-173326175 on Build GRCh38
rs766693155 Chr.5: 173326042-173326042 on Build GRCh38
rs755389339 Chr.5: 173326050-173326050 on Build GRCh38
rs998585486 Chr.5: 173326118-173326118 on Build GRCh38
rs937403328 Chr.5: 173325973-173325973 on Build GRCh38
rs994300199 Chr.5: 173326003-173326003 on Build GRCh38
rs74328847 Chr.5: 173326113-173326113 on Build GRCh38
rs768098012 Chr.5: 173325857-173325857 on Build GRCh38
rs764227728 Chr.5: 173325905-173325905 on Build GRCh38
rs114346665 Chr.5: 173325979-173325979 on Build GRCh38
rs758598823 Chr.5: 173326017-173326017 on Build GRCh38
rs775794210 Chr.5: 173325903-173325903 on Build GRCh38
rs978806988 Chr.5: 173326227-173326227 on Build GRCh38
rs915169431 Chr.5: 173326182-173326186 on Build GRCh38
rs370833041 Chr.5: 173325963-173325963 on Build GRCh38
rs753736356 Chr.5: 173326003-173326004 on Build GRCh38
rs757329205 Chr.5: 173325917-173325917 on Build GRCh38

View Full Product Details