Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 71683429-71683960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
UniGene
Hs.494146
Gene Chromosome Location
Chr.9: 71683366-71769532 on Build GRCh38
Transcripts
NM_001135820.1 NM_013390.2 XM_005251869.4

SNP Detail

SNP ID Chromosome Location
rs147500253 Chr.9: 71683847-71683847 on Build GRCh38
rs7872063 Chr.9: 71683553-71683553 on Build GRCh38
rs1054457568 Chr.9: 71683921-71683921 on Build GRCh38
rs13296471 Chr.9: 71683506-71683506 on Build GRCh38
rs968255626 Chr.9: 71683834-71683834 on Build GRCh38
rs946137229 Chr.9: 71683940-71683940 on Build GRCh38
rs10683269 Chr.9: 71683631-71683632 on Build GRCh38
rs966262405 Chr.9: 71683503-71683503 on Build GRCh38
rs951727777 Chr.9: 71683529-71683529 on Build GRCh38
rs34812277 Chr.9: 71683619-71683620 on Build GRCh38
rs13295855 Chr.9: 71683535-71683535 on Build GRCh38
rs947936375 Chr.9: 71683691-71683691 on Build GRCh38
rs73646776 Chr.9: 71683632-71683632 on Build GRCh38
rs540882646 Chr.9: 71683920-71683920 on Build GRCh38
rs28521499 Chr.9: 71683541-71683541 on Build GRCh38
rs551304042 Chr.9: 71683628-71683629 on Build GRCh38
rs769668612 Chr.9: 71683552-71683552 on Build GRCh38
rs183467551 Chr.9: 71683904-71683904 on Build GRCh38
rs7854914 Chr.9: 71683546-71683546 on Build GRCh38
rs763330415 Chr.9: 71683521-71683522 on Build GRCh38
rs573929059 Chr.9: 71683519-71683520 on Build GRCh38
rs368721910 Chr.9: 71683619-71683619 on Build GRCh38
rs1019563389 Chr.9: 71683453-71683453 on Build GRCh38
rs575841937 Chr.9: 71683937-71683937 on Build GRCh38
rs962488921 Chr.9: 71683547-71683547 on Build GRCh38
rs59888879 Chr.9: 71683551-71683552 on Build GRCh38
rs376007736 Chr.9: 71683902-71683902 on Build GRCh38
rs1015039419 Chr.9: 71683869-71683869 on Build GRCh38
rs749059190 Chr.9: 71683554-71683554 on Build GRCh38
rs375077098 Chr.9: 71683520-71683521 on Build GRCh38
rs372441179 Chr.9: 71683872-71683872 on Build GRCh38
rs905835817 Chr.9: 71683710-71683710 on Build GRCh38
rs902048042 Chr.9: 71683824-71683824 on Build GRCh38
rs997359537 Chr.9: 71683778-71683778 on Build GRCh38
rs542380300 Chr.9: 71683901-71683901 on Build GRCh38
rs761477519 Chr.9: 71683830-71683830 on Build GRCh38
rs1054912423 Chr.9: 71683792-71683792 on Build GRCh38
rs917795928 Chr.9: 71683638-71683638 on Build GRCh38
rs980661913 Chr.9: 71683908-71683908 on Build GRCh38
rs10657343 Chr.9: 71683626-71683627 on Build GRCh38
rs753981877 Chr.9: 71683632-71683632 on Build GRCh38
rs3208833 Chr.9: 71683575-71683575 on Build GRCh38
rs73646777 Chr.9: 71683794-71683794 on Build GRCh38
rs535325453 Chr.9: 71683855-71683855 on Build GRCh38
rs974987605 Chr.9: 71683511-71683511 on Build GRCh38
rs909108901 Chr.9: 71683523-71683523 on Build GRCh38
rs192920734 Chr.9: 71683870-71683870 on Build GRCh38
rs570167193 Chr.9: 71683742-71683742 on Build GRCh38
rs1019091780 Chr.9: 71683815-71683815 on Build GRCh38
rs202089728 Chr.9: 71683519-71683519 on Build GRCh38
rs936144845 Chr.9: 71683897-71683897 on Build GRCh38
rs947686512 Chr.9: 71683498-71683498 on Build GRCh38
rs572163979 Chr.9: 71683857-71683857 on Build GRCh38
rs187858354 Chr.9: 71683709-71683709 on Build GRCh38
rs984923797 Chr.9: 71683543-71683543 on Build GRCh38
rs891231300 Chr.9: 71683799-71683799 on Build GRCh38
rs6560226 Chr.9: 71683804-71683804 on Build GRCh38
rs34237025 Chr.9: 71683549-71683549 on Build GRCh38
rs961819208 Chr.9: 71683892-71683892 on Build GRCh38
rs1006722636 Chr.9: 71683544-71683544 on Build GRCh38
rs536383342 Chr.9: 71683661-71683661 on Build GRCh38
rs35210493 Chr.9: 71683549-71683550 on Build GRCh38
rs147197801 Chr.9: 71683553-71683554 on Build GRCh38
rs925194897 Chr.9: 71683918-71683918 on Build GRCh38
rs992303546 Chr.9: 71683894-71683894 on Build GRCh38

View Full Product Details