Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62678229-62678724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBXN1
Entrez Gene ID
51035
Gene Name
UBX domain protein 1
Gene Aliases
2B28, SAKS1, UBXD10
UniGene
Hs.351296
Gene Chromosome Location
Chr.11: 62676498-62679117 on Build GRCh38
Transcripts
NM_001286077.1 NM_001286078.1 NM_015853.4 XM_005274033.4 XM_011545090.2 XM_017017874.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4198984 c.230C>T p.S77F Chr.11: 62678399-62678399 on Build GRCh38
6069855 c.145G>T p.D49Y Chr.11: 62678570-62678570 on Build GRCh38
6069857 c.217G>C p.E73Q Chr.11: 62678498-62678498 on Build GRCh38
7429469 c.196T>G p.S66A Chr.11: 62678519-62678519 on Build GRCh38
6069851 c.97G>A p.E33K Chr.11: 62678706-62678706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777385951 Chr.11: 62678557-62678557 on Build GRCh38
rs761124667 Chr.11: 62678271-62678271 on Build GRCh38
rs899246157 Chr.11: 62678335-62678335 on Build GRCh38
rs767857290 Chr.11: 62678622-62678622 on Build GRCh38
rs1045730176 Chr.11: 62678580-62678580 on Build GRCh38
rs768924326 Chr.11: 62678260-62678260 on Build GRCh38
rs760422382 Chr.11: 62678448-62678448 on Build GRCh38
rs748564352 Chr.11: 62678523-62678523 on Build GRCh38
rs763648423 Chr.11: 62678352-62678352 on Build GRCh38
rs940697283 Chr.11: 62678318-62678318 on Build GRCh38
rs758064181 Chr.11: 62678288-62678288 on Build GRCh38
rs761022908 Chr.11: 62678696-62678696 on Build GRCh38
rs769025417 Chr.11: 62678553-62678553 on Build GRCh38
rs367762053 Chr.11: 62678537-62678537 on Build GRCh38
rs200509247 Chr.11: 62678452-62678452 on Build GRCh38
rs780840991 Chr.11: 62678646-62678646 on Build GRCh38
rs773974516 Chr.11: 62678355-62678355 on Build GRCh38
rs776196357 Chr.11: 62678578-62678578 on Build GRCh38
rs573473975 Chr.11: 62678291-62678291 on Build GRCh38
rs754123655 Chr.11: 62678630-62678630 on Build GRCh38
rs746425748 Chr.11: 62678423-62678423 on Build GRCh38
rs745568868 Chr.11: 62678648-62678648 on Build GRCh38
rs768652978 Chr.11: 62678661-62678661 on Build GRCh38
rs753215567 Chr.11: 62678406-62678406 on Build GRCh38
rs763843203 Chr.11: 62678281-62678281 on Build GRCh38
rs373658424 Chr.11: 62678531-62678531 on Build GRCh38
rs374673900 Chr.11: 62678482-62678482 on Build GRCh38
rs553738145 Chr.11: 62678287-62678287 on Build GRCh38
rs770391851 Chr.11: 62678425-62678425 on Build GRCh38
rs199667995 Chr.11: 62678518-62678518 on Build GRCh38
rs761083702 Chr.11: 62678336-62678336 on Build GRCh38
rs762986383 Chr.11: 62678609-62678609 on Build GRCh38
rs766902589 Chr.11: 62678273-62678273 on Build GRCh38
rs756075011 Chr.11: 62678299-62678299 on Build GRCh38
rs201705756 Chr.11: 62678679-62678679 on Build GRCh38
rs776082556 Chr.11: 62678497-62678497 on Build GRCh38
rs746471196 Chr.11: 62678315-62678316 on Build GRCh38
rs762490673 Chr.11: 62678280-62678280 on Build GRCh38
rs753739263 Chr.11: 62678384-62678384 on Build GRCh38
rs771574845 Chr.11: 62678504-62678504 on Build GRCh38
rs777128896 Chr.11: 62678344-62678344 on Build GRCh38
rs763933578 Chr.11: 62678610-62678610 on Build GRCh38
rs749021415 Chr.11: 62678658-62678658 on Build GRCh38
rs752586581 Chr.11: 62678521-62678521 on Build GRCh38
rs766507871 Chr.11: 62678699-62678699 on Build GRCh38
rs766235589 Chr.11: 62678377-62678377 on Build GRCh38
rs370802907 Chr.11: 62678334-62678334 on Build GRCh38
rs772765104 Chr.11: 62678505-62678505 on Build GRCh38
rs1015956881 Chr.11: 62678303-62678303 on Build GRCh38
rs773240253 Chr.11: 62678420-62678423 on Build GRCh38
rs771351553 Chr.11: 62678341-62678341 on Build GRCh38
rs748323768 Chr.11: 62678326-62678326 on Build GRCh38
rs754951734 Chr.11: 62678453-62678453 on Build GRCh38
rs765143859 Chr.11: 62678457-62678457 on Build GRCh38
rs775420938 Chr.11: 62678657-62678657 on Build GRCh38
rs2276297 Chr.11: 62678437-62678437 on Build GRCh38
rs183444513 Chr.11: 62678436-62678436 on Build GRCh38
rs777299895 Chr.11: 62678408-62678408 on Build GRCh38
rs370333215 Chr.11: 62678502-62678502 on Build GRCh38
rs759353634 Chr.11: 62678581-62678581 on Build GRCh38
rs933823877 Chr.11: 62678585-62678585 on Build GRCh38
rs755578162 Chr.11: 62678631-62678631 on Build GRCh38
rs1047441794 Chr.11: 62678278-62678278 on Build GRCh38
rs755076465 Chr.11: 62678391-62678391 on Build GRCh38
rs766152223 Chr.11: 62678450-62678450 on Build GRCh38
rs772388946 Chr.11: 62678266-62678266 on Build GRCh38
rs749366736 Chr.11: 62678259-62678259 on Build GRCh38
rs746231930 Chr.11: 62678486-62678486 on Build GRCh38
rs769404807 Chr.11: 62678649-62678649 on Build GRCh38
rs369515395 Chr.11: 62678645-62678645 on Build GRCh38
rs140112798 Chr.11: 62678342-62678342 on Build GRCh38
rs368461519 Chr.11: 62678625-62678625 on Build GRCh38
rs976362958 Chr.11: 62678306-62678306 on Build GRCh38
rs557002078 Chr.11: 62678411-62678414 on Build GRCh38
rs750203711 Chr.11: 62678295-62678295 on Build GRCh38
rs758434170 Chr.11: 62678472-62678472 on Build GRCh38
rs143908470 Chr.11: 62678362-62678362 on Build GRCh38
rs370914438 Chr.11: 62678617-62678617 on Build GRCh38
rs775124680 Chr.11: 62678600-62678600 on Build GRCh38
rs550723710 Chr.11: 62678596-62678596 on Build GRCh38
rs746530793 Chr.11: 62678637-62678637 on Build GRCh38
rs757139007 Chr.11: 62678289-62678289 on Build GRCh38
rs115234949 Chr.11: 62678265-62678265 on Build GRCh38
rs746499157 Chr.11: 62678565-62678565 on Build GRCh38
rs777152521 Chr.11: 62678302-62678302 on Build GRCh38
rs751721689 Chr.11: 62678613-62678613 on Build GRCh38
rs200124299 Chr.11: 62678623-62678623 on Build GRCh38
rs777991221 Chr.11: 62678403-62678403 on Build GRCh38
rs776465200 Chr.11: 62678511-62678511 on Build GRCh38
rs749384873 Chr.11: 62678305-62678305 on Build GRCh38
rs770482012 Chr.11: 62678493-62678493 on Build GRCh38
rs749758621 Chr.11: 62678552-62678552 on Build GRCh38
rs752203326 Chr.11: 62678276-62678276 on Build GRCh38
rs145840266 Chr.11: 62678538-62678538 on Build GRCh38
rs762582295 Chr.11: 62678346-62678346 on Build GRCh38
rs774074053 Chr.11: 62678426-62678426 on Build GRCh38
rs200272023 Chr.11: 62678517-62678517 on Build GRCh38
rs780029540 Chr.11: 62678302-62678302 on Build GRCh38
rs201490859 Chr.11: 62678399-62678399 on Build GRCh38
rs201175971 Chr.11: 62678574-62678574 on Build GRCh38
rs771286702 Chr.11: 62678407-62678407 on Build GRCh38
rs764283196 Chr.11: 62678528-62678528 on Build GRCh38
rs757396813 Chr.11: 62678475-62678475 on Build GRCh38
rs202070522 Chr.11: 62678539-62678539 on Build GRCh38
rs192772658 Chr.11: 62678476-62678476 on Build GRCh38
rs926118532 Chr.11: 62678432-62678432 on Build GRCh38
rs754290527 Chr.11: 62678704-62678704 on Build GRCh38
rs756602937 Chr.11: 62678331-62678332 on Build GRCh38
rs985208179 Chr.11: 62678420-62678420 on Build GRCh38
rs780738842 Chr.11: 62678550-62678550 on Build GRCh38
rs575585285 Chr.11: 62678392-62678392 on Build GRCh38
rs978829425 Chr.11: 62678469-62678469 on Build GRCh38
rs779227862 Chr.11: 62678636-62678636 on Build GRCh38
rs11543359 Chr.11: 62678549-62678549 on Build GRCh38
rs772318723 Chr.11: 62678330-62678330 on Build GRCh38
rs747203915 Chr.11: 62678474-62678474 on Build GRCh38
rs774400270 Chr.11: 62678368-62678368 on Build GRCh38
rs112860804 Chr.11: 62678315-62678315 on Build GRCh38
rs767196813 Chr.11: 62678364-62678364 on Build GRCh38
rs762014718 Chr.11: 62678673-62678673 on Build GRCh38
rs373339711 Chr.11: 62678376-62678376 on Build GRCh38
rs774091437 Chr.11: 62678669-62678669 on Build GRCh38
rs773793618 Chr.11: 62678269-62678269 on Build GRCh38
rs779141529 Chr.11: 62678324-62678324 on Build GRCh38
rs770541744 Chr.11: 62678506-62678506 on Build GRCh38
rs767656506 Chr.11: 62678674-62678674 on Build GRCh38
rs886404873 Chr.11: 62678279-62678279 on Build GRCh38
rs372123032 Chr.11: 62678626-62678626 on Build GRCh38

View Full Product Details