Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65108730-65109236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS51
Entrez Gene ID
738
Gene Name
VPS51, GARP complex subunit
Gene Aliases
ANG2, ANG3, C11orf2, C11orf3, FFR
UniGene
Hs.277517
Gene Chromosome Location
Chr.11: 65096115-65111860 on Build GRCh38
Transcripts
NM_013265.3 NR_073519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1194312 c.1300G>A p.V434M Chr.11: 65108771-65108771 on Build GRCh38
690059 c.1406C>T p.A469V Chr.11: 65108877-65108877 on Build GRCh38
1676171 c.1441C>T p.R481W Chr.11: 65108912-65108912 on Build GRCh38
5624198 c.1404C>T p.T468T Chr.11: 65108875-65108875 on Build GRCh38
3810011 c.1396C>A p.L466I Chr.11: 65108867-65108867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772539666 Chr.11: 65108782-65108782 on Build GRCh38
rs758822492 Chr.11: 65108887-65108887 on Build GRCh38
rs772952505 Chr.11: 65108930-65108930 on Build GRCh38
rs1012897726 Chr.11: 65109128-65109128 on Build GRCh38
rs527307869 Chr.11: 65108847-65108847 on Build GRCh38
rs912258308 Chr.11: 65108978-65108978 on Build GRCh38
rs144845424 Chr.11: 65108827-65108827 on Build GRCh38
rs563771562 Chr.11: 65108913-65108913 on Build GRCh38
rs760670592 Chr.11: 65108934-65108934 on Build GRCh38
rs963653761 Chr.11: 65108988-65108988 on Build GRCh38
rs749867580 Chr.11: 65108754-65108754 on Build GRCh38
rs758592314 Chr.11: 65108936-65108936 on Build GRCh38
rs760096968 Chr.11: 65108839-65108839 on Build GRCh38
rs1057056916 Chr.11: 65109107-65109107 on Build GRCh38
rs1041977202 Chr.11: 65109048-65109048 on Build GRCh38
rs899209650 Chr.11: 65109194-65109194 on Build GRCh38
rs752415010 Chr.11: 65108962-65108962 on Build GRCh38
rs1037791296 Chr.11: 65109147-65109147 on Build GRCh38
rs914286624 Chr.11: 65109038-65109038 on Build GRCh38
rs777607408 Chr.11: 65108912-65108912 on Build GRCh38
rs752863502 Chr.11: 65108890-65108892 on Build GRCh38
rs753040548 Chr.11: 65108880-65108880 on Build GRCh38
rs140186079 Chr.11: 65108835-65108835 on Build GRCh38
rs143862116 Chr.11: 65108911-65108911 on Build GRCh38
rs537983133 Chr.11: 65109160-65109160 on Build GRCh38
rs758697422 Chr.11: 65108964-65108964 on Build GRCh38
rs567558898 Chr.11: 65108949-65108949 on Build GRCh38
rs776178886 Chr.11: 65108950-65108950 on Build GRCh38
rs577686094 Chr.11: 65109208-65109208 on Build GRCh38
rs753590445 Chr.11: 65108756-65108756 on Build GRCh38
rs759098381 Chr.11: 65108951-65108951 on Build GRCh38
rs201089305 Chr.11: 65108841-65108841 on Build GRCh38
rs747556945 Chr.11: 65108895-65108895 on Build GRCh38
rs774317852 Chr.11: 65108822-65108822 on Build GRCh38
rs373780734 Chr.11: 65108875-65108875 on Build GRCh38
rs771110751 Chr.11: 65108798-65108798 on Build GRCh38
rs377512003 Chr.11: 65108929-65108929 on Build GRCh38
rs761738641 Chr.11: 65108830-65108830 on Build GRCh38
rs946541516 Chr.11: 65108968-65108968 on Build GRCh38
rs947249949 Chr.11: 65109043-65109043 on Build GRCh38
rs778303315 Chr.11: 65108787-65108787 on Build GRCh38
rs768521247 Chr.11: 65108814-65108814 on Build GRCh38
rs747940507 Chr.11: 65108919-65108919 on Build GRCh38
rs556367817 Chr.11: 65109102-65109102 on Build GRCh38
rs754660795 Chr.11: 65108865-65108865 on Build GRCh38
rs548997589 Chr.11: 65108941-65108941 on Build GRCh38
rs755264716 Chr.11: 65108761-65108761 on Build GRCh38
rs778140195 Chr.11: 65108890-65108890 on Build GRCh38
rs201622389 Chr.11: 65108770-65108770 on Build GRCh38
rs987723898 Chr.11: 65108945-65108945 on Build GRCh38
rs894493155 Chr.11: 65109124-65109124 on Build GRCh38
rs750430648 Chr.11: 65108837-65108837 on Build GRCh38
rs747048701 Chr.11: 65108792-65108792 on Build GRCh38
rs536145001 Chr.11: 65109045-65109045 on Build GRCh38
rs118099554 Chr.11: 65109146-65109146 on Build GRCh38
rs932109005 Chr.11: 65108936-65108936 on Build GRCh38
rs149391431 Chr.11: 65108800-65108800 on Build GRCh38
rs924464764 Chr.11: 65109063-65109063 on Build GRCh38
rs1042291138 Chr.11: 65109011-65109011 on Build GRCh38
rs755696138 Chr.11: 65108755-65108755 on Build GRCh38
rs372162170 Chr.11: 65108958-65108958 on Build GRCh38
rs756774498 Chr.11: 65109031-65109031 on Build GRCh38
rs557845230 Chr.11: 65109181-65109181 on Build GRCh38
rs770859008 Chr.11: 65108908-65108908 on Build GRCh38
rs780720381 Chr.11: 65108931-65108931 on Build GRCh38
rs72932836 Chr.11: 65108918-65108918 on Build GRCh38

View Full Product Details