Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15719388-15719883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF286A
Entrez Gene ID
57335
Gene Name
zinc finger protein 286A
Gene Aliases
ZNF286
UniGene
Hs.585799
Gene Chromosome Location
Chr.17: 15699577-15720787 on Build GRCh38
Transcripts
NM_001130842.1 NM_001288642.1 NM_001288643.1 NM_001288644.1 NM_001288645.1 NM_001288646.1 NM_001288647.1 NM_001288648.1 NM_001288649.1 NM_020652.2

SNP Detail

SNP ID Chromosome Location
rs8079347 Chr.17: 15719757-15719757 on Build GRCh38
rs551411910 Chr.17: 15719551-15719551 on Build GRCh38
rs565735837 Chr.17: 15719675-15719675 on Build GRCh38
rs141138905 Chr.17: 15719643-15719643 on Build GRCh38
rs536988737 Chr.17: 15719836-15719836 on Build GRCh38
rs918187614 Chr.17: 15719559-15719559 on Build GRCh38
rs567841005 Chr.17: 15719797-15719797 on Build GRCh38
rs2530058 Chr.17: 15719623-15719623 on Build GRCh38
rs2530059 Chr.17: 15719591-15719591 on Build GRCh38
rs949605903 Chr.17: 15719572-15719572 on Build GRCh38
rs965294471 Chr.17: 15719655-15719655 on Build GRCh38
rs188012109 Chr.17: 15719644-15719644 on Build GRCh38
rs576289626 Chr.17: 15719807-15719807 on Build GRCh38
rs936349437 Chr.17: 15719808-15719808 on Build GRCh38
rs879198854 Chr.17: 15719465-15719465 on Build GRCh38
rs1026353380 Chr.17: 15719691-15719691 on Build GRCh38
rs377321175 Chr.17: 15719803-15719803 on Build GRCh38
rs985273327 Chr.17: 15719454-15719454 on Build GRCh38
rs950778949 Chr.17: 15719696-15719696 on Build GRCh38
rs959418479 Chr.17: 15719758-15719758 on Build GRCh38
rs976082960 Chr.17: 15719424-15719424 on Build GRCh38
rs542476653 Chr.17: 15719802-15719802 on Build GRCh38
rs991007537 Chr.17: 15719770-15719770 on Build GRCh38
rs527423831 Chr.17: 15719614-15719614 on Build GRCh38
rs570281628 Chr.17: 15719666-15719666 on Build GRCh38
rs534606472 Chr.17: 15719445-15719446 on Build GRCh38
rs373188421 Chr.17: 15719667-15719667 on Build GRCh38
rs554654341 Chr.17: 15719728-15719728 on Build GRCh38
rs556298793 Chr.17: 15719804-15719804 on Build GRCh38
rs376484531 Chr.17: 15719420-15719420 on Build GRCh38
rs182491423 Chr.17: 15719637-15719637 on Build GRCh38
rs200361865 Chr.17: 15719810-15719811 on Build GRCh38
rs558572278 Chr.17: 15719674-15719674 on Build GRCh38
rs889556370 Chr.17: 15719815-15719815 on Build GRCh38
rs1040890477 Chr.17: 15719823-15719823 on Build GRCh38
rs918074901 Chr.17: 15719791-15719791 on Build GRCh38
rs1006583648 Chr.17: 15719823-15719823 on Build GRCh38
rs192365189 Chr.17: 15719725-15719725 on Build GRCh38
rs909266264 Chr.17: 15719527-15719527 on Build GRCh38
rs938164947 Chr.17: 15719533-15719533 on Build GRCh38
rs545047837 Chr.17: 15719562-15719562 on Build GRCh38
rs919233547 Chr.17: 15719437-15719437 on Build GRCh38
rs993975287 Chr.17: 15719848-15719848 on Build GRCh38
rs186317070 Chr.17: 15719540-15719540 on Build GRCh38

View Full Product Details