Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 118209193-118209690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC2
Entrez Gene ID
255082
Gene Name
cancer susceptibility candidate 2 (non-protein coding)
Gene Aliases
C10orf5
UniGene
Hs.89387
Gene Chromosome Location
Chr.10: 118046821-118210153 on Build GRCh38
Transcripts
NR_026939.1 NR_026940.1

SNP Detail

SNP ID Chromosome Location
rs1023198761 Chr.10: 118209413-118209413 on Build GRCh38
rs142097910 Chr.10: 118209622-118209622 on Build GRCh38
rs1014497678 Chr.10: 118209411-118209411 on Build GRCh38
rs933243379 Chr.10: 118209247-118209247 on Build GRCh38
rs747952394 Chr.10: 118209448-118209448 on Build GRCh38
rs562488931 Chr.10: 118209426-118209426 on Build GRCh38
rs1008354677 Chr.10: 118209253-118209253 on Build GRCh38
rs115633161 Chr.10: 118209656-118209656 on Build GRCh38
rs181034168 Chr.10: 118209455-118209455 on Build GRCh38
rs1043081517 Chr.10: 118209535-118209535 on Build GRCh38
rs746248477 Chr.10: 118209366-118209366 on Build GRCh38
rs1037198221 Chr.10: 118209506-118209506 on Build GRCh38
rs573839629 Chr.10: 118209355-118209355 on Build GRCh38
rs375129311 Chr.10: 118209587-118209590 on Build GRCh38
rs564601823 Chr.10: 118209481-118209481 on Build GRCh38
rs12252936 Chr.10: 118209663-118209663 on Build GRCh38
rs770378093 Chr.10: 118209638-118209638 on Build GRCh38
rs34884433 Chr.10: 118209284-118209284 on Build GRCh38
rs575609236 Chr.10: 118209440-118209440 on Build GRCh38
rs758631922 Chr.10: 118209386-118209386 on Build GRCh38
rs954726111 Chr.10: 118209494-118209494 on Build GRCh38
rs904614770 Chr.10: 118209377-118209377 on Build GRCh38
rs746049587 Chr.10: 118209304-118209304 on Build GRCh38
rs1015926271 Chr.10: 118209450-118209450 on Build GRCh38
rs896006603 Chr.10: 118209396-118209396 on Build GRCh38
rs1051708624 Chr.10: 118209282-118209282 on Build GRCh38
rs58917327 Chr.10: 118209331-118209331 on Build GRCh38
rs897396958 Chr.10: 118209323-118209323 on Build GRCh38
rs746978427 Chr.10: 118209508-118209508 on Build GRCh38
rs60396676 Chr.10: 118209492-118209492 on Build GRCh38
rs370171761 Chr.10: 118209596-118209599 on Build GRCh38
rs367699429 Chr.10: 118209630-118209630 on Build GRCh38
rs551454472 Chr.10: 118209394-118209394 on Build GRCh38
rs771976615 Chr.10: 118209449-118209449 on Build GRCh38
rs186371480 Chr.10: 118209532-118209532 on Build GRCh38
rs573381281 Chr.10: 118209333-118209334 on Build GRCh38
rs960149941 Chr.10: 118209378-118209378 on Build GRCh38
rs969824702 Chr.10: 118209342-118209342 on Build GRCh38
rs1001162997 Chr.10: 118209625-118209625 on Build GRCh38
rs925986445 Chr.10: 118209576-118209576 on Build GRCh38
rs778765952 Chr.10: 118209372-118209372 on Build GRCh38
rs189236274 Chr.10: 118209387-118209387 on Build GRCh38
rs963155060 Chr.10: 118209451-118209451 on Build GRCh38
rs910483943 Chr.10: 118209530-118209530 on Build GRCh38
rs1037697581 Chr.10: 118209269-118209269 on Build GRCh38
rs778961902 Chr.10: 118209332-118209332 on Build GRCh38

View Full Product Details