Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23301540-23302080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R3E
Entrez Gene ID
90673
Gene Name
protein phosphatase 1 regulatory subunit 3E
Gene Aliases
-
UniGene
Hs.601513
Gene Chromosome Location
Chr.14: 23295921-23302848 on Build GRCh38
Transcripts
NM_001276318.1

SNP Detail

SNP ID Chromosome Location
rs940898034 Chr.14: 23301793-23301793 on Build GRCh38
rs781433173 Chr.14: 23302011-23302011 on Build GRCh38
rs977433648 Chr.14: 23301691-23301691 on Build GRCh38
rs546199819 Chr.14: 23301699-23301699 on Build GRCh38
rs982423632 Chr.14: 23301977-23301977 on Build GRCh38
rs943610446 Chr.14: 23301570-23301570 on Build GRCh38
rs908323519 Chr.14: 23301922-23301922 on Build GRCh38
rs990021167 Chr.14: 23301579-23301579 on Build GRCh38
rs10130210 Chr.14: 23301946-23301946 on Build GRCh38
rs894278358 Chr.14: 23301792-23301792 on Build GRCh38
rs902130436 Chr.14: 23301795-23301795 on Build GRCh38
rs115298373 Chr.14: 23301986-23301986 on Build GRCh38
rs1047281978 Chr.14: 23301595-23301595 on Build GRCh38
rs886841327 Chr.14: 23301598-23301598 on Build GRCh38
rs1046711792 Chr.14: 23301735-23301735 on Build GRCh38
rs915654036 Chr.14: 23301765-23301765 on Build GRCh38
rs775685361 Chr.14: 23301829-23301829 on Build GRCh38
rs540190699 Chr.14: 23301585-23301585 on Build GRCh38
rs527781300 Chr.14: 23301823-23301823 on Build GRCh38
rs887996425 Chr.14: 23301628-23301628 on Build GRCh38
rs777212182 Chr.14: 23301851-23301851 on Build GRCh38
rs1002701120 Chr.14: 23301600-23301600 on Build GRCh38
rs754862629 Chr.14: 23301589-23301589 on Build GRCh38
rs1042890879 Chr.14: 23302016-23302016 on Build GRCh38
rs903005677 Chr.14: 23301836-23301836 on Build GRCh38
rs747599770 Chr.14: 23301841-23301841 on Build GRCh38
rs943636116 Chr.14: 23301718-23301718 on Build GRCh38
rs761998334 Chr.14: 23301858-23301858 on Build GRCh38
rs752724791 Chr.14: 23301626-23301626 on Build GRCh38
rs910688548 Chr.14: 23301714-23301714 on Build GRCh38
rs983820671 Chr.14: 23301879-23301879 on Build GRCh38
rs933307037 Chr.14: 23301797-23301797 on Build GRCh38
rs901826715 Chr.14: 23301664-23301664 on Build GRCh38
rs763057072 Chr.14: 23301907-23301907 on Build GRCh38
rs568209030 Chr.14: 23301788-23301788 on Build GRCh38
rs1012847322 Chr.14: 23301809-23301809 on Build GRCh38
rs548411535 Chr.14: 23301762-23301762 on Build GRCh38
rs984900548 Chr.14: 23301785-23301785 on Build GRCh38
rs1021645099 Chr.14: 23301672-23301672 on Build GRCh38
rs1049252904 Chr.14: 23301617-23301617 on Build GRCh38
rs969092574 Chr.14: 23301688-23301688 on Build GRCh38
rs773637435 Chr.14: 23301893-23301893 on Build GRCh38
rs960018346 Chr.14: 23301845-23301845 on Build GRCh38
rs538036764 Chr.14: 23302003-23302003 on Build GRCh38
rs987512675 Chr.14: 23301884-23301884 on Build GRCh38
rs778548239 Chr.14: 23301769-23301769 on Build GRCh38
rs1050424011 Chr.14: 23301807-23301807 on Build GRCh38
rs1011379363 Chr.14: 23301701-23301701 on Build GRCh38
rs560201471 Chr.14: 23301668-23301668 on Build GRCh38
rs769074563 Chr.14: 23301842-23301842 on Build GRCh38
rs959958489 Chr.14: 23301743-23301743 on Build GRCh38
rs1035643248 Chr.14: 23301742-23301742 on Build GRCh38
rs1012504043 Chr.14: 23301865-23301865 on Build GRCh38
rs1039379000 Chr.14: 23301571-23301571 on Build GRCh38
rs1013212404 Chr.14: 23301636-23301636 on Build GRCh38
rs920844183 Chr.14: 23301727-23301727 on Build GRCh38
rs1021468688 Chr.14: 23301720-23301720 on Build GRCh38
rs965276781 Chr.14: 23301916-23301916 on Build GRCh38
rs1002545893 Chr.14: 23301775-23301775 on Build GRCh38
rs958625014 Chr.14: 23301631-23301631 on Build GRCh38
rs938454522 Chr.14: 23301989-23301989 on Build GRCh38
rs915686642 Chr.14: 23301605-23301605 on Build GRCh38
rs765396697 Chr.14: 23301877-23301877 on Build GRCh38
rs987671437 Chr.14: 23301710-23301710 on Build GRCh38
rs920884510 Chr.14: 23301973-23301973 on Build GRCh38
rs767215827 Chr.14: 23301616-23301616 on Build GRCh38
rs948221658 Chr.14: 23301613-23301613 on Build GRCh38
rs554807099 Chr.14: 23302059-23302059 on Build GRCh38
rs1021987977 Chr.14: 23301813-23301813 on Build GRCh38
rs940957499 Chr.14: 23301630-23301630 on Build GRCh38
rs930997612 Chr.14: 23301730-23301730 on Build GRCh38
rs910740934 Chr.14: 23301891-23301891 on Build GRCh38
rs369376909 Chr.14: 23301744-23301744 on Build GRCh38
rs569176208 Chr.14: 23301990-23301990 on Build GRCh38
rs547344365 Chr.14: 23301840-23301840 on Build GRCh38
rs570780508 Chr.14: 23301904-23301904 on Build GRCh38
rs779089690 Chr.14: 23302021-23302021 on Build GRCh38
rs998737962 Chr.14: 23302058-23302058 on Build GRCh38
rs756687089 Chr.14: 23301766-23301766 on Build GRCh38
rs995281248 Chr.14: 23301592-23301592 on Build GRCh38
rs745491936 Chr.14: 23301816-23301816 on Build GRCh38
rs929567100 Chr.14: 23301974-23301974 on Build GRCh38
rs532805281 Chr.14: 23301681-23301681 on Build GRCh38
rs924710711 Chr.14: 23301698-23301698 on Build GRCh38
rs962519350 Chr.14: 23301927-23301927 on Build GRCh38
rs563224661 Chr.14: 23301724-23301724 on Build GRCh38

View Full Product Details