Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58291583-58292117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMA3-AS1
Entrez Gene ID
379025
Gene Name
PSMA3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.531089
Gene Chromosome Location
Chr.14: 58265365-58298137 on Build GRCh38
Transcripts
NR_029434.1 NR_029435.1

SNP Detail

SNP ID Chromosome Location
rs994423684 Chr.14: 58291763-58291763 on Build GRCh38
rs397739100 Chr.14: 58291614-58291615 on Build GRCh38
rs914843898 Chr.14: 58291692-58291692 on Build GRCh38
rs185690039 Chr.14: 58292086-58292086 on Build GRCh38
rs865859038 Chr.14: 58291818-58291818 on Build GRCh38
rs939007183 Chr.14: 58291854-58291854 on Build GRCh38
rs577574749 Chr.14: 58291920-58291920 on Build GRCh38
rs894881314 Chr.14: 58291931-58291931 on Build GRCh38
rs533968786 Chr.14: 58291723-58291723 on Build GRCh38
rs1042454482 Chr.14: 58291754-58291754 on Build GRCh38
rs950608410 Chr.14: 58291786-58291786 on Build GRCh38
rs751648447 Chr.14: 58291976-58291976 on Build GRCh38
rs536877620 Chr.14: 58291688-58291688 on Build GRCh38
rs75445767 Chr.14: 58291790-58291790 on Build GRCh38
rs567019887 Chr.14: 58291702-58291702 on Build GRCh38
rs755103289 Chr.14: 58292001-58292001 on Build GRCh38
rs932165691 Chr.14: 58291656-58291656 on Build GRCh38
rs79702010 Chr.14: 58291843-58291843 on Build GRCh38
rs1051349134 Chr.14: 58292074-58292074 on Build GRCh38
rs763522961 Chr.14: 58291905-58291905 on Build GRCh38
rs988780344 Chr.14: 58291918-58291918 on Build GRCh38
rs980630127 Chr.14: 58291855-58291855 on Build GRCh38
rs926027929 Chr.14: 58291862-58291863 on Build GRCh38
rs1053376530 Chr.14: 58291682-58291682 on Build GRCh38
rs556091398 Chr.14: 58291916-58291916 on Build GRCh38
rs934253261 Chr.14: 58291954-58291954 on Build GRCh38
rs925987929 Chr.14: 58291643-58291643 on Build GRCh38
rs1027417493 Chr.14: 58291959-58291959 on Build GRCh38
rs11461913 Chr.14: 58291841-58291842 on Build GRCh38
rs193016764 Chr.14: 58292002-58292002 on Build GRCh38
rs545040470 Chr.14: 58291972-58291972 on Build GRCh38
rs766864636 Chr.14: 58291974-58291974 on Build GRCh38
rs747633258 Chr.14: 58291823-58291823 on Build GRCh38
rs969916096 Chr.14: 58291843-58291843 on Build GRCh38
rs1038773319 Chr.14: 58291758-58291758 on Build GRCh38
rs1004766159 Chr.14: 58292031-58292031 on Build GRCh38
rs537773302 Chr.14: 58291893-58291893 on Build GRCh38
rs773535639 Chr.14: 58291757-58291757 on Build GRCh38
rs1026207510 Chr.14: 58291781-58291781 on Build GRCh38
rs199556603 Chr.14: 58291615-58291615 on Build GRCh38
rs765719250 Chr.14: 58291745-58291745 on Build GRCh38
rs188714702 Chr.14: 58291781-58291781 on Build GRCh38
rs545259192 Chr.14: 58291869-58291869 on Build GRCh38
rs935982030 Chr.14: 58291646-58291646 on Build GRCh38
rs553446368 Chr.14: 58292027-58292027 on Build GRCh38
rs11461912 Chr.14: 58291613-58291614 on Build GRCh38
rs182931221 Chr.14: 58291647-58291647 on Build GRCh38
rs898842641 Chr.14: 58291762-58291762 on Build GRCh38
rs386364092 Chr.14: 58291842-58291843 on Build GRCh38
rs1012054593 Chr.14: 58291955-58291955 on Build GRCh38
rs551911065 Chr.14: 58291698-58291698 on Build GRCh38
rs573962774 Chr.14: 58291829-58291829 on Build GRCh38
rs773906324 Chr.14: 58291727-58291727 on Build GRCh38

View Full Product Details