Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 66491526-66492027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00238
Entrez Gene ID
440184
Gene Name
long intergenic non-protein coding RNA 238
Gene Aliases
C14orf53, NCRNA00238
UniGene
Hs.412138
Gene Chromosome Location
Chr.14: 66486371-66498553 on Build GRCh38
Transcripts
NR_024338.2 NR_110307.1 NR_110308.1

SNP Detail

SNP ID Chromosome Location
rs527457081 Chr.14: 66491998-66491998 on Build GRCh38
rs976902974 Chr.14: 66491605-66491605 on Build GRCh38
rs938884493 Chr.14: 66491564-66491564 on Build GRCh38
rs773502166 Chr.14: 66491913-66491913 on Build GRCh38
rs983004554 Chr.14: 66491684-66491684 on Build GRCh38
rs918291591 Chr.14: 66491969-66491969 on Build GRCh38
rs578102215 Chr.14: 66491676-66491676 on Build GRCh38
rs892189566 Chr.14: 66491864-66491864 on Build GRCh38
rs960341262 Chr.14: 66491915-66491915 on Build GRCh38
rs1006698057 Chr.14: 66491686-66491686 on Build GRCh38
rs71422423 Chr.14: 66491930-66491930 on Build GRCh38
rs779144248 Chr.14: 66491799-66491799 on Build GRCh38
rs553804209 Chr.14: 66491790-66491790 on Build GRCh38
rs1036727439 Chr.14: 66491594-66491594 on Build GRCh38
rs1457419 Chr.14: 66491888-66491888 on Build GRCh38
rs938989504 Chr.14: 66491818-66491818 on Build GRCh38
rs1000364766 Chr.14: 66491945-66491945 on Build GRCh38
rs775488122 Chr.14: 66491980-66491980 on Build GRCh38
rs930978025 Chr.14: 66491997-66491997 on Build GRCh38
rs1048628926 Chr.14: 66491658-66491658 on Build GRCh38
rs151184618 Chr.14: 66491729-66491729 on Build GRCh38
rs759676456 Chr.14: 66491919-66491919 on Build GRCh38
rs201971949 Chr.14: 66491655-66491662 on Build GRCh38
rs140212943 Chr.14: 66491761-66491761 on Build GRCh38
rs542579088 Chr.14: 66491835-66491835 on Build GRCh38
rs896793844 Chr.14: 66491609-66491609 on Build GRCh38
rs1019379696 Chr.14: 66491716-66491716 on Build GRCh38
rs1457420 Chr.14: 66491958-66491958 on Build GRCh38
rs980279231 Chr.14: 66491906-66491906 on Build GRCh38
rs765082664 Chr.14: 66491787-66491787 on Build GRCh38
rs1018196874 Chr.14: 66491574-66491574 on Build GRCh38
rs182923969 Chr.14: 66491627-66491627 on Build GRCh38
rs964048875 Chr.14: 66491991-66491991 on Build GRCh38
rs931114760 Chr.14: 66491640-66491640 on Build GRCh38
rs538394067 Chr.14: 66491975-66491975 on Build GRCh38
rs370828816 Chr.14: 66491576-66491576 on Build GRCh38
rs999831574 Chr.14: 66491793-66491793 on Build GRCh38
rs558376785 Chr.14: 66491662-66491662 on Build GRCh38
rs187218877 Chr.14: 66491810-66491810 on Build GRCh38
rs909820941 Chr.14: 66491743-66491743 on Build GRCh38
rs889496201 Chr.14: 66491664-66491664 on Build GRCh38
rs1036656631 Chr.14: 66491646-66491646 on Build GRCh38
rs1011542747 Chr.14: 66491836-66491836 on Build GRCh38
rs113458528 Chr.14: 66491889-66491889 on Build GRCh38

View Full Product Details