Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176150197-176150707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD3
Entrez Gene ID
3232
Gene Name
homeobox D3
Gene Aliases
HOX1D, HOX4, HOX4A, Hox-4.1
UniGene
Hs.93574
Gene Chromosome Location
Chr.2: 176136921-176173102 on Build GRCh38
Transcripts
XM_005246510.4 XM_006712477.2 XM_005246514.4 NR_029609.1

Gene Symbol
HOXD4
Entrez Gene ID
3233
Gene Name
homeobox D4
Gene Aliases
HHO.C13, HOX-5.1, HOX4, HOX4B, Hox-4.2
UniGene
Hs.591609
Gene Chromosome Location
Chr.2: 176150394-176156078 on Build GRCh38
Transcripts
XM_005246510.4 XM_006712477.2 XM_005246514.4 NR_029609.1

Gene Symbol
MIR10B
Entrez Gene ID
406903
Gene Name
microRNA 10b
Gene Aliases
MIRN10B, hsa-mir-10b, miRNA10B, mir-10b
UniGene
Gene Chromosome Location
Chr.2: 176150303-176150412 on Build GRCh38
Transcripts
XM_005246510.4 XM_006712477.2 XM_005246514.4 NR_029609.1

SNP Detail

SNP ID Chromosome Location
rs776938328 Chr.2: 176150410-176150410 on Build GRCh38
rs750914364 Chr.2: 176150308-176150308 on Build GRCh38
rs758869665 Chr.2: 176150310-176150310 on Build GRCh38
rs77628401 Chr.2: 176150300-176150300 on Build GRCh38
rs113754306 Chr.2: 176150261-176150261 on Build GRCh38
rs946819560 Chr.2: 176150502-176150502 on Build GRCh38
rs1018785492 Chr.2: 176150221-176150221 on Build GRCh38
rs971963283 Chr.2: 176150632-176150632 on Build GRCh38
rs773513030 Chr.2: 176150446-176150447 on Build GRCh38
rs761097016 Chr.2: 176150223-176150223 on Build GRCh38
rs760930932 Chr.2: 176150269-176150269 on Build GRCh38
rs919068530 Chr.2: 176150639-176150639 on Build GRCh38
rs982387279 Chr.2: 176150399-176150399 on Build GRCh38
rs138423463 Chr.2: 176150514-176150514 on Build GRCh38
rs765856096 Chr.2: 176150302-176150302 on Build GRCh38
rs967661659 Chr.2: 176150485-176150485 on Build GRCh38
rs1022620853 Chr.2: 176150312-176150312 on Build GRCh38
rs946842787 Chr.2: 176150662-176150662 on Build GRCh38
rs746695011 Chr.2: 176150314-176150314 on Build GRCh38
rs1020863140 Chr.2: 176150466-176150466 on Build GRCh38
rs760873052 Chr.2: 176150384-176150384 on Build GRCh38
rs200015643 Chr.2: 176150411-176150411 on Build GRCh38
rs1014433772 Chr.2: 176150438-176150438 on Build GRCh38
rs769243214 Chr.2: 176150337-176150337 on Build GRCh38
rs993260962 Chr.2: 176150241-176150241 on Build GRCh38
rs1036564604 Chr.2: 176150271-176150271 on Build GRCh38
rs1023096373 Chr.2: 176150625-176150628 on Build GRCh38
rs751011495 Chr.2: 176150417-176150417 on Build GRCh38
rs1034795682 Chr.2: 176150414-176150414 on Build GRCh38
rs751955022 Chr.2: 176150444-176150444 on Build GRCh38
rs774478768 Chr.2: 176150626-176150627 on Build GRCh38
rs774183104 Chr.2: 176150381-176150381 on Build GRCh38
rs764138704 Chr.2: 176150270-176150270 on Build GRCh38
rs141593566 Chr.2: 176150625-176150626 on Build GRCh38
rs747667744 Chr.2: 176150332-176150332 on Build GRCh38
rs34537138 Chr.2: 176150486-176150487 on Build GRCh38
rs753964284 Chr.2: 176150286-176150286 on Build GRCh38
rs552375106 Chr.2: 176150265-176150265 on Build GRCh38
rs553803111 Chr.2: 176150621-176150621 on Build GRCh38
rs770679233 Chr.2: 176150369-176150369 on Build GRCh38
rs879174632 Chr.2: 176150285-176150285 on Build GRCh38
rs930592555 Chr.2: 176150642-176150642 on Build GRCh38
rs766158071 Chr.2: 176150254-176150254 on Build GRCh38
rs535486989 Chr.2: 176150560-176150560 on Build GRCh38
rs556450863 Chr.2: 176150481-176150481 on Build GRCh38
rs760678943 Chr.2: 176150523-176150523 on Build GRCh38
rs1048980200 Chr.2: 176150654-176150654 on Build GRCh38
rs749191671 Chr.2: 176150359-176150359 on Build GRCh38
rs1035301654 Chr.2: 176150640-176150640 on Build GRCh38
rs866116872 Chr.2: 176150264-176150264 on Build GRCh38
rs765372245 Chr.2: 176150413-176150413 on Build GRCh38
rs868047830 Chr.2: 176150682-176150682 on Build GRCh38
rs897667260 Chr.2: 176150238-176150238 on Build GRCh38
rs896186738 Chr.2: 176150433-176150433 on Build GRCh38
rs764383388 Chr.2: 176150394-176150394 on Build GRCh38
rs915352596 Chr.2: 176150486-176150486 on Build GRCh38
rs755236598 Chr.2: 176150460-176150460 on Build GRCh38
rs986237086 Chr.2: 176150666-176150666 on Build GRCh38
rs1033039568 Chr.2: 176150509-176150509 on Build GRCh38
rs1056501222 Chr.2: 176150665-176150665 on Build GRCh38
rs1000430812 Chr.2: 176150499-176150499 on Build GRCh38
rs1027440319 Chr.2: 176150624-176150625 on Build GRCh38
rs926518133 Chr.2: 176150658-176150658 on Build GRCh38
rs770929781 Chr.2: 176150517-176150517 on Build GRCh38
rs564940769 Chr.2: 176150278-176150278 on Build GRCh38
rs780439437 Chr.2: 176150311-176150311 on Build GRCh38
rs897732456 Chr.2: 176150613-176150616 on Build GRCh38
rs930561308 Chr.2: 176150329-176150329 on Build GRCh38
rs937682103 Chr.2: 176150659-176150659 on Build GRCh38
rs1867863 Chr.2: 176150242-176150242 on Build GRCh38
rs780639575 Chr.2: 176150324-176150324 on Build GRCh38
rs1040279948 Chr.2: 176150599-176150602 on Build GRCh38
rs538164178 Chr.2: 176150426-176150426 on Build GRCh38
rs570521775 Chr.2: 176150317-176150317 on Build GRCh38
rs757340886 Chr.2: 176150295-176150295 on Build GRCh38
rs911981606 Chr.2: 176150573-176150573 on Build GRCh38
rs754106408 Chr.2: 176150354-176150354 on Build GRCh38
rs766716253 Chr.2: 176150435-176150435 on Build GRCh38

View Full Product Details