Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 55656886-55657431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERC2
Entrez Gene ID
26059
Gene Name
ELKS/RAB6-interacting/CAST family member 2
Gene Aliases
CAST, CAST1, ELKSL, SPBC110, Spc110
UniGene
Hs.476389
Gene Chromosome Location
Chr.3: 55508311-56468473 on Build GRCh38
Transcripts
NM_015576.2 NR_132749.1 XM_017006138.1 XM_017006139.1 XM_017006140.1 XM_017006141.1 XM_017006142.1 XM_017006143.1 XM_017006144.1 XM_017006145.1 XM_017006146.1 XM_017006147.1 XM_017006148.1 XM_017006149.1 XM_017006150.1 XM_017006151.1 XM_017006152.1 XM_017006153.1 XM_017006154.1 XM_017006155.1 XM_017006156.1 XM_017006157.1 XM_017006158.1 XM_017006159.1 NR_024615.1

Gene Symbol
ERC2-IT1
Entrez Gene ID
711
Gene Name
ERC2 intronic transcript 1
Gene Aliases
C1orf1, C3orf51, Po42
UniGene
Hs.667161
Gene Chromosome Location
Chr.3: 55657206-55659469 on Build GRCh38
Transcripts
NM_015576.2 NR_132749.1 XM_017006138.1 XM_017006139.1 XM_017006140.1 XM_017006141.1 XM_017006142.1 XM_017006143.1 XM_017006144.1 XM_017006145.1 XM_017006146.1 XM_017006147.1 XM_017006148.1 XM_017006149.1 XM_017006150.1 XM_017006151.1 XM_017006152.1 XM_017006153.1 XM_017006154.1 XM_017006155.1 XM_017006156.1 XM_017006157.1 XM_017006158.1 XM_017006159.1 NR_024615.1

SNP Detail

SNP ID Chromosome Location
rs936721758 Chr.3: 55656922-55656922 on Build GRCh38
rs577371980 Chr.3: 55657163-55657163 on Build GRCh38
rs1033523399 Chr.3: 55657191-55657191 on Build GRCh38
rs796105249 Chr.3: 55657336-55657336 on Build GRCh38
rs562760802 Chr.3: 55657260-55657260 on Build GRCh38
rs901553954 Chr.3: 55657079-55657079 on Build GRCh38
rs374168679 Chr.3: 55657028-55657028 on Build GRCh38
rs894949269 Chr.3: 55657209-55657209 on Build GRCh38
rs928954491 Chr.3: 55657150-55657150 on Build GRCh38
rs557770268 Chr.3: 55657094-55657094 on Build GRCh38
rs191367774 Chr.3: 55657164-55657164 on Build GRCh38
rs918825071 Chr.3: 55657363-55657363 on Build GRCh38
rs958149728 Chr.3: 55657287-55657287 on Build GRCh38
rs1008494626 Chr.3: 55657080-55657080 on Build GRCh38
rs966725268 Chr.3: 55657263-55657263 on Build GRCh38
rs1013544048 Chr.3: 55657216-55657216 on Build GRCh38
rs1001268606 Chr.3: 55656954-55656954 on Build GRCh38
rs556806793 Chr.3: 55657224-55657224 on Build GRCh38
rs376589881 Chr.3: 55657112-55657112 on Build GRCh38
rs776897661 Chr.3: 55657281-55657281 on Build GRCh38
rs577644361 Chr.3: 55657108-55657108 on Build GRCh38
rs116135987 Chr.3: 55657365-55657365 on Build GRCh38
rs951303128 Chr.3: 55657075-55657075 on Build GRCh38
rs995096709 Chr.3: 55657060-55657060 on Build GRCh38
rs3732751 Chr.3: 55656961-55656961 on Build GRCh38
rs944301542 Chr.3: 55657065-55657065 on Build GRCh38
rs538557796 Chr.3: 55657177-55657177 on Build GRCh38
rs139684373 Chr.3: 55657252-55657252 on Build GRCh38
rs976993782 Chr.3: 55657359-55657359 on Build GRCh38
rs530103796 Chr.3: 55657145-55657146 on Build GRCh38
rs978105936 Chr.3: 55657266-55657266 on Build GRCh38
rs185895649 Chr.3: 55657151-55657151 on Build GRCh38
rs73069877 Chr.3: 55657369-55657369 on Build GRCh38
rs985914618 Chr.3: 55657298-55657298 on Build GRCh38
rs760157223 Chr.3: 55657146-55657146 on Build GRCh38
rs911340397 Chr.3: 55657047-55657047 on Build GRCh38
rs555596418 Chr.3: 55656935-55656935 on Build GRCh38
rs552746517 Chr.3: 55657158-55657158 on Build GRCh38
rs1019465581 Chr.3: 55657105-55657105 on Build GRCh38
rs763387525 Chr.3: 55657273-55657273 on Build GRCh38
rs182494474 Chr.3: 55657304-55657304 on Build GRCh38
rs944205582 Chr.3: 55657332-55657332 on Build GRCh38
rs534859716 Chr.3: 55657070-55657070 on Build GRCh38
rs573833143 Chr.3: 55657231-55657231 on Build GRCh38
rs1055473686 Chr.3: 55657305-55657305 on Build GRCh38
rs761222323 Chr.3: 55657205-55657206 on Build GRCh38

View Full Product Details